Ion AmpliSeq™ HD Dual Barcode Kit 1-24
Ion Torrent™

Ion AmpliSeq™ HD Dual Barcode Kit 1-24

The Ion AmpliSeq HD Dual Barcode Kit 1-24 provides a set of 24 unique dual-barcode primers designed and validated for optimal performance with Ion GeneStudio S5 series sequencers.
Have Questions?
Catalog NumberQuantity
A3769524 Reactions
Catalog number A37695
Price (INR)
30,194.00
Each
Quantity:
24 Reactions
Price (INR)
30,194.00
Each

The Ion AmpliSeq HD Dual Barcode Kit 1-24 provides a set of 24 unique dual-barcode primers designed and validated for optimal performance with Ion GeneStudio S5 series sequencers. This barcode kit enables up to 24 samples to be multiplexed on a single chip. The barcode kit is only compatible with the Ion AmpliSeq HD Library Kit (Cat. No. A37694) and should not be used with any other library kit.

Features of the Ion AmpliSeq HD Dual Barcode Kit 1-24 include:

  • Enables multiplexing of up to 24 library samples on a single sequencing chip
  • Dual barcoding at both the 5' and 3' end for enhanced sample tracking
  • Barcodes have been sequence- and flow-optimized for equal representation of all barcodes within a pool
  • Provided in a single-use 96-well plate for ease of use

Sample multiplexing

Sample multiplexing using the Ion AmpliSeq HD Dual Barcode Kit 1-24 allows for multiple samples to be run on the same chip, thus decreasing the amount of reagent used on a per sample basis.

Dual-barcoding

Use of dual-barcodes has been shown to provide greater confidence in sample tracking and enhances the ability to detect any cross-barcode events. The barcodes are added at the 5' and 3' ends as part of the library preparation protocol.

Sequence- and flow-optimized

The Ion AmpliSeq HD Dual Barcode Kit 1-24 contains primers with unique sequences that have been optimized to provide equal representation of all barcodes in a pool, with a minimum number of flows. This helps achieve economical multiplexed sequencing runs.

24 single-use dual-barcodes in a 96-well plate format

The dual barcodes come in a sealed 96-well plate. Spacing of the barcodes prevents cross-barcode mixing and enables easy access to individual barcodes. A single reaction is provided in each well, which prevents the contamination challenges faced when using multi-use tubes.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)Ion GeneStudio™ S5 Series Sequencer, Ion Chef™ System
LibrariesTargeted Sequencing Library
No. of Reactions24
Product LineAmpliSeq
Product TypeHD Dual Barcode Kit
Quantity24 Reactions
Sample TypeDNA
Shipping ConditionRoom Temperature
Workflow StepLibrary Barcoding
FormatKit
Sequencing TypeGenome and DNA Sequencing
Unit SizeEach
Contents & Storage
One 96-well plate containing dual barcodes

Store at -5°C to -30°C.

Frequently asked questions (FAQs)

Can the 96-well plate in the Ion AmpliSeq HD Dual Barcode Kit 1-24 (Cat. No. A37695) be freeze/thawed multiple times?

We have taken the 96-well plate in the Ion AmpliSeq HD Dual Barcode Kit 1-24 through several freeze-thaw cycles with little to no reduction in performance.
Note: Sealing the plate between uses is key when using the plate for multiple experiments.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

What version of Torrent Suite Software do I need in order to run the Ion AmpliSeq HD Dual Barcode Kit 1-24?

The Ion AmpliSeq HD Dual Barcode Kit 1-24 is compatible with Torrent Suite Software version 5.10 and above.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

Is the Ion AmpliSeq HD Dual Barcode Kit 1-24 shipped on ice or at room temperature?

The kit is shipped at room temperature, but should be stored at -20 degrees C.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

How many samples is one Ion AmpliSeq HD Dual Barcode Kit 1-24 good for?

The Ion AmpliSeq HD Dual Barcode Kit 1-24 is good for 24 samples. Each barcode is good for one sample.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

Is the Ion AmpliSeq HD Dual Barcode Kit 1-24 compatible with the Ion Chef System?

No, it is only compatible with manual sample preparation.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.