Experience nuclei counting with fluorescent-mode counting algorithms, simplified workflows, built-in dilution calculators and reports using the latest version of Countess software with optional 21 CFR Part 11 compliance. Download now!
Countess™ Reusable Slide and Holder, 1 holder
Invitrogen™
Countess™ Reusable Slide and Holder, 1 holder
Countess Cell Counting Chamber Slides are conveniently packaged for use with the Countess Automated Cell Counters. Each disposable and reusable slide features two enclosed chambers.
Have Questions?
Catalog number A48208
Price (INR)
5,012.00
Online offer
Ends: 31-Dec-2026
7,159.00
Save 2,147.00 (30%)
Each
Price (INR)
5,012.00
Online offer
Ends: 31-Dec-2026
7,159.00
Save 2,147.00 (30%)
Each
The Countess Chamber Slides are designed for use with the Countess 3 and 3FL automated cell counters.
The disposable Countess Cell Counting Chamber Slides offer convenience in automated cell counting for processing multiple cell samples, busy cell culture facilities, and easy disposal of contaminated or hazardous samples. They also eliminate the risk of contamination from residual samples. After use, Countess slides can be disposed of as standard biohazard waste.
The Countess Reusable Slide is a green and cost-efficient alternative to disposable slides for automated cell counting. The reusable slide enables researchers and cell culture facilities to reduce costs and generate less waste per year, making it affordable for labs to automate their cell counts. The reusable slide is cleaned and dried between counts, just as with a hemocytometer.
Each Countess Cell Counting Chamber Slide has two separate, enclosed chambers for counting one sample in duplicate or two different samples. Cell counting occurs in a central location of the counting chamber. The volume counted is 0.4 μL, the same as counting four 1-mm2 squares in a standard hemocytometer.
The Countess Automated Cell Counter works in three simple steps:
  • Mix 10 μL of sample with 10 μL of a cell viability stain or other desired reagent, including ReadyCount™ stains, and pipet into a Countess chamber slide
  • Insert the slide into the instrument
  • Press 'Count'
That's it—get the total cell count, percent viable cells, and average cell size in as little as 30 seconds.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
CompatibilityCountess™ Reusable Slide (Cat. No. A5750)
For Use With (Equipment)Countess II FL, Countess 3 & Countess 3 FL Automated Cell Counters
Quantity1 Holder
Product LineCountess
TypeSlide Holder
Unit SizeEach
Contents & Storage
Countess™ 3 and 3 FL Reusable Slide Holder, 1 each
Frequently asked questions (FAQs)
Do you offer reusable slides and the appropriate holders for Countess II FL and Countess 3/3 FL instruments?
The Countess Reusable Slide (Cat. No. A25750) is compatible with the Countess II FL and the Countess 3/3 FL instruments. However, they require a reusable slide holder specific to the instrument. The Countess 3 & 3 FL Reusable Slide Holder (Cat. No. A48208) and the Countess II FL Reusable Slide Holder (Cat No. AMEP4746) are shipped with the instrument and can be purchased separately.
The slide holder can be held inside the back of the instrument, next to the light cube removal tool.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Where may I purchase the coverslips provided with the Countess Reusable Slide (Cat no. A25750) as a stand-alone product?
Hausser Scientific Cover Glasses/Slips (Cat. No. 02-617-581) can be purchased through Fisher Scientific; these are 20 x 26 x 0.5 mm borosilicate glass coverslips.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Can the Countess 3 & 3 FL Reusable Slide Holder (Cat no. A48208) be used on a Countess II FL Automated Cell Counter (Cat. No. AMQAF1000)?
Yes. The Countess 3 & 3 FL Reusable Slide Holder (Cat. No. A48208) be used on a Countess II FL Automated Cell Counter (Cat. No. AMQAF1000).
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Can the Countess II FL Reusable Slide Holder (Cat no. AMEP4746) be used on a Countess 3 and 3 FL Automated Cell Counter (Cat. Nos. AMQAX2000 and AMQAF2000)?
Yes. The Countess II FL Reusable Slide Holder (Cat no. AMEP4746) can be used on a Countess 3 and 3 FL Automated Cell Counter (Cat. Nos. AMQAX2000 and AMQAF2000).
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Can a conventional cell counting chamber be used with the Countess 3 & 3 FL Reusable Slide Holder (Cat. No. A48208)?
No. The Countess 3 & 3 FL Reusable Slide Holder (Cat. No. A48208) and the Countess instruments will only work with the Countess Reusable Slide (Cat. No. A25750). Using other slides can damage the instrument, due to differences in the height and width of the slides.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.