TrueMark™ Respiratory Panel 2.0 PreAmp Primers
Applied Biosystems™
TrueMark™ Respiratory Panel 2.0 PreAmp Primers
The TrueMark Respiratory Panel 2.0 PreAmp Primers kit contains primers for all respiratory tract microbiota assays plus the TaqMan UniversalRead more
Have Questions?
Catalog NumberQuantity
A490491 Kit
Catalog number A49049
Price (INR)
54,314.00
Each
Quantity:
1 Kit
Price (INR)
54,314.00
Each
The TrueMark Respiratory Panel 2.0 PreAmp Primers kit contains primers for all respiratory tract microbiota assays plus the TaqMan Universal RNA Spike In/Reverse Transcription (Xeno) Control and the RNase P RPPH1 gene, but does not contain primers for the TaqMan Universal Extraction Control Organism (B. atrophaeus).
The primers are intended to be used in the preamplification step of the TrueMark Respiratory Panel 2.0, OpenArray plate and TrueMark Respiratory Panel 2.0, TaqMan array card workflows to help achieve the highest analytical sensitivity with the added benefit of sample conservation.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use WithTrueMark™ Respiratory Panel 2.0, TaqMan™ Array Card, TrueMark™ Respiratory Panel 2.0, OpenArray™ Plate
Product LineTrueMark
Quantity1 Kit
Shipping ConditionDry Ice
Detection MethodPrimer-probe
Unit SizeEach
Contents & Storage
Store at -30°C to -5°C