Tubes, strips of 8, red
Thermo Scientific™

Tubes, strips of 8, red

The Thermo Scientific 0.2 mL Strip Tubes are designed for PCR and qPCR applications, and are compatible with 0.2 mLRead more
Have Questions?
Catalog NumberQuantity
AB0264R250 strips
Catalog number AB0264R
Price (INR)
21,914.00
Online offer
Ends: 31-Dec-2026
36,523.00
Save 14,609.00 (40%)
Each
Quantity:
250 strips
Request bulk or custom format
Price (INR)
21,914.00
Online offer
Ends: 31-Dec-2026
36,523.00
Save 14,609.00 (40%)
Each
The Thermo Scientific 0.2 mL Strip Tubes are designed for PCR and qPCR applications, and are compatible with 0.2 mL thermal cycler blocks.While these tubes are sold without caps, Thermo Scientific offers multiple cap options, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL

Features of The Thermo Scientific 0.2 mL Strip Tubes, red

• Compatible with 0.2 mL thermal cycler blocks
• 8 tube strips
• Colored tubes are ideal for keeping track of samples
• Sealable using flat and domed cap strips. Ultra-Clear cap options available for use in qPCR assays

Applications

• PCR
• qPCR/Real-Time PCR

Related Products
Tubes, strips of 8
Tubes, strips of 8, white
Tubes, strips of 8, blue
Tubes, strips of 8, green
Tubes, strips of 8, purple
Tubes, strips of 8, yellow
Tubes, strips of 12
Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Equipment)Thermal Cyclers
Format250 Strip Tubes
Quantity250 strips
Reaction SpeedStandard
Volume (Metric) Working0.2 mL
Closure TypeNot Included
ColorRed
MaterialPolypropylene
Product Type8-strip Tube
SterilityNon-sterile
Volume (Metric)0.25 mL
Unit SizeEach