A more sustainable equivalent is available.
EasyStrip™ Plus Tube Strip with Attached Flat Caps
Thermo Scientific™

EasyStrip™ Plus Tube Strip with Attached Flat Caps

The Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Flat Caps is designed to reduce the risk of cross-contamination inRead more
Have Questions?
Catalog NumberQuantity
AB2000250 strips
Catalog number AB2000
Price (INR)
11,576.00
온라인 행사
Ends: 31-Dec-2026
16,537.00
Save 4,961.00 (30%)
Each
Quantity:
250 strips
Request bulk or custom format
Price (INR)
11,576.00
온라인 행사
Ends: 31-Dec-2026
16,537.00
Save 4,961.00 (30%)
Each
The Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Flat Caps is designed to reduce the risk of cross-contamination in PCR assays. The tube strip is compatible with 0.2 mL thermal cycler blocks and features individually attached flat caps for quick and easy access to individual samples. In addition, graduated 20 μL markers on each tube enable visual volume verification and measurement of reagent and sample additions.

For tube strips with ultra clear caps, see Cat. No. AB-2005.

Features:
• Avoid cross contamination with the individually attached caps
• Verify tube reaction volumes with graduated 20 μL markings on each tube
• Track samples using the end tabs to label each strip
• Identify individual wells quickly and prevent pipetting errors using the labels (A-H) on each tube cap and cap hinge (visible when cap is open and closed)

Applications:
• PCR

Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Equipment)Thermal Cyclers
Format250 Strip Tubes w/ Attached Caps
Product LineEasyStrip™ 
Quantity250 strips
Shipping ConditionRoom Temperature
Volume (Metric) Working0.2 mL
Closure TypeFlat Cap
ColorNatural
MaterialPolypropylene
Product Type8-strip Tube
Volume (Metric)0.25 mL
Unit SizeEach

Frequently asked questions (FAQs)

What are the benefits of using EasyStrip Plus Tube Strip with Attached Flat Caps?

These new strips have new features including a graduated 20 µL measuring mark for visual verification and dual end tabs for better labeling and handling. In addition, the individual caps help prevent cross contamination and reduce sample evaporation.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Is the EasyStrip Plus Tube Strip with Attached Flat Caps DNase free?

Yes. The tubes are DNase/RNase/Human DNA free, as they are manufactured in a Class 100K ISO certified clean room UK production facility.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Which instruments are compatible with the EasyStrip Plus Tube Strip with Attached Flat Caps?

Please refer to the table on Page 2 of this flyer (https://tools.thermofisher.com/content/sfs/brochures/easystrip-plus-tube-flyer.pdf) for compatibility information.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.