Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
A more sustainable equivalent is available.
EasyStrip™ Plus Tube Strip with Attached Ultra Clear Caps
Thermo Scientific™

EasyStrip™ Plus Tube Strip with Attached Ultra Clear Caps

The Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Ultra Clear Caps is designed to reduce the risk of cross-contaminationRead more
Have Questions?
Catalog NumberQuantity
AB2005250 strips
Catalog number AB2005
Price (USD)
414.65
Online exclusive
532.00
Save 117.35
Each
Quantity:
250 strips
Price (USD)
414.65
Online exclusive
532.00
Save 117.35
Each
The Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Ultra Clear Caps is designed to reduce the risk of cross-contamination in qPCR assays. The tube strip is compatible with 0.2 mL thermal cycler blocks and features individually attached optical caps for quick and easy access to individual samples. In addition, graduated 20 μL markers on each tube enable visual volume verification and measurement of reagent and sample additions.

For tube strips with flat caps, please see Cat. No. AB-2000.

Features:
• Avoid cross contamination with the individually attached caps
• Verify tube reaction volumes with graduated 20 μL markings on each tube
• Track samples easily using the end tabs to label each strip
• Identify individual wells quickly and prevent pipetting errors using the labels (A-H) on each tube cap and cap hinge (visible when cap is open and closed)

Applications
• qPCR/Real-Time PCR

Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Equipment)Thermal Cyclers or Real-Time PCR Systems
Format250 Strip Tubes w/ Attached Caps
Product LineEasyStrip™ 
Quantity250 strips
Shipping ConditionRoom Temperature
Volume (Metric) Working0.2 mL
Closure TypeFlat Cap
ColorNatural, Ultra Clear
MaterialPolypropylene
Product Type8-strip Tube
Volume (Metric)0.25 mL
Unit SizeEach

Frequently asked questions (FAQs)

What are the benefits of using EasyStrip Plus Tube Strip with Attached Ultra Clear Caps?

These new strips have new features including a graduated 20 µL measuring mark for visual verification and dual end tabs for better labeling and handling. In addition, the individual caps help prevent cross contamination and reduce sample evaporation.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Is the EasyStrip Plus Tube Strip with Attached Ultra Clear Caps DNase free?

Yes. The tubes are DNase/RNase/Human DNA free, as they are manufactured in a Class 100K ISO certified clean room UK production facility.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Which instruments are compatible with EasyStrip Plus Tube Strip with Attached Ultra Clear Caps?

Please refer to the table on Page 2 of this flyer (https://tools.thermofisher.com/content/sfs/brochures/easystrip-plus-tube-flyer.pdf) for compatibility information.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.