Armadillo PCR Plate, 96-well, orange, clear wells
Thermo Scientific™

Armadillo PCR Plate, 96-well, orange, clear wells

The Thermo Scientific Armadillo PCR plates are the ultimate in high performance PCR plastics. Combining the rigidity of a polycarbonateRead more
Have Questions?
Catalog NumberQuantity
AB2396O25 Plates
Catalog number AB2396O
Price (INR)
38,283.00
Each
Quantity:
25 Plates
Price (INR)
38,283.00
Each
The Thermo Scientific Armadillo PCR plates are the ultimate in high performance PCR plastics. Combining the rigidity of a polycarbonate frame with thin-walled polypropylene wells, the Armadillo plate provides superior thermal cycling performance under all conditions without warping. These 96-well plates are orange with clear wells.

Features of Armadillo PCR plates:

Optically clear skirt—aids in visualization of liquids and improves stacking capabilities in automated handling systems
Raised rim design—improved well surface area improves seal integrity in automated heat sealing
ANSI:SBS footprint—with rigidity of polycarbonate skirt, the ANSI footprint ensures complete compatibility with robotic systems for high-throughput applications
Thermostable polycarbonate frame design—warp-resistant thermal cycling and enhanced mechanical stability in robotic handling
Thin walled U-bottom wells—optimal heat transfer and maximum sample recovery
Indented Alphanumeric Grid reference lettering—integrated frame and well design for improved heat sealing
Low profile design—allows for maximum heat transfer and minimal evaporation when utilizing low volume samples (200 μL working volume)
Barcoding available—all plates are available already barcoded and can be provided barcoded to customer specifications for fully-automated applications

Applications:
• PCR

Related Products
Armadillo PCR Plate, 96-well, blue, clear wells
Armadillo PCR Plate, 96-well, green, clear wells
Armadillo PCR Plate, 96-well, orange, clear wells
Armadillo PCR Plate, 96-well, red, clear wells
Armadillo PCR Plate, 96-well, yellow, clear wells
Armadillo PCR Plate, 96-well, clear, white wells
Armadillo PCR Plate, 96-well, blue, white wells
Armadillo PCR Plate, 96-well, green, white wells
Armadillo PCR Plate, 96-well, orange, white wells
Armadillo PCR Plate, 96-well, red, white wells
Armadillo PCR Plate, 96-well, yellow, white wells, barcoded
Armadillo PCR Plate, 96-well, clear, clear wells, barcoded
Armadillo PCR Plate, 96-well, blue, clear wells, barcoded
Armadillo PCR Plate, 96-well, green, clear wells, barcoded
Armadillo PCR Plate, 96-well, orange, clear wells, barcoded
Armadillo PCR Plate, 96-well, red, clear wells, barcoded
Armadillo PCR Plate, 96-well, yellow, clear wells, barcoded
Armadillo PCR Plate, 96-well, blue, white wells, barcoded
Armadillo PCR Plate, 96-well, green, white wells, barcoded
Armadillo PCR Plate, 96-well, orange, white wells, barcoded
Armadillo PCR Plate, 96-well, red, white wells, barcoded
This product is sold under license from Eppendorf AG.
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging25 Plate
Product TypePCR Microplate
Quantity25 Plates
Reaction SpeedLow profile
Skirt StyleSkirted
Volume (Metric) Working0.1 mL
Well ColorClear
BarcodeNo Barcode
ColorOrange, Clear
MaterialPolycarbonate, Polypropylene
No. of Wells96
Product LineArmadillo
Volume (Metric) Well0.2 mL
Unit SizeEach