Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Armadillo PCR Plate, 96-well, green, white wells, barcoded
Thermo Scientific™

Armadillo PCR Plate, 96-well, green, white wells, barcoded

The Thermo Scientific Armadillo PCR plates, Barcoded, are the ultimate in high performance PCR plastics. Combining the rigidity of aRead more
Have Questions?
Catalog NumberQuantity
BC3396G25 plates
Catalog number BC3396G
Price (USD)
238.00
Each
Quantity:
25 plates
Request bulk or custom format
Price (USD)
238.00
Each
The Thermo Scientific Armadillo PCR plates, Barcoded, are the ultimate in high performance PCR plastics. Combining the rigidity of a polycarbonate frame with thin-walled polypropylene wells, the Armadillo plate provides superior thermal cycling performance under all conditions without warping. These 96-well plates are green with white wells.

Features of Armadillo PCR plates:

• Optically clear skirt—aids in visualization of liquids and improves stacking capabilities in automated handling systems
• Raised rim design—improved well surface area improves seal integrity in automated heat sealing
• ANSI:SBS footprint—with rigidity of polycarbonate skirt, the ANSI footprint ensures complete compatibility with robotic systems for high-throughput applications
• Thermostable polycarbonate frame design—warp-resistant thermal cycling and enhanced mechanical stability in robotic handling
• Thin walled U-bottom wells—optimal heat transfer and maximum sample recovery
• Indented Alphanumeric Grid reference lettering—integrated frame and well design for improved heat sealing
• Low profile design—allows for maximum heat transfer and minimal evaporation when utilizing low volume samples (200 μL working volume)
• White colored wells—ensures the highest level of sensitivity for all PCR/qPCR reactions
• Barcoded—all plates are barcoded and can be provided barcoded to customer specifications for fully-automated applications

Applications:
• PCR
• qPCR/Real-Time PCR
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging25 Plate
Product TypePCR Microplate
Quantity25 plates
Reaction SpeedLow profile
Skirt StyleSkirted
Volume (Metric) Working0.1 mL
Well ColorWhite
BarcodeIncludes Barcode
ColorGreen, White
MaterialPolycarbonate, Polypropylene
No. of Wells96
Product LineArmadillo
Unit SizeEach