Armadillo PCR Plate, 96-well, blue, white wells
Thermo Scientific™

Armadillo PCR Plate, 96-well, blue, white wells

Thermo Scientific Armadillo PCR plates are the ultimate in high performance PCR plastics. Combining the rigidity of a polycarbonate frameRead more
Have Questions?
Catalog NumberQuantity
AB3396B25 plates
Catalog number AB3396B
Price (INR)
14,790.00
Online offer
Ends: 31-Dec-2026
24,650.00
Save 9,860.00 (40%)
Each
Quantity:
25 plates
Request bulk or custom format
Price (INR)
14,790.00
Online offer
Ends: 31-Dec-2026
24,650.00
Save 9,860.00 (40%)
Each
Thermo Scientific Armadillo PCR plates are the ultimate in high performance PCR plastics. Combining the rigidity of a polycarbonate frame with thin-walled polypropylene wells, the Armadillo plate provides superior thermal cycling performance under all conditions without warping. These 96-well plates are blue with white wells.

Features of Armadillo PCR plates:

• Optically clear skirt—aids in visualization of liquids and improves stacking capabilities in automated handling systems
• Raised rim design—improved well surface area improves seal integrity in automated heat sealing
• ANSI:SBS footprint—with rigidity of polycarbonate skirt, the ANSI footprint ensures complete compatibility with robotic systems for high-throughput applications
• Thermostable polycarbonate frame design—warp-resistant thermal cycling and enhanced mechanical stability in robotic handling
• Thin walled U-bottom wells—optimal heat transfer and maximum sample recovery
• Indented Alphanumeric Grid reference lettering—integrated frame and well design for improved heat sealing
• Low profile design—allows for maximum heat transfer and minimal evaporation when utilizing low volume samples (200 μL working volume)
• White wells—Ensures the highest level of sensitivity for qPCR reactions
• Barcoding available—all plates are available already barcoded and can be provided barcoded to customer specifications for fully-automated applications

Applications

• PCR
• qPCR/Real-Time PCR

Related Products
Armadillo PCR Plate, 96-well, clear, clear wells
Armadillo PCR Plate, 96-well, blue, clear wells
Armadillo PCR Plate, 96-well, green, clear wells
Armadillo PCR Plate, 96-well, orange, clear wells
Armadillo PCR Plate, 96-well, red, clear wells
Armadillo PCR Plate, 96-well, yellow, clear wells
Armadillo PCR Plate, 96-well, blue, white wells
Armadillo PCR Plate, 96-well, green, white wells
Armadillo PCR Plate, 96-well, orange, white wells
Armadillo PCR Plate, 96-well, red, white wells
Armadillo PCR Plate, 96-well, yellow, white wells, barcoded
Armadillo PCR Plate, 96-well, clear, clear wells, barcoded
Armadillo PCR Plate, 96-well, blue, clear wells, barcoded
Armadillo PCR Plate, 96-well, green, clear wells, barcoded
Armadillo PCR Plate, 96-well, orange, clear wells, barcoded
Armadillo PCR Plate, 96-well, red, clear wells, barcoded
Armadillo PCR Plate, 96-well, yellow, clear wells, barcoded
Armadillo PCR Plate, 96-well, blue, white wells, barcoded
Armadillo PCR Plate, 96-well, green, white wells, barcoded
Armadillo PCR Plate, 96-well, orange, white wells, barcoded
Armadillo PCR Plate, 96-well, red, white wells, barcoded
This product is sold under license from Eppendorf AG.
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging25 Plate
Product TypePCR Microplate
Quantity25 plates
Reaction SpeedLow profile
Skirt StyleSkirted
Volume (Metric) Working0.1 mL
Well ColorWhite
BarcodeNo Barcode
ColorBlue, White
MaterialPolycarbonate, Polypropylene
No. of Wells96
Product LineArmadillo
Unit SizeEach