Armadillo PCR Plate, 384-well, blue, clear wells, barcoded
Thermo Scientific™

Armadillo PCR Plate, 384-well, blue, clear wells, barcoded

The Thermo Scientific Armadillo PCR plates, Barcoded, are the ultimate in high performance PCR plastics. Combining the rigidity of aRead more
Have Questions?
Catalog NumberQuantity
BC2384B50 Plates
Catalog number BC2384B
Price (INR)
62,486.00
Each
Quantity:
50 Plates
Price (INR)
62,486.00
Each
The Thermo Scientific Armadillo PCR plates, Barcoded, are the ultimate in high performance PCR plastics. Combining the rigidity of a polycarbonate frame with thin-walled polypropylene wells, the Armadillo plate provides superior thermal cycling performance under all conditions without warping. These 384-well plates are blue with clear wells.

Features of Armadillo Barcoded PCR plates:

• Optically clear skirt—aids in visualization of liquids and improves stacking capabilities in automated handling systems
• Raised rim design—improved well surface area improves seal integrity in automated heat sealing
• ANSI:SBS footprint—with rigidity of polycarbonate skirt, the ANSI footprint ensures complete compatibility with robotic systems for high-throughput applications
• Thermostable polycarbonate frame design—warp-resistant thermal cycling and enhanced mechanical stability in robotic handling
• Thin walled U-bottom wells—optimal heat transfer and maximum sample recovery
• Indented Alphanumeric Grid reference lettering—integrated frame and well design for improved heat sealing
• Low profile design—allows for maximum heat transfer and minimal evaporation when utilizing low volume samples (30 μL working volume)
• Barcoded—all plates are provided with barcoding and can be barcoded to customer specifications for fully-automated applications

Applications:
• PCR

Related Products
Armadillo PCR Plate, 384-well, clear, clear wells
Armadillo PCR Plate, 384-well, blue, clear wells
Armadillo PCR Plate, 384-well, green, clear wells
Armadillo PCR Plate, 384-well, orange, clear wells
Armadillo PCR Plate, 384-well, red, clear wells
Armadillo PCR Plate, 384-well, yellow, clear wells
Armadillo PCR Plate, 384-well, red, white wells
Armadillo PCR Plate, 384-well, orange, clear wells, barcoded
Armadillo PCR Plate, 384-well, red, clear wells, barcoded
Armadillo PCR Plate, 384-well, yellow, clear wells, barcoded
Armadillo PCR Plate, 384-well, clear, white wells, barcoded
Armadillo PCR Plate, 384-well, blue, white wells, barcoded
Armadillo PCR Plate, 384-well, green, white wells, barcoded
Armadillo PCR Plate, 384-well, orange, white wells, barcoded
Armadillo PCR Plate, 384-well, red, white wells, barcoded
Armadillo PCR Plate, 384-well, yellow, white wells, barcoded
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging50 Plate
Product TypePCR Microplate
Quantity50 Plates
Reaction SpeedStandard
Skirt StyleSkirted
Volume (Metric) Working25 μL
Well ColorClear
BarcodeIncludes Barcode
ColorBlue, Clear
MaterialPolycarbonate, Polypropylene
No. of Wells384
Product LineArmadillo
Unit SizeEach