Run Invitrogen Mini Protein Gels in Bio-Rad's Mini-PROTEAN® Tetra Cell. Click here for more information.
Attention: BN2012BX10 is the equivalent SKU for BN1003BOX. Please contact Technical Support with any questions.
NativePAGE™ Bis-Tris Mini Protein Gels, 3 to 12%, 1.0 mm
Invitrogen™

NativePAGE™ Bis-Tris Mini Protein Gels, 3 to 12%, 1.0 mm

The Invitrogen NativePAGE Bis-Tris Gel System is a precast polyacrylamide mini-gel system that provides sensitive, high-resolution analysis of native proteins and protein complexes for molecular mass estimations, and assessment of purity.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWells
BN2012BX1015-well
BN2011BX1010-well
Catalog number BN2012BX10
Price (INR)
15,492.00
Online offer
Ends: 30-Sep-2026
28,165.83
Save 12,673.83 (45%)
Each
Wells:
15-well
Price (INR)
15,492.00
Online offer
Ends: 30-Sep-2026
28,165.83
Save 12,673.83 (45%)
Each
Invitrogen NativePAGE Bis-Tris Protein gels are precast polyacrylamide mini-gels that provides sensitive, high-resolution analysis of native proteins and protein complexes for molecular mass estimations and assessment of purity. They are based on the blue native polyacrylamide gel electrophoresis (BN PAGE) technique developed by Schägger and von Jagow, which overcomes the limitations of traditional native electrophoresis by providing a near-neutral operating pH and detergent compatibility.

Invitrogen NativePAGE Bis-Tris Protein gels are precast polyacrylamide mini-gels that provides sensitive, high-resolution analysis of native proteins and protein complexes for molecular mass estimations and assessment of purity. They are based on the blue native polyacrylamide gel electrophoresis (BN PAGE) technique developed by Schägger and von Jagow, which overcomes the limitations of traditional native electrophoresis by providing a near-neutral operating pH and detergent compatibility.

Features of the NativePAGE Bis-Tris Protein gels include:

  • Wide molecular weight resolving range of 15–10,000 kDa
  • Neutral-pH separation, which better preserves the native state of protein complexes
  • Resolution of all proteins regardless of their isoelectric point (pI)
  • Ability to analyze membrane-protein complexes in their native conformations
  • Ability to obtain better resolution than with traditional tris-glycine native electrophoresis

How NativePAGE Bis-Tris Protein gels work

In SDS-PAGE, SDS functions as a charge-shift molecule that denatures proteins by conferring on them a net negative charge and enables proteins to migrate towards the anode. BN PAGE uses Coomassie G-250 as the charge-shift molecule, which binds to proteins and confers a net negative charge while maintaining the proteins in their nondenatured, native state. The near-neutral pH of the NativePAGE Bis-Tris Protein gels provides maximum stability of both the proteins and the gel matrix, resulting in a highly sensitive method for analysis of native membrane-protein complexes and offering superior band resolution over traditional Tris-glycine gel systems.

For transferring proteins to a membrane, we recommend using NuPAGE transfer buffers for traditional wet transfer and PVDF membranes. NuPAGE transfer buffers maintain the neutral pH environment established during electrophoresis. Nitrocellulose membranes are not compatible for blotting since the membrane will bind the Coomassie G-250 dye very tightly. Alternatively, rapid semi-dry transfer can be done using the Power Blotter (Cat. No. PB0013) or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001) using PVDF membranes.

For Research Use Only.
Specifications
For Use With (Equipment)XCell SureLock Mini-Cell
Gel Thickness1.0 mm
Length (Metric)8 cm
Mode of SeparationMolecular Weight
Quantity10 gels (1 box)
Sample Loading VolumeUp to 15 μL
Shelf Life12 Months
Shipping ConditionWet Ice
Width (Metric)8 cm
Gel Percentage3 to 12%
Gel SizeMini
Gel TypeBis-Tris
Product LineNativePAGE
Separation Range30 to 10,000 kDa
Separation TypeNative
Wells15-well
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C. Do not freeze.

Frequently asked questions (FAQs)

Are NativePAGE gels and buffers compatible with Mini Gel tank?

Yes, NativePAGE gels are compatible with our Mini Gel tank, however there is a small variation from the original protocol.

The following protocol are for using NativePage gels with the Mini Gel Tank depending on if you are also performing a western transfer or not:
If you are NOT performing a western transfer:
1. Prepare 250 mL of each buffer (Anode and Dark Blue Cathode) per gel
2. After inserting a loaded NativePAGE gel into the Mini Gel Tank and pulling the clamp forward, fill the front cathode buffer chamber to the Fill Line with Dark Blue Cathode Buffer (~200 mL per gel)
3. Add 220 mL of Anode Buffer to the back anode buffer chamber for that gel. Start the gel run

If you are performing a western transfer:
1. Prepare 250 mL of Dark Blue Cathode Buffer, 250 mL of Light Blue Cathode Buffer, and 500 mL of Anode Buffer per gel
2. After inserting a loaded NativePAGE gel into the Mini Gel Tank and pulling the clamp forward, fill the front cathode buffer chamber to the Fill Line with Dark Blue Cathode Buffer (~200 mL per gel
3. Add 220 mL of Anode Buffer to the back anode buffer chamber for that gel
4. Start the gel run; pause the run after the dark blue dye has run ~1/3 of the way through gel
a. Pour out the buffers from the Mini Gel Tank
b. Refill the back anode buffer chamber with 220 mL of Anode Buffer per gel
c. Fill the front cathode buffer chamber to the Fill Line with Light Blue Cathode Buffer (~200 mL per gel)
5. Resume the gel run

Find additional tips, troubleshooting help, and resources within our Protein Biology Support Centers .

Citations & References (1)

Citations & References
Abstract
Active gamma-secretase complexes contain only one of each component.
Authors:Sato T, Diehl TS, Narayanan S, Funamoto S, Ihara Y, De Strooper B, Steiner H, Haass C, Wolfe MS,
Journal:J Biol Chem
PubMed ID:17911105
Gamma-secretase is an intramembrane aspartyl protease complex that cleaves type I integral membrane proteins, including the amyloid beta-protein precursor and the Notch receptor, and is composed of presenilin, Pen-2, nicastrin, and Aph-1. Although all four of these membrane proteins are essential for assembly and activity, the stoichiometry of the complex ... More