Click-iT™ RNA Alexa Fluor™ 594 Imaging Kit
Invitrogen™

Click-iT™ RNA Alexa Fluor™ 594 Imaging Kit

The Click-iT® RNA Alexa Fluor® 594 Imaging Kit enables detection of global RNA transcription temporally and spatially in cells andRead more
Have Questions?
Catalog NumberQuantity
C103301 kit
Catalog number C10330
Price (INR)
-
Quantity:
1 kit
The Click-iT® RNA Alexa Fluor® 594 Imaging Kit enables detection of global RNA transcription temporally and spatially in cells and tissues (PNAS (2008) 105:15779-84).The ability to detect newly synthesized RNA or changes in RNA levels resulting from disease, environmental damage or drug treatments is an important aspect of toxicological profiling. Utilizing an alkyne-modified nucleoside, 5-ethynyl uridine (EU), and powerful click chemistry, newly synthesized RNA can be detected without the use of radioactivity or antibodies with a simple, two-step procedure. In step one, the alkyne-containing nucleoside is fed to cells or animals and actively incorporated into nascent RNA. The small size of the tag enables efficient incorporation of the modified nucleoside into RNA, but not into DNA. Detection utilizes the chemoselective ligation or “click" reaction between and azide and an alkyne where the modified RNA is detected with a corresponding azide-containing dye. With its dimunitive “footprint", the Click-iT® detection molecule can easily penetrate complex samples and leaves open the possibility of multiplex analyses with other probes, including antibodies for the detection of RNA-interactive proteins for deeper biological insights.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorRed
Excitation/Emission590/617
For Use With (Equipment)Confocal Microscope, Fluorescence Microscope
Product LineAlexa Fluor, Click-iT, Molecular Probes
Product TypeNew RNA Synthesis Assay
Quantity1 kit
Shipping ConditionRoom Temperature
Detection MethodFluorescence
FormatSlide
Label or DyeAlexa Fluor 594
Unit SizeEach
Contents & Storage
The kit contains sufficient material for 25, 18 X 18 coverslips. Store at 2-6°C, dessicate and protect from light.

Frequently asked questions (FAQs)

Can the components of the Click-iT Plus EdU Alexa Fluor Imaging Kits be used for labeling samples for flow cytometry?

Other than the EdU (Component A), DMSO (Component C), and Click-iT EdU buffer additive (Component F or G), all other components in the respective kits are not interchangeable. The types of reagents and amounts provided per kit were optimized either for imaging or flow cytometry. Using the imaging reagents may result in an excessive level of signal for flow cytometry detection and using the flow cytometry reagents may result in sub-optimal signal for imaging.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can any type of cell incorporate EU?

No. The cell must possess a pyrimidine salvage pathway; without this pathway, EU does not become phosphorylated to allow incorporation during RNA synthesis.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is the approximate fluorescence excitation/emission maxima for Alexa Fluor 594 azide?

The fluorescence excitation/emission maxima is 590/615 nm.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (13)

Citations & References
Abstract
Visualization of actin filaments and monomers in somatic cell nuclei.
Authors:Belin BJ, Cimini BA, Blackburn EH, Mullins RD,
Journal:Mol Biol Cell
PubMed ID:23447706
'In addition to its long-studied presence in the cytoplasm, actin is also found in the nuclei of eukaryotic cells. The function and form (monomer, filament, or noncanonical oligomer) of nuclear actin are hotly debated, and its localization and dynamics are largely unknown. To determine the distribution of nuclear actin in ... More
Bookmarking by specific and nonspecific binding of FoxA1 pioneer factor to mitotic chromosomes.
Authors:Caravaca JM, Donahue G, Becker JS, He X, Vinson C, Zaret KS,
Journal:Genes Dev
PubMed ID:23355396
While most transcription factors exit the chromatin during mitosis and the genome becomes silent, a subset of factors remains and
Visualizing coronavirus RNA synthesis in time by using click chemistry.
Authors:Hagemeijer MC, Vonk AM, Monastyrska I, Rottier PJ, de Haan CA,
Journal:J Virol
PubMed ID:22438542
Coronaviruses induce in infected cells the formation of replicative structures, consisting of double-membrane vesicles (DMVs) and convoluted membranes, where viral RNA synthesis supposedly takes place and to which the nonstructural proteins (nsp's) localize. Double-stranded RNA (dsRNA), the presumed intermediate in RNA synthesis, is localized to the DMV interior. However, as ... More
Dynamics and Spatial Genomics of the Nascent Transcriptome by Intron seqFISH.
Authors:
Journal:Cell
PubMed ID:29887381
Topoisomerase II-Induced Chromosome Breakage and Translocation Is Determined by Chromosome Architecture and Transcriptional Activity.
Authors:
Journal:Mol Cell
PubMed ID:31202577