Pair with TrueCut Cas9 Protein v2 to help achieve higher editing efficiency.
Lipofectamine™ CRISPRMAX™ Cas9 Transfection Reagent
Invitrogen™

Lipofectamine™ CRISPRMAX™ Cas9 Transfection Reagent

Lipofectamine CRISPRMAX Cas9 Transfection Reagent is an optimized lipid-based transfection reagent for CRISPR-Cas9 protein delivery, providing the cleavage efficiency of electroporation with simplicity and scalability.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
CMAX000030.3 mL
CMAX000080.75 mL
CMAX000010.1 mL
CMAX000151.5 mL
Catalog number CMAX00003
Price (INR)
18,221.00
Online offer
Ends: 31-Dec-2026
30,367.00
Save 12,146.00 (40%)
Each
Quantity:
0.3 mL
Price (INR)
18,221.00
Online offer
Ends: 31-Dec-2026
30,367.00
Save 12,146.00 (40%)
Each

Lipofectamine CRISPRMAX Cas9 Transfection Reagent is the first optimized lipid-based transfection reagent for CRISPR knockout applications. It effectively delivers the Cas9 protein and guide RNA (gRNA) ribonucleoprotein (RNP) complex, ensuring high transfection efficiency and effective gene knockout. The proprietary lipid-based transfection reagent minimizes cellular toxicity, making it suitable to for sensitive cells. Its ease of use and compatibility with high-throughput screening make it a cost-effective alternative to electroporation.

Combined with our TrueCut Cas9 Protein v2 and TrueGuide Synthetic gRNA, it provides:

  • Optimized for CRISPR—specially formulated for CRISPR-Cas9 RNP applications
  • Superior gene knockout—efficient Cas9 transfection ensures higher genomic cleavage
  • Broad cell type applicability—versatile for use in a wide range of cell lines, including primary cells and stem cells
  • Minimal cytotoxicity—viable, healthy cells maintained throughout Cas9 transfection
  • High-throughput compatible—an ideal delivery solution for functional genomic studies with CRISPR-Cas9

Achieve high transfection efficiency in novel genome editing applications

Lipofectamine CRISPRMAX Transfection Reagent enables successful Cas9 transfection in a wide range of cell types, including iPSC, mESC, N2A, CHO, A549, HCT116, HeLa, HEK293, and more. Unlike CRISPR plasmid or Cas9 mRNA, this protein-based approach eliminates the need for transcription or translation, reducing the risk of genomic integration and making the process cell cycle-independent. When combined with our TrueCut Cas9 Protein v2 and TrueGuide Synthetic gRNA, it ensures superior cleavage efficiency and effective gene editing.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ClassificationAnimal Origin-Free
For Use With (Application)Transfection
Product LineCRISPRMAX, Lipofectamine
Product TypeTransfection Reagent
Quantity0.3 mL
Serum CompatibleYes
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Cell TypeEstablished Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample TypeProtein, Ribonucleoprotein, Cas9 Ribonucleoprotein, Cas9, Plasmid DNA
Transfection TechniqueLipid-based Transfection
Unit SizeEach
Contents & Storage
Contains one vial of Lipofectamine CRISPRMAX Reagent and one vial of Cas9 Plus Reagent.

Store at 2–8°C. Do not freeze.

Frequently asked questions (FAQs)

Can I use Lipofectamine CRISPRMAX Cas9 Transfection Reagent with one-cell embryos?

We have no data regarding the use of CRISPRMAX in one-celled embryos, therefore we do not recommend using this product for this application.

The shipping for my Lipofectamine CRISPRMAX Cas9 Transfection Reagent was delayed. Can I still use it?

All of the Lipofectamine reagents, including Lipofectamine CRISPRMAX Cas9 Transfection Reagent, are stable at room temperature for months. What is most important is that the reagent did not freeze, because that will alter the integrity and composition of the lipid particles and will compromise usability. Previous freezing can be evidenced by precipitation and excessive cloudiness of the reagent. A limited amount of cloudiness is normal, but excessive cloudiness is a sign that the reagent froze at some point. If the reagent you received looks clear or a bit cloudy the product should still be usable. Some additional data on room temperature stability of some Lipofectamine reagents, in the context of shipping, can be found in this white paper.

Find additional tips, troubleshooting help, and resources within our Transfection Support Center.

How much Lipofectamine CRISPRMAX Reagent, Cas9 protein, and gRNA should I complex for gene editing?

Please see the Lipofectamine CRISPRMAX Reagent manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0014545_lipofectamine_crispermax_QR.pdf) for protocol details or contact techsupport@thermofisher.com.

How does the Lipofectamine CRISPRMAX Reagent work?

The Lipofectamine CRISPRMAX Reagent combined with the proprietary enhancement properties of the Lipofectamine Cas9 Plus Reagent leads to efficient complex formation with the Cas9-gRNA ribonucleoprotein, for best delivery to the nucleus, helping to ensure high gene editing frequency for a wide range of cell types.

Can the Lipofectamine CRISPRMAX Reagent be used to deliver proteins other than the Cas9 nuclease?

Although the Lipofectamine CRISPRMAX Reagent was developed for the delivery of the Cas9-gRNA ribonucleoprotein, there is potential for other protein complex applications.