Search
| Catalog Number | Description |
|---|---|
| D14031 | Discontinued; replacement product is 50132131 |
| D14041 | Descontinued; replacement product is 50132132 |
This item is no longer available and has been replaced by the next generation of Barnstead Water Purification Systems. The recommended replacement for this product is the Thermo Scientific™ Barnstead™ Pacific™ TII water purification system.
Produce Type 2 pure water that exceeds ASTM and CLSI-CLRW standards with the smart and flexible Thermo Scientific™ Barnstead™ TII Type 2 Water System. This system is ideal for all applications requiring pure water such as the preparation of microbiological media and buffers, glassware washing and as feed water for Type 1 ultrapure water systems.
Features:
Smart, Easy to Use, Low-Maintenance System:
Ordering Information:
Optional accessories include remote dispensers, printer, cutoff float, interlock cable, storage reservoirs, bottles and replacement consumables. These products are available separately. For details, contact your sales representative.
Includes:
Requires:
Recommended for: