Run Invitrogen Mini Protein Gels in Bio-Rad's Mini-PROTEAN® Tetra Cell. Click here for more information.
Novex™ IEF Protein Gels, pH 3-7, 1.0 mm
Invitrogen™

Novex™ IEF Protein Gels, pH 3-7, 1.0 mm

Novex™ pH 3-7 IEF Gels are used for pI determination and are excellent for native applications using soluble proteins. Isoelectric focusing (IEF) is an electrophoresis technique that separates proteins based on their isoelectric point (pI).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWells
EC6645BOX10-well
EC66452BOX12-well
Catalog number EC6645BOX
Price (INR)
-
Wells:
10-well
Novex™ pH 3-7 IEF Gels are used for pI determination and are excellent for native applications using soluble proteins. Isoelectric focusing (IEF) is an electrophoresis technique that separates proteins based on their isoelectric point (pI). The pI is the pH at which a protein has no net charge and does not move in an electric field. Vertical IEF gels effectively create a pH gradient so proteins separate according to their unique pI.

IEG gels can be used to readily detect minor changes in a protein due to deamination, phosphorylation, or glycosylation, and can resolve different proteins of similar size that cannot be resolved on standard SDS-PAGE gels. Pre-focusing is not required. Total run time is approximately 2.5 hours.

Formulation
Novex™ IEF gels are 5% polyacrylamide and consist of high-purity acrylamide, bisacrylamide, TEMED, APS, ultrapure water, and 2% ampholytes. They do not contain denaturing reagents.

Recommended buffers
IEF is a sensitive technique which is affected by many factors. Our optimized, premixed IEF buffers reduce variability and help you obtain consistent results.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)XCell SureLock Mini-Cell
Gel Thickness1.0 mm
Mode of SeparationpI (Charge)
Quantity5 Gels/Box
Sample Loading Volume25 μL
Shelf Life6 Months
Shipping ConditionWet Ice
Storage RequirementsStore at 2°C to 8°C. Do not freeze.
Width (Metric)8 cm
Gel Percentage5%
Gel SizeMini
Gel TypeIEF
Product LineNovex
Separation TypeIsoelectric Focusing
Wells10-well
pH Range3.5 to 6.8
Unit SizeEach

Frequently asked questions (FAQs)

What may cause streaking on the 2nd dimension gel after IEF?

There are several reasons why streaking may occur.

(1) Sample is not completely solubilized prior to application.

(2) Sample is poorly soluble in rehydration solution.

(3) Non-protein impurities in the sample can interfere with IEF, causing horizontal streaking in the final 2-D result, particularly toward the acidic side of the gel.

(4) Ionic impurities are present in sample.

(5) Ionic detergent is present in sample.

(6) Sample load is too high.

(7) Underfocusing. Focusing time was not long enough to achieve steady state focusing.

(8) Overfocusing. Extended focusing times (over 100,000 Vh) may result in electroendosmotic water and protein movement, which can produce horizontal smearing.

What should be done?

(1) Be sure that the sample is completely and stably solubilized. Note: Repeated precipitation-resolubilization cycles produce or increase horizontal streaking.

(2) Increase the concentration of the solubilizing components in the rehydration solution.

(3) Modify sample preparation to limit these contaminants or dialyze protein.

(4) Reduce salt concentration to below 10 mM by dilution or desalt the sample by dialysis. Precipitation with TCA and acetone and subsequent resuspension is another effective desalting technique that removes lipids, nucleotides and other small molecules.
Note: Specific and non-specific losses of proteins can occur with dialysis, gel chromatography, and precipitation/resuspension of samples. If the sample preparation cannot be modified, the effect of ionic impurities can be reduced by modifying the IEF protocol. Limit the voltage to 100-150 V for 2 hours, then resume a normal voltage step program. This pre-step allows the ions in the sample to move to the ends of the IPG strip.

(5) If the ionic detergent SDS is used in sample preparation, the final concentration must not exceed 0.25% after dilution into the rehydration solution. Additionally, the concentration of the non-ionic detergent present must be at least 8 times higher than the concentration of any ionic detergent to ensure complete removal of SDS from the proteins.

(6) Extend focusing time. Load less sample.

(7) Prolong focusing time.

(8) Reduce focusing time.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Why would a monoclonal antibody run as a smear on an Invitrogen IEF gel while the IEF marker focuses perfectly?

It is not unusual for antibodies (even monoclonals) to be differentially glycosylated, and therefore not focus well on IEF gels. Sometimes a monoclonal antibody can even run more smeary or unfocused than a polyclonal, over a range of pH 6.5 to 8.0. Try running the gel longer to improve the focus, but often the improvement is minimal if anything.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

When running an Invitrogen IEF gel, why does the voltage have to be adjusted from 100 to 200 to 500 V?

The initial voltage is to set up the ampholytes in a pH gradient, the second step actually drives the proteins to their pI. The third step is to "fine focus" or sharpen the protein bands.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What does it mean when bands appear to be getting narrower (or "funneling") as they progress down a protein gel?

There may be too much beta-mercaptoethanol (BME), sample buffer salts, or dithiothreitol (DTT) in your samples. If the proteins are over-reduced, they can be negatively charged and actually repel each other across the lanes causing the bands to get narrower as they progress down the gel.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Will NP-40 affect the migration of the samples in the SDS-PAGE gel?

Yes. All detergents and even phospholipids in cell extracts will form mixed micelles with SDS and migrate down into the gel.

They can also interfere with the SDS:protein binding equilibrium. Most of the nonionic detergents significantly interfere with SDS-PAGE.

We recommend that you keep the ratio of SDS to lipid or other detergent at 10:1 (or greater) to minimize these effects.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.