Run Invitrogen Mini Protein Gels in Bio-Rad's Mini-PROTEAN® Tetra Cell. Click here for more information.
Novex™ Tricine Mini Protein Gels, 16%, 1.0 mm
Invitrogen™

Novex™ Tricine Mini Protein Gels, 16%, 1.0 mm

Invitrogen Novex Tricine Gels provide separation of low molecular weight proteins and peptides. In this system tricine replaces glycine in the running buffer, resulting in more efficient stacking and destacking for low molecular weight proteins and higher resolution of smaller peptides.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWells
EC66952BOX12-well
EC6695BOX10-well
EC66955BOX15-well
Catalog number EC66952BOX
Price (INR)
17,414.00
Online offer
Ends: 30-Sep-2026
31,661.00
Save 14,247.00 (45%)
Each
Wells:
12-well
Price (INR)
17,414.00
Online offer
Ends: 30-Sep-2026
31,661.00
Save 14,247.00 (45%)
Each
Invitrogen Novex Tricine Gels provide separation of low molecular weight proteins and peptides. The Tricine system is a modification of the tris-glycine discontinuous buffer system developed by Schaegger and von Jagow (Schaegger and von Jagow, 1987) specifically for resolving peptides and low molecular weight proteins. In this system the tricine replaces the glycine in the running buffer, resulting in more efficient stacking and destacking for low molecular weight proteins and higher resolution of smaller peptides.

Features of Novex Tricine protein gels:

  • Increased resolution of proteins with molecular weights as low as 2 kDa
  • Improved compatibility with direct sequencing of proteins after transferring to PVDF
  • Minimized protein modification due to the lower pH of the tricine buffering system

Formulation

Invitrogen Tricine gels are made with high-purity, strictly quality-controlled reagents: Tris base, HCl, acrylamide, bisacrylamide, TEMED, APS, and highly purified water. Our Tricine gels have a 4% stacking gel and do not contain SDS. The Tricine system requires SDS in sample and running buffers for best results.

Choose the right Tricine gel for your protein separation

Invitrogen Tricine gels come in three polyacrylamide concentrations of 10%, 16%, and a gradient of 10–20%. Select from our many well formats, including 10-, 12-, and 15-well. Tricine gels are formulated for denaturing gel electrophoresis applications. For optimal sample preparation, we recommend Tricine SDS Sample Buffer (LC1676) and optimal separation use Tricine SDS Running Buffer (LC1675).

For transfer of proteins to a membrane, we recommend using the Novex Tris-Glycine Transfer Buffer (LC3675) if performing a traditional wet transfer using the XCell II Blot Module (EI9051) or the Mini Blot Module (B1000). Rapid semi-dry transfer can be performed using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Mini Gel Tank
Gel Thickness1.0 mm
Length (Metric)8 cm
Mode of SeparationMolecular Weight
Quantity10 Gels/Box
Recommended ApplicationsDenaturing
Sample Loading VolumeUp to 20 μL
Shelf Life12 Weeks
Shipping ConditionWet Ice
Storage RequirementsStore at 2°C to 8°C. Do not freeze.
Width (Metric)8 cm
Gel Percentage16%
Gel SizeMini
Gel TypeTricine
Product LineNovex
Separation Range2.5 to 200 kDa
Separation TypeDenaturing
Wells12-well
Unit SizeEach

Frequently asked questions (FAQs)

What does it mean when bands appear to be getting narrower (or "funneling") as they progress down a protein gel?

There may be too much beta-mercaptoethanol (BME), sample buffer salts, or dithiothreitol (DTT) in your samples. If the proteins are over-reduced, they can be negatively charged and actually repel each other across the lanes causing the bands to get narrower as they progress down the gel.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

If a Tricine gel heats up to around 37°C during a run, should any precautions be taken?

A temperature increase to 35°C to 40°C during electrophoresis is not uncommon for Tricine gels. If you want to run the gels at a cooler temperature, the lower (outer) buffer chamber can be filled higher or they can be run at a lower voltage, for example 100 V.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What type of transfer buffer should be used with Invitrogen Tricine gels?

For non-sequencing applications, any transfer buffer used with Tris-Glycine gels can be used with Tricine gels including Tris-Glycine transfer buffer. For sequencing applications, the buffer should be chemically compatible with sequencing protocols. Non-glycine based transfer buffers such as the NuPAGE Transfer buffer, 1/2X TBE Transfer buffer, or CAPS Buffer can be used for N-terminal sequencing . Generally, a pH which is close to neutral is desirable to maintain gel and protein stability. High current should be avoided because it can lead to heat generation and instability.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

If a Tricine gel is accidentally run with buffers used in the Tris-Glycine system, what will happen and why?

If the Tricine gel is run with Tris-Glycine sample buffer, the bands will behave abnormally and resolve poorly. If the Tricine gel is accidentally run with Tris-Glycine running buffer, the gel will take longer to run and the resolution, especially for smaller proteins, will be worse than when the proteins are run on a Tris-Glycine gel with Tris-Glycine buffers. This is due to a combination of increase in stack area size (glycine is a slower ion than Tricine) and the higher ionic strength of the Tricine gel.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What is the cause of smeary artifacts down the lanes of a Tricine gel and how can this be prevented?

Protein samples are possibly reoxidizing before the run is complete in the Tricine gel system. Since Tricine is a glycine derivative, the running pH ranges of the two systems are different. As a consequence, reduced samples tend to oxidize more in the Tricine system. Adding more reducing agent will not solve the problem.

One option is to alkylate the sample by reducing with 20 mM DTT at 70°C for 30 min, followed by 50 mM iodoacetic acid to alkylate.

Another method which inhibits oxidation is the addition of thioglycolic acid (TGA) to the running buffer. The reference to this is described by Hunkapiller et al, Methods of Enzymology, (91), 399, 1983.

Caution should be taken when using this method since this compound is both toxic and expensive. In addition, the TGA must be fresh as it tends to become oxidized itself over time. Oxidized TGA will actually promote sample re-oxidation.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Citations & References (4)

Citations & References
Abstract
The Solution Structures of the Human beta -Defensins Lead to a Better Understanding of the Potent Bactericidal Activity of HBD3 against Staphylococcus aureus.
Authors: Schibli David J.; Hunter Howard N.; Aseyev Vladimir; Starner Timothy D.; Wiencek John M.; McCray Paul B. Jr; Tack Brian F.; Vogel Hans J.;
Journal:J Biol Chem
PubMed ID:11741980
'The three human beta-defensins, HBD1--3, are 33--47-residue, cationic antimicrobial proteins expressed by epithelial cells. All three proteins have broad spectrum antimicrobial activity, with HBD3 consistently being the most potent. Additionally, HBD3 has significant bactericidal activity against Gram-positive Staphylococcus aureus at physiological salt concentrations. We have compared the multimeric state of ... More
Phosphatidylinositol mannoside from Mycobacterium tuberculosis binds alpha5beta1 integrin (VLA-5) on CD4+ T cells and induces adhesion to fibronectin.
Authors:Rojas RE, Thomas JJ, Gehring AJ, Hill PJ, Belisle JT, Harding CV, Boom WH,
Journal:J Immunol
PubMed ID:16920931
'The pathological hallmark of the host response to Mycobacterium tuberculosis is the granuloma where T cells and macrophages interact with the extracellular matrix (ECM) to control the infection. Recruitment and retention of T cells within inflamed tissues depend on adhesion to the ECM. T cells use integrins to adhere to ... More
Molecular characterization of major cat allergen Fel d 1: expression of heterodimer by use of a baculovirus expression system.
Authors:Seppälä U, Hägglund P, Wurtzen PA, Ipsen H, Thorsted P, Lenhard T, Roepstorff P, Spangfort MD,
Journal:J Biol Chem
PubMed ID:15546862
Fel d 1 is a major cat allergen inducing allergic rhinitis and asthma in sensitized individuals. It has a more complex structure when compared with other allergens and therefore expression of recombinant Fel d 1 has been considered a challenge. The present study shows for the first time that a ... More
Active gamma-secretase complexes contain only one of each component.
Authors:Sato T, Diehl TS, Narayanan S, Funamoto S, Ihara Y, De Strooper B, Steiner H, Haass C, Wolfe MS,
Journal:J Biol Chem
PubMed ID:17911105
Gamma-secretase is an intramembrane aspartyl protease complex that cleaves type I integral membrane proteins, including the amyloid beta-protein precursor and the Notch receptor, and is composed of presenilin, Pen-2, nicastrin, and Aph-1. Although all four of these membrane proteins are essential for assembly and activity, the stoichiometry of the complex ... More