TOPO™ XL-2 Complete PCR Cloning Kit, without competent cells
TOPO™ XL-2 Complete PCR Cloning Kit, without competent cells
Invitrogen™

TOPO™ XL-2 Complete PCR Cloning Kit, without competent cells

The TOPO™ XL-2 Complete PCR Cloning Kit provides all the necessary elements for efficient cloning of extra-long PCR products (upRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
K8040-1010 Reactions
K8040-2020 Reactions
Catalog number K8040-10
Price (INR)
21,003.00
線上優惠
Ends: 31-Mar-2026
31,115.00
Save 10,112.00 (32%)
Each
Add to cart
No. of Reactions:
10 Reactions
Price (INR)
21,003.00
線上優惠
Ends: 31-Mar-2026
31,115.00
Save 10,112.00 (32%)
Each
Add to cart
The TOPO™ XL-2 Complete PCR Cloning Kit provides all the necessary elements for efficient cloning of extra-long PCR products (up to 13 kb). The kit uses the linearized and topoisomerase 1-activated pCR™-XL-2-TOPO™ vector, which is compatible with the cloning of blunt-end PCR fragments. Amplification of long PCR fragments is enabled by Platinum™ SuperFi™ Green PCR Master Mix, which is included in the kit. Topoisomerase I activation of the vector allows PCR products to be ligated in just 5 minutes on your bench–top, resulting in high cloning efficiency (up to 90% positive recombinants with 10 kb fragments).

The pCR-XL-2-TOPO vector includes:
• ccdB gene for positive selection
• EcoR I site flanking the PCR product insertion site for easy excision of inserts
• Ampicillin- and kanamycin-resistance genes for your choice of selection in E. coli
• T7 promoter/priming site for in vitro RNA transcription and sequencing
• T7, T3, and M13 forward and reverse primer sites for sequencing

The TOPO XL-2 Complete PCR Cloning Kit includes:
• TOPO XL-2 PCR Cloning Kits containing the pCR-XL-2-TOPO vector
Platinum SuperFi Green PCR Master Mix—featuring a proofreading DNA polymerase (>100X fidelity compared to Taq) with high processivity and a density gradient to generate accurate, long PCR amplicons ready to load onto an agarose gel for gel extraction
PureLink™ Quick Gel Extraction and PCR Purification Combo Kit—designed to purify DNA fragments from agarose gels or direct PCR purification using a silica-based spin cartridge in < 30 minutes

For a kit that includes competent cells, please see Cat. Nos. K805010 and K805020.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialAmpicillin (AmpR), Kanamycin (KanR)
Bacterial or Yeast StrainNot Included
Cloning MethodZero Blunt TOPO
For Use With (Application)PCR Cloning
No. of Reactions10 Reactions
Product LineTOPO
Product TypeCloning Kit
PromoterT3, T7
Quantity10 reactions
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
VectorpCR-XL-2-TOPO
FormatKit
Unit SizeEach
Contents & Storage
Box 1: Store at -5 to -30°C.
• pCR XL-2-TOPO vector
• Salt Solution
• T3 Primer
• T7 Primer
• XL Control PCR Template
• XL Control PCR Primers
• Platinum SuperFi Green PCR Master Mix
• SuperFi GC Enhancer
• Nuclease-free water

Box 2: Store at room temperature.
• Gel Solubilization Buffer (L3)
• Binding Buffer (B2)
• Wash Buffer (W1)
• Elution Buffer (E1)
• PureLink Clean-up Spin Columns (in Wash Tubes)
• PureLink Recovery/Elution Tubes