Run Invitrogen Mini Protein Gels in Bio-Rad's Mini-PROTEAN® Tetra Cell. Click here for more information.
Bolt™ Bis-Tris Plus Mini Protein Gels, 8%, 1.0 mm, WedgeWell™ format
Invitrogen™

Bolt™ Bis-Tris Plus Mini Protein Gels, 8%, 1.0 mm, WedgeWell™ format

Invitrogen Bolt Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of a wide range of protein sizes under denaturing conditions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWells
NW00085BOX15-well
NW00080BOX10-well
NW00082BOX12-well
NW00087BOX17-well
Catalog number NW00085BOX
Price (INR)
-
Wells:
15-well
Invitrogen Bolt Bis-Tris Plus gels are precast polyacrylamide gels designed for optimal separation of a wide range of protein sizes under denaturing conditions. Similar to NuPAGE Bis-Tris gels, Bolt Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications with increased loading capacity. Bolt gels are ideal for western blot transfer and analysis, and any other techniques where protein integrity is crucial.

Use Bolt Bis-Tris Plus gels to obtain optimal results for your day-to-day protein separation needs. Bolt Bis-Tris Plus gels come in four gel types and multiple well formats. Each box contains 10 gels. A Mini Gel Tank (A25977) is required for use of all Bolt gels.

Features include:
Optimized run conditions—separate your proteins using constant voltage in approximately 20 minutes
Long shelf life—store gels for at least 16 months at 4–25°C
Better protein integrity—neutral pH sample preparation process preserves your proteins
Wedge-shaped wells—for easier sample loading and up to two times higher sample volume

Choose the right Bolt Bis-Tris Plus gels for your protein separation
Obtain optimal separation of your proteins by choosing the right combination of gel and running buffer. Bolt Bis-Tris Plus gels come in four polyacrylamide concentrations: 8%, 10%, 12%, and 4–12% gradient. The gels can be run using Bolt MES SDS (B000202) or Bolt MOPS SDS (B000102) running buffer to obtain different separation ranges. Bolt Bis-Tris Plus gels consist of a neutral pH-buffered (pH 6.4) polyacrylamide gel and have an operating pH of about 7.0. Bolt Bis-Tris Plus gels also come in four different well formats: 10, 12, 15, and 17 wells.

Convenient to use
Bolt Bis-Tris Plus gels can be run using constant voltage in approximately 20 minutes with the Bolt MES running buffer. Bolt Bis-Tris Plus gels offer sharper bands, high resolution, longer shelf life, and higher accuracy of results than Tris-glycine–based gels. In addition, Bolt Bis-Tris Plus gels have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0-mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier. And Bolt Bis-Tris Plus gels have a shelf life of at least 16 months at 4°C to 25°C.

For transferring your proteins to a membrane, we recommend using for Bolt Transfer buffer (BT0006) for traditional wet transfer using the Mini Blot Module (B1000) . Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the Blot 2 Gel Transfer Device (IB21001).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)XCell SureLock Mini-Cell
Gel Thickness1.0 mm
Length (Metric)8 cm
Mode of SeparationMolecular Weight
Quantity10 gels (1 box)
Recommended ApplicationsDenaturing
Sample Loading VolumeUp to 35 μL
Shelf Life16 Months
Shipping ConditionRoom Temperature
Storage RequirementsStore at 4–25°C. Do not freeze.
Width (Metric)8 cm
Gel Percentage8%
Gel SizeMini
Gel TypeBis-Tris Plus
Product LineBolt, WedgeWell
Separation Range3.5 to 260 kDa
Separation TypeDenaturing
Wells15-well
Unit SizeEach

Frequently asked questions (FAQs)

Can I prepare my protein sample with the reducing agent and store it for future use?

DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

My LDS or SDS sample buffer precipitates when stored at 4 degrees C. Can I warm it up? Can I store it at room temperature?

Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

What are the storage conditions for Bolt gels?

Similar to NuPAGE gels with storage temperatures of 4 to 25 degrees C.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What does it mean when bands appear to be getting narrower (or "funneling") as they progress down a protein gel?

There may be too much beta-mercaptoethanol (BME), sample buffer salts, or dithiothreitol (DTT) in your samples. If the proteins are over-reduced, they can be negatively charged and actually repel each other across the lanes causing the bands to get narrower as they progress down the gel.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How much methanol is required per transfer when using the Mini Blot Module and Bolt Bis-Tris Plus gels?

We recommend using 1x Bolt Transfer buffer with 10% methanol.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.