Power Blotter XL Welcome Pack, PVDF
Invitrogen™
Power Blotter XL Welcome Pack, PVDF
The Invitrogen Power Blotter XL System (Power Blotter Station + Power Blotter Cassette XL) is designed specifically for rapid semi-dryRead more
Have Questions?
Catalog NumberQuantity
PB01171 kit
Catalog number PB0117
Price (INR)
208,126.00
線上優惠
Ends: 31-Dec-2026
378,410.00
Save 170,284.00 (45%)
Each
Quantity:
1 kit
Price (INR)
208,126.00
線上優惠
Ends: 31-Dec-2026
378,410.00
Save 170,284.00 (45%)
Each
The Invitrogen Power Blotter XL System (Power Blotter Station + Power Blotter Cassette XL) is designed specifically for rapid semi-dry transfer of proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 12 minutes. The Power Blotter System features an integrated power supply optimized to enable consistent and high-efficiency protein transfer.
Learn more about semi-dry transfer with the Power Blotter XL System ›
The Power Blotter XL Welcome Pack, PVDF, includes:
• Power Blotter Station
• Power Blotter Cassette XL
• Power Blotter Select Transfer Stacks, PVDF, regular size
• Power Blotter Pre-cut Membranes and Filters, PVDF, regular size
• Power Blotter 1-Step Transfer Buffer
Features of the Power Blotter XL System include:
• Efficient—high transfer efficiency with a broad range of protein sizes compared to conventional semi-dry or wet (tank) transfer methods
• Fast—5- to 12-minute transfer when used with Power Blotter Select Transfer Stacks or Power Blotter 1-Step Transfer Buffer
• Integrated power supply—seamless operation between control unit and cassette; no need for external power supply
• Easy-touch programming—convenient pre-programmed transfer methods, or create, save, and run customized transfer methods
• Flexible—simultaneous transfer of 1 to 4 mini-size or 1 or 2 midi-size gels using the Power Blotter XL Cassette
• Versatile—compatible with Power Blotter Select Transfer Stacks and Pierce 1-Step Transfer Buffer for rapid blotting programs or Towbin transfer buffer for conventional semi-dry transfer methods
The Power Blotter System achieves highly efficient transfer in seven minutes when used with Power Blotter Select Transfer Stacks or ten minutes with Power Blotter 1-Step Transfer Buffer. The instrument is also effective for standard 30- to 60-minute semi-dry transfer protocols based on traditional buffers.
Related products
Power Blotter XL Welcome Pack
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Acrylamide Gels
Mode of TransferSemi-Dry
Quantity1 kit
Shipping ConditionRoom Temperature
MaterialPVDF
Product LinePower Blotter
Unit SizeEach
Contents & Storage
• Power Blotter Station
• Power Blotter Cassette XL
• Power Blotter Select Transfer Stacks, PVDF, regular-size
• Power Blotter Pre-cut Membranes and Filters, PVDF, regular-size
• Power Blotter 1-Step Transfer Buffer