Power Blotter Welcome Pack
Invitrogen™
Power Blotter Welcome Pack
The Power Blotter Welcome Pack includes:Power Blotter StationPower Blotter CassettePower Blotter Select Transfer Stacks, nitrocellulose, regular sizePower Blotter Pre-cut MembranesRead more
Have Questions?
Catalog NumberQuantity
PB01121 Kit
Catalog number PB0112
Price (USD)
4,120.00
Each
Quantity:
1 Kit
Request bulk or custom format
Price (USD)
4,120.00
Each
The Power Blotter Welcome Pack includes:
Power Blotter Station
Power Blotter Cassette
Power Blotter Select Transfer Stacks, nitrocellulose, regular size
Power Blotter Pre-cut Membranes and Filters, nitrocellulose, regular size
Power Blotter 1-Step Transfer Buffer
The Invitrogen Power Blotter System (Power Blotter Station + Power Blotter Cassette) is designed specifically for rapid semi-dry transfer of proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 12 minutes. The Power Blotter System features an integrated power supply optimized to enable consistent and high-efficiency protein transfer.
Features of the Power Blotter System include:
• Efficient—high transfer efficiency with a broad range of protein sizes compared to conventional semi-dry or wet (tank) transfer methods
• Fast—5- to 12-minute transfer when used with Power Blotter Select Transfer Stacks or Power Blotter 1-Step Transfer Buffer
• Integrated power supply—seamless operation between control unit and cassette; no need for external power supply
• Easy-touch programming—convenient pre-programmed transfer methods or create, save, and run customized transfer methods
• Flexible—simultaneous transfer of 1 to 2 mini-size or 1 midi-size gel using the Power Blotter Cassette
• Versatile—compatible with Power Blotter Select Transfer Stacks and Pierce 1-Step Transfer Buffer for rapid blotting programs or Towbin transfer buffer for conventional semi-dry transfer methods
Learn more about the semi-dry transfer device: Power Blotter System
The Power Blotter System achieves highly efficient transfer in seven minutes when used with Power Blotter Select Transfer Stacks or ten minutes with Power Blotter 1-Step Transfer Buffer. The instrument is also effective for standard 30- to 60-minute semi-dry transfer protocols based on traditional buffers.
Related products
Power Blotter XL Welcome Pack
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Novex Bolt gels, NuPage Protein gels
Mode of TransferSemi-Dry
Quantity1 Kit
Shipping ConditionRoom Temperature
MaterialNitrocellulose
Product LinePower Blotter
Unit SizeEach
Contents & Storage
• Power Blotter Station
• Power Blotter Cassette
• Power Blotter Select Transfer Stacks, nitrocellulose, regular-size
• Power Blotter Pre-cut Membranes and Filters, nitrocellulose, regular-size
• Power Blotter 1-Step Transfer Buffer
Frequently asked questions (FAQs)
What is the difference between the Power Blotter System and the Power Blotter XL System?
The Power Blotter System (Cat. No. PB0012) comes with the Power Blotter Station (Cat. No. PB0010) plus Power Blotter Cassette (Cat. No. PB0002). The Power Blotter System can transfer one midi or up to two mini gels at the same time. The Power Blotter XL System (Cat. No. PB0013) on the other hand comes with the Power Blotter Station (Cat. No. PB0010) plus Power Blotter XL Cassette (Cat. No. PB0003). The Power Blotter XL System can transfer up to two midi or four mini gels at the same time.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Is it possible to do gel staining with the Power Blotter System/Power Blotter XL System?
Even though the Power Blotter System/Power Blotter XL System does not have the staining program, this can be introduced. Please see the user guide for our Pierce Mini/Midi Gel Power Staining Kit (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017799_22839_22840_powerstain_midi_minigel_UG.pdf) for how to create a Custom Staining Method.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
What is the material used for the anode and cathode plates of the Power Blotter Cassette/Power Blotter Cassette XL?
The top (cathode) is stainless steel and the bottom (anode) is platinum-coated stainless steel.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
The central European or UK power cord for the Power Blotter System/Power Blotter XL System does not work in Switzerland. Do you offer a Swiss power cord?
Yes, you may purchase the Universal Cable Kit (Cat. No. 4377117).
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
I have lost the power cord for the Power Blotter Station. Is it sold as a stand-alone item?
Here are the SKUs for the power cord. Please order the one appropriate for your region:
- Cat. No. 84858: Power Cord with C13 Connector, North America
- Cat. No. 84859: Power Cord with C13 Connector, Continental Europe and South Korea
- Cat. No. 84860: Power Cord with C13 Connector, United Kingdom and Singapore
- Cat. No. 84855: Chinese 10 Amp Cordset w/ C13 Connector
- Cat. No. 84856: Japanese 15 Amp Cordset w/ C13 Connector
- Cat. No. 84857: Australian 10 Amp Cordset w/ C13 Connector
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.