Pierce™ Premium Grade Sulfo-NHS (N-hydroxysulfosuccinimide)
Thermo Scientific™

Pierce™ Premium Grade Sulfo-NHS (N-hydroxysulfosuccinimide)

Thermo Scientific Pierce Premium Grade Sulfo-NHS is our highest quality formulation of this N-hydroxysuccinimide chemical modification reagent, specially characterized forRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PG8207210 g
PG82071500 mg
Catalog number PG82072
Price (INR)
166,530.00
Online offer
Ends: 31-Mar-2026
256,199.00
Save 89,669.00 (35%)
Each
Quantity:
10 g
Request bulk or custom format
Price (INR)
166,530.00
Online offer
Ends: 31-Mar-2026
256,199.00
Save 89,669.00 (35%)
Each
Thermo Scientific Pierce Premium Grade Sulfo-NHS is our highest quality formulation of this N-hydroxysuccinimide chemical modification reagent, specially characterized for applications where product integrity and risk minimization are paramount.

Features of Premium Grade Sulfo-NHS:

Sulfo-NHS—use with EDC crosslinker to prepare amine-reactive NHS ester reagents
High quality—identity and purity confirmed by several tests, including quantitative NMR
Product integrity—enhanced level of testing and characterization compared to standard grade
Lot retention—ample supply of past lots retained to ensure future process testing
Change management—Change Control Notification (CCN) service
Consistent manufacture—batch-specific manufacturing documentation review

Applications:
• Improve efficiency of EDC coupling reactions
• Convert carboxyls to amine-reactive sulfo-NHS esters
• Membrane-impermable for cell surface labeling
• Crosslink proteins to carboxyl-coated beads or surfaces more efficiently
• Activate nanoparticles with amine-reactive sulfo-NHS esters

Compared to the standard grade product, Premium Grade Sulfo-NHS provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. Sulfo-NHS (N-hydroxysulfosuccinimide) enables control and modification of carbodiimide crosslinking reactions involving activation of carboxylates (—COOH) for conjugation with primary amines (—NH2). Derivatives are easily synthesized by mixing the sulfo-NHS with a carboxyl-containing molecule and a dehydrating agent such as the carbodiimide EDC (EDAC). The method is the basis for generating many types of protein labeling reagents, including amine-reactive fluorescent dyes, biotin affinity tags and pegylation compounds.

Product References:
Crosslinker Application Guide -- search for recent literature references for this product

Related Products
Sulfo-NHS (N-hydroxysulfosuccinimide)
Pierce™ Premium Grade EDC (1-ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityNo
FormPowder
Labeling MethodChemical Labeling
Molecular Weight (g/mol)217.13
PEGylatedNo
Product LinePierce
Quantity10 g
Reactive MoietyCarbodiimide
Shipping ConditionAmbient
SolubilityWater
Water SolubleYes
Chemical ReactivityAmine-Carboxyl
CleavableNo
Crosslinker TypeHeterobifunctional
FormatStandard, Single-use, Premium-grade
Product TypeCrosslinker
SpacerShort (<10 Å)
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C.

Frequently asked questions (FAQs)

Can you provide the shelf-life for Premium Grade Sulfo-NHS (N-hydroxysulfosuccinimide)?

Premium Grade Sulfo-NHS (N-hydroxysulfosuccinimide) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.