Pierce™ Premium Grade Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate)
Pierce™ Premium Grade Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate)
Thermo Scientific™

Pierce™ Premium Grade Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate)

Thermo Scientific Pierce Premium Grade Sulfo-SMCC is our highest quality formulation of amine-to-sulfhydryl crosslinker, specially characterized for applications where productRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PG820861 g
PG82085100 mg
Catalog number PG82086
Price (INR)
256,985.00
Online offer
Ends: 31-Mar-2026
395,361.00
Save 138,376.00 (35%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (INR)
256,985.00
Online offer
Ends: 31-Mar-2026
395,361.00
Save 138,376.00 (35%)
Each
Add to cart
Thermo Scientific Pierce Premium Grade Sulfo-SMCC is our highest quality formulation of amine-to-sulfhydryl crosslinker, specially characterized for applications where product integrity and risk minimization are paramount.

Compared to the standard grade product, Premium Grade Sulfo-SMCC provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. Sulfo-SMCC is a water-soluble, non-cleavable and membrane-impermeable amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane spacer arm (8.3 angstroms). NHS esters react with primary amines at pH 7-9 to form stable amide bonds. Maleimides react with sulfhydryl groups at pH 6.5-7.5 to form stable thioether bonds. The maleimide groups of Sulfo-SMCC are unusually stable up to pH 7.5 because of the cyclohexane bridge in the spacer arm. Because it contains the hydrophilic sulfonyl moiety, Sulfo-SMCC is soluble up to nearly 10 mM in water and many commonly used buffers, thus avoiding the use of organic solvents which may perturb protein structure.

Features of Sulfo-SMCC:

Sulfo-SMCC—a water-soluble amine-to-sulfhydryl crosslinker with a stabilizing cyclohexane spacer arm
High quality—identity and purity confirmed by several tests, including quantitative NMR
Product integrity—enhanced level of testing and characterization compared to standard grade
Lot retention—ample supply of past lots retained to ensure future process testing
Change management—Change Control Notification (CCN) service
Consistent manufacture—batch-specific manufacturing documentation review

Applications:
• Enzyme labelling of antibodies - both enzyme and antibody specificity can be preserved
• Create specific bioconjugates via one- or two-step crosslinking reactions
• Create sulfhydryl-reactive, maleimide-activated carrier proteins for coupling haptens

Product References:
Crosslinker Application Guide -- search for recent literature references for this product

Related Products
Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityNo
FormPowder
Labeling MethodChemical Labeling
Molecular Weight (g/mol)436.37
PEGylatedNo
Product LinePierce
Quantity1 g
Reactive MoietyMaleimide, Sulfo-NHS Ester
Shipping ConditionAmbient
SolubilityWater
Spacer Arm Length8.3 Å
Water SolubleYes
Chemical ReactivityAmine-Sulfhydryl
CleavableNo
Crosslinker TypeHeterobifunctional
FormatStandard, Single-use, Premium-grade
Product TypeCrosslinker
SpacerShort (<10 Å)
Unit SizeEach
Contents & Storage
Upon receipt store desiccated at -20°C.

Frequently asked questions (FAQs)

Can you provide the shelf-life for Premium Grade Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate)?

Premium Grade Sulfo-SMCC (sulfosuccinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Is the cyclohexane ring in Sulfo-SMCC crosslinker cis or trans?

The cyclohexane ring in Sulfo-SMCC crosslinker is trans.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Can Sulfo-SMCC crosslinker reagent be reconstituted in an organic solvent (DMF or DMSO) and added to an aqueous reaction mixture?

Yes, Sulfo-SMCC can be reconstituted in an organic solvent (DMF or DMSO) and added to an aqueous reaction mixture.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Is Sulfo-SMCC crosslinking reagent water soluble?

Yes, Sulfo-SMCC is water soluble.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Can the Sulfo-SMCC cross-linking agent be added directly to the reaction mixture?

Yes, Sulfo-SMCC can be added directly to the reaction mixture.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.