Pierce™ Premium Grade TCEP-HCl
Actual product may vary
Pierce™ Premium Grade TCEP-HCl
Pierce™ Premium Grade TCEP-HCl
Pierce™ Premium Grade TCEP-HCl
Thermo Scientific™
Pierce™ Premium Grade TCEP-HCl
Thermo Scientific Pierce Premium Grade TCEP-HCl is our highest quality formulation of this disulfide reducing agent, specially characterized for applicationsRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PG82090100 g
PG820801 g
PG8208910 g
Catalog number PG82090
Price (INR)
405,841.00
Each
Quantity:
100 g
Request bulk or custom format
Price (INR)
405,841.00
Each
Thermo Scientific Pierce Premium Grade TCEP-HCl is our highest quality formulation of this disulfide reducing agent, specially characterized for applications where product integrity and risk minimization are paramount.
Features of Premium Grade TCEP-HCl:
TCEP—a thiol-free, odorless yet potent reducing agent for cleaving disulfide bonds
High quality—identity and purity confirmed by several tests, including quantitative NMR
Product integrity—enhanced level of testing and characterization compared to standard grade
Lot retention—ample supply of past lots retained to ensure future process testing
Change management—Change Control Notification (CCN) service
Consistent manufacture—batch-specific manufacturing documentation review
Compared to the standard grade product, Premium Grade TCEP-HCl provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. TCEP-HCl is pure, crystalline Tris(2-carboxyethyl)phosphine hydrochloride, a thiol-free compound that is highly effective at reducing protein and peptide disulfide bonds, as well as being odorless and stable. TCEP selectively and completely reduces even the most stable water-soluble alkyl disulfides over a wide pH range.
Advantages of TCEP-HCl over other reducing agents:
Odorless—Unlike DTT or BME, TCEP is odor-free, so reductions can be carried out conveniently on the bench top.
Stable in air—The inherent stability of the TCEP moiety eliminates the need for any special precautions to avoid oxidation when handling, using or storing TCEP.
Efficient—For most applications, 5 to 50 mM TCEP provides sufficient molar excess to effectively reduce peptide or protein disulfide bonds within a few minutes at room temperature.
Compatible—With TCEP, removal of the reducing agent is not necessary prior to most applications, (e.g. histidine-tagged protein purification, maleimide conjugations).
Related Products
Pierce™ TCEP-HCl
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormPowder
Product LinePierce
Quantity100 g
Product TypeDisulfide Reducing Reagent
Unit SizeEach
Contents & Storage
Upon receipt store product at room temperature in sealed container to prevent oxidation. Product is shipped at ambient temperature.
Frequently asked questions (FAQs)
Can other reducing agents other than DTT or BME be used to reduce proteins prior to electrophoresis? For example, what about TCEP (Tris Carboxy Ethyl Phosphene)?
TCEP, Tris Carboxy Ethyl Phosphene is an alternative sulfhydryl reducing agent for protein samples. It is an extremely potent and effective reducing agent for particularly ‘difficult' proteins. It is compatible with the Tris-Glycine gels and NuPAGE gels. It should be added to the sample buffer for these systems. 20 mM final (maximum) concentration is sufficient for samples. You may add alkylating agents, e.g. Iodine (50 mM Iodoacetic acid), to prevent re-forming of S-S bonds but it is not necessary. Do not heat because this will hydrolyze much of your sample. Instead let the sample sit for several minutes at RT and then load.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.