Orion Star™ A222 Conductivity Portable Meter
Thermo Scientific™

Orion Star™ A222 Conductivity Portable Meter

Reliably measure conductivity, TDS, salinity, resistivity and temperature with this conductivity portable meter for advanced field analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProbe Type
STARA22254-cell Conductivity Sensor
STARA2220Not included
Catalog number STARA2225
Price (INR)
-
Probe Type:
4-cell Conductivity Sensor

Record accurate and reliable conductivity, total dissolved solids (TDS), salinity, resistivity and temperature measurements with the Thermo Scientific™ Orion Star™ A222 Conductivity Portable Meter. This instrument is ideal for a wide range of applications and advanced conductivity analysis in the field. Obtain temperature compensated readings using linear, non-linear or EP options with 15°C, 20°C or 25°C reference temperatures. Log up to 1000 data point sets with time/date stamp and transfer via USB or RS232 to a printer or computer. Waterproof meter ensures durable, rugged performance with an IP67-rated housing.

Don't leave your measurements to chance when analyzing samples outside the lab. Orion Star A222 conductivity portable meters are designed to offer the reliability you need in a user-friendly package for your dedicated conductivity testing needs. This waterproof portable meter displays critical data with your measurement including temperature, date and time, read type, measurement stability, active calibration info and power source status.

  • Measure conductivity, TDS, salinity or resistivity with temperature
  • Perform up to a five point conductivity calibration
  • Auto-ranging conductivity values offer consistent readings across entire measurement range without need to recalibrate
  • Use linear, non-linear or off conductivity temperature compensation with 15°C, 20°C or 25°C reference temperatures
  • Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
  • Capture your critical readings with measurement stability indicator and selectable read modes – Auto-Read holds a stable reading and Continuous shows changing readings with hold option
  • Data log collects and date/time stamps up to 1000 measurement sets
  • Non-volatile meter memory ensures your data and settings are protected, even with loss of power
  • Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
  • Flexible power options with AA batteries or universal power adapter
  • IP67-rated housing is waterproof for rugged field operation
  • Meter kits include hard-sided field case and protective meter armor with electrode holders, adjustable wrist/belt strap and built-in meter stand
  • Meters offer CE, TUV 3-in-1 and FCC Class A certifications and 3-year replacement warranty
Specifications
AC AdapterUniversal 50-60 Hz, 100-240 VAC power adapter (sold separately, Cat. No. 1010003)
Accuracy (Conductivity)0.5% of reading ±1 digit > 3 μS; 0.5% of reading ±0.01 μS ≤ 3 μS
Accuracy (Resistivity)0.5% of reading ±1 digit
Accuracy (Salinity)0.5% of reading ±1 digit
Accuracy (TDS)0.5% of reading ±1 digit
Accuracy (Temperature)±0.1°C
Alarm OutputCalibration due alarm (fixed 12 hour)
Backlight OptionYes (Selectable)
Battery Life800 hr.
Battery Type4 x AA (included)
CalibrationConductivity and temperature
Calibration Points (Conductivity)5
Cell Constant0.001 to 199.9cm-1
Certifications/ComplianceCE, TUV 3-1, FCC Class A
Channels1
Compatible Probes2-cell or 4-cell conductivity sensors with built-in temperature
Data Management DetailsTransfer single, range or all data points to printer or computer
DescriptionOrion Star A222 conductivity/TDS/salinity/resistivity portable meter kit with DuraProbe 4-cell conductivity sensor, solutions, meter armor and field case
Dimensions (L x W x H)24 x 10 x 6.5 cm (9.5 x 3.9 x 2.6 in.)
IncludesStar A222 Conductivity Meter with Batteries; 013010MD DuraProbe Epoxy 4-Probe (K=0.475) Conductivity Probe with 3 Meter Cable; 01100710 Conductivity 1413uS Standard Pouches; 911110 Rinse Solution Pouches; STARA-CS Star A Meter Hard Carrying Case; STARA-AR Protective Armor with Electrode Holder
Inputs8 pin MiniDIN (conductivity sensor with built-in temperature)
KeypadComprehensive with menu-specific function keys and dual purpose scroll/shortcut keys
LanguagesEnglish, Spanish, French, Italian, German and Chinese; Portuguese and Korean available with software update
Log Function TypeAutomatic with Auto-Read Measure Mode, Manual with Continuous Measure Mode
MemoryNon-volatile memory preserves data log, calibration log and meter settings
Methods10 per channel with password protection
ModelOrion Star A222 Conductivity/TDS/Salinity/Resistivity/Temperature Portable Meter
OutputsUSB, RS232
Power Supply4 AA batteries (included) or universal AC adapter
Probe Type4-cell Conductivity Sensor
Relative Humidity Range5 to 85%, non-condensing
Resolution (Conductivity)0.001 μS minimum, auto ranging up to 4 significant digits
Resolution (Resistivity)1 ohm or 0.1 megohm, auto ranging
Resolution (Salinity)0.01 psu or 0.01 ppt minimum, auto ranging
Resolution (TDS)0.001 ppm minimum, auto ranging up to 4 significant digits
Resolution (Temperature)0.1°C (0.1°F)
Salinity Measurement ModesPractical salinity (psu)
StabilitySmart Stability and Smart Averaging
TDS Factor RangeLinear (0.02 to 9.99)
Temperature CompensationLinear, nLFn (non-linear ultra pure water) or off
Temperature (English) Operating41°F to 113°F (ambient)
Temperature (Metric) Operating5°C to 45°C (ambient)
Temperature (Metric) Reference15, 20, 25°C
Temperature Probe Calibration1 point temperature offset calibration
Temperature SelectionManual or automatic with ATC temperature probe
TypePortable Conductivity Meter
Warranty3 Year Meter
Weight (English)1.2 lb.
Weight (Metric)0.5 kg
Data Points1000 with Date and Time Stamp
Display TypeGraphic LCD with Backlight
IP RatingIP67
Measurement ModesConductivity, TDS, Salinity, Resistivity with Temperature
Range (Conductivity)0.001 μS/cm to 3000 mS/cm
Range (Resistivity)2 ohm to 100.0 megohm
Range (Salinity)0.06 to 80.00 psu, 0.05 to 42.00 ppt
Range (TDS)0.001 to 200.0 ppt
Temperature Range (Metric)-5°C to +105°C
Unit SizeEach

Frequently asked questions (FAQs)

How long is the warranty period for Orion conductivity meters?

At Thermo Fisher Scientific, we stand behind the quality and reliability of our instruments. That's why we offer a minimum two-year (2) warranty on all Orion benchtop and portable conductivity meters. Obtain peace of mind knowing that your investment is protected. Should any issues arise within the warranty period, our team of experts is committed to providing you with prompt and efficient service. Be sure to check instrument specifications to confirm the warranty coverage period.

How do I perform a calibration using Orion conductivity meters?

Calibration is an important step to obtaining accurate results on your Orion benchtop or portable conductivity meter. Before beginning the calibration process, ensure that you are utilizing freshly poured, non-expired calibration standards to ensure accurate results. Follow the steps below to calibrate your conductivity meter.

• Prepare the calibration standards: Obtain at least one conductivity reference standard with a known conductivity value near the expected sample conductivity range. Ideally, use one standard close to the expected conductivity of your sample, and additional standards for different conductivity ranges. Ensure that the standards are fresh and have not expired.
• Clean the probe: Rinse the probe with distilled or deionized water to remove any debris or contaminants. If necessary, gently wipe the probe with a soft cloth or tissue.
• Turn on the conductivity meter: Power on the conductivity meter and allow it to warm up, if needed. • Immerse the probe: Immerse the probe into a beaker containing the first calibration standard. Ensure that the probe is fully submerged and that it is not touching the sides or bottom of the beaker.
• Allow stabilization: Allow the conductivity reading to stabilize. This may take a few moments, depending on the specific meter and standard being used. The reading should become steady and remain constant.
• Adjust the conductivity value: Using the calibration controls on the conductivity meter, adjust the reading to match the known conductivity value of the calibration standard. Reference the user manual for specific guidance on adjusting the conductivity.
• Rinse the probe: After calibration, rinse the probe with distilled or deionized water to remove any residual calibration solution.
• Repeat calibration: Repeat the calibration steps with the second calibration standard, following the same process as before.
• Verify accuracy: After calibration, it is recommended to check the conductivity meter's accuracy by measuring a conductivity solution of known value. This can help ensure the reliability of your measurements.

To learn more about conductivity calibration, maintenance and beyond, download our user guide: Orion Conductivity Probe User Guide

How do I perform an analysis using Orion conductivity meters?

Before beginning an analysis on your Orion benchtop or portable conductivity meter, assemble your materials. You will need a conductivity probe suitable for your application, calibration standards, and a test sample. Follow the steps below to obtain your conductivity analysis.

• Calibrate the conductivity meter: Start by calibrating your conductivity meter using at least one calibration standard. Typically, you will need at least one standard with a known conductivity value near the expected sample conductivity range. Be sure to reference the user manual for instructions on how to properly calibrate your conductivity meter.
• Prepare your sample: Ensure that your sample is at the desired temperature (most frequently room temperature).
• Immerse the electrode: Dip the conductivity probe into the sample. Avoid touching the sides or bottom of the container to prevent inaccurate readings.
• Allow for stabilization: Give the conductivity meter some time to stabilize and reach a steady reading. This may take a few seconds to a minute depending on your meter and the characteristics of the sample.
• Record the conductivity value: Once the conductivity meter stabilizes, record the conductivity value and temperature value displayed on the meter. Take note of any temperature compensation settings if applicable. • Rinse the probe: After each measurement, rinse the probe with distilled or deionized water to remove any residue or contaminants. This ensures accurate subsequent readings.
• Repeat (if necessary): If you need to analyze multiple samples, repeat the process with each sample, recalibrating the conductivity meter if required.

To learn more about how to get accurate conductivity measurements, download the smart note: Top 10 Mistakes to Avoid When Measuring Conductivity in Liquids.

How do I choose an Orion conductivity probe for my application?

When choosing an Orion conductivity probe for your application there are many factors to consider. Measurement range, sample type, container dimensions, accuracy/ precision requirements, temperature compensation needs, cell constant, durability, maintenance, and meter compatibility are all key features to evaluate.

To learn more about Orion conductivity probes, download the brochure: Orion Conductivity Probes

What supplies do I need to perform an analysis with Orion conductivity meters?

To perform an analysis using Orion benchtop and portable conductivity meters, you will need the following supplies: conductivity probe, conductivity standards, distilled or deionized water, sample containers, and a stirrer probe (optional).