Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • SLC38A3 Antibodies

          Abnova

          SLC38A3 Polyclonal Antibody, MaxPab™

          View all (6) SLC38A3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SLC38A3 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          SLC38A3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SLC38A3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SLC38A3 Antibody (H00010991-B01P) in WB

          Western Blot analysis of SLC38A3 expression in transfected 293T cell line (H00010991-T01) by SLC38A3 MaxPab polyclonal antibody.

          Lane 1: SLC38A3 transfected lysate(55.44 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SLC38A3 Antibody in Western Blot (WB)
          SLC38A3 Antibody in Western Blot (WB)
          SLC38A3 Antibody in Western Blot (WB)
          SLC38A3 Polyclonal Antibody, MaxPab™

          Product Details

          H00010991-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SLC38A3 (NP_006832.1, 1 a.a. approximately 504 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MEAPLQTEMV ELVPNGKHSE GLLPVITPMA GNQRVEDPAR SCMEGKSFLQ KSPSKEPHFT DFEGKTSFGM SVFNLSNAIM GSGILGLAYA MANTGIILFL FLLTAVALLS SYSIHLLLKS SGVVGIRAYE QLGYRAFGTP GKLAAALAIT LQNIGAMSSY LYIIKSELPL VIQTFLNLEE KTSDWYMNGN YLVILVSVTI ILPLALMRQL GYLGYSSGFS LSCMVFFLIA VIYKKFHVPC PLPPNFNNTT GNFSHVEIVK EKVQLQVEPE ASAFCTPSYF TLNSQTAYTI PIMAFAFVCH PEVLPIYTEL KDPSKKKMQH ISNLSIAVMY IMYFLAALFG YLTFYNGVES ELLHTYSKVD PFDVLILCVR VAVLTAVTLT VPIVLFPVRR AIQQMLFPNQ EFSWLRHVLI AVGLLTCINL LVIFAPNILG IFGVIGATSA PFLIFIFPAI FYFRIMPTEK EPARSTPKIL ALCFAMLGFL LMTMSLSFII IDWASGTSRH GGNH

          Target Information

          As a sodium-dependent amino acid/proton antiporter, SLC38A3 mediates electrogenic cotransport of glutamine and sodium ions in exchange for protons. It also recognizes histidine, asparagine and alanine. It may mediate amino acid transport in either direction under physiological conditions and play a role in nitrogen metabolism and synaptic transmission.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: hSN1; N-system amino acid transporter 1; Na(+)-coupled neutral amino acid transporter 3; SLC38A3; Sodium-coupled neutral amino acid transporter 3; Solute carrier family 38 member 3; System N amino acid transporter 1; system N1 Na+ and H+-coupled glutamine transporter; unnamed protein product

          View more View less

          Gene Aliases: DEE102; G17; NAT1; SLC38A3; SN1; SNAT3

          View more View less

          UniProt ID: (Human) Q99624

          View more View less

          Entrez Gene ID: (Human) 10991

          View more View less

          Function(s)
          L-histidine transmembrane transporter activity neutral amino acid:sodium symporter activity amino acid transmembrane transporter activity L-alanine transmembrane transporter activity L-asparagine transmembrane transporter activity L-glutamine transmembrane transporter activity symporter activity antiporter activity L-glutamine, sodium:proton antiporter activity L-asparagine, sodium:proton antiporter activity L-histidine, sodium:proton antiporter activity
          Process(es)
          amino acid transmembrane transport ion transport sodium ion transport amino acid transport asparagine transport glutamine transport female pregnancy glutamine secretion L-alanine transport histidine transport positive regulation of transcription from RNA polymerase II promoter cellular response to potassium ion starvation amino acid homeostasis transport across blood-brain barrier L-histidine transport L-glutamine import across plasma membrane L-histidine import across plasma membrane L-asparagine import across plasma membrane positive regulation of glutamine transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline