Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • ORP8 Antibodies

          Abnova

          OSBPL8 Polyclonal Antibody, MaxPab™

          View all (9) ORP8 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite OSBPL8 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          OSBPL8 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          OSBPL8 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          OSBPL8 Antibody (H00114882-B01P) in WB

          Western Blot analysis of OSBPL8 expression in transfected 293T cell line by OSBPL8 MaxPab polyclonal antibody. Lane 1: OSBPL8 transfected lysate(93.17 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          OSBPL8 Antibody in Western Blot (WB)
          OSBPL8 Polyclonal Antibody, MaxPab™

          Product Details

          H00114882-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          OSBPL8 (NP_001003712.1, 1 a.a. approximately 847 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSQRQGKEAY PTPTKDLHQP SLSPASPHSQ GFERGKEDIS QNKDESSLSM SKSKSESKLY NGSEKDSSTS SKLTKKESLK VQKKNYREEK KRATKELLST ITDPSVIVMA DWLKIRGTLK SWTKLWCVLK PGVLLIYKTQ KNGQWVGTVL LNACEIIERP SKKDGFCFKL FHPLEQSIWA VKGPKGEAVG SITQPLPSSY LIIRATSESD GRCWMDALEL ALKCSSLLKR TMIREGKEHD LSVSSDSTHV TFYGLLRANN LHSGDNFQLN DSEIERQHFK DQDMYSDKSD KENDQEHDES DNEVMGKSEE SDTDTSERQD DSYIEPEPVE PLKETTYTEQ SHEELGEAGE ASQTETVSEE NKSLIWTLLK QVRPGMDLSK VVLPTFILEP RSFLDKLSDY YYHADFLSEA ALEENPYFRL KKVVKWYLSG FYKKPKGLKK PYNPILGETF RCLWIHPRTN SKTFYIAEQV SHHPPISAFY VSNRKDGFCL SGSILAKSKF YGNSLSAILE GEARLTFLNR GEDYVMTMPY AHCKGILYGT MTLELGGTVN ITCQKTGYSA ILEFKLKPFL GSSDCVNQIS GKLKLGKEVL ATLEGHWDSE VFITDKKTDN SEVFWNPTPD IKQWRLIRHT VKFEEQGDFE SEKLWQRVTR AINAKDQTEA TQEKYVLEEA QRQAARDRKT KNEEWSCKLF ELDPLTGEWH YKFADTRPWD PLNDMIQFEK DGVIQTKVKH RTPMVSVPKM KHKPTRQQKK VAKGYSSPEP DIQDSSGSEA QSVKPSTRRK KGIELGDIQS SIESIKQTQE EIKRNIMALR NHLVSSTPAT DYFLQQKDYF IIFLLILLQV IINFMFK

          Target Information

          This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Two transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ORP-8; OSBP-related protein 8; OSBP10; Oxysterol-binding protein-related protein 8; unnamed protein product

          View more View less

          Gene Aliases: KIAA1451; MST120; MSTP120; ORP8; OSBP10; OSBPL8

          View more View less

          UniProt ID: (Human) Q9BZF1

          View more View less

          Entrez Gene ID: (Human) 114882

          View more View less

          Function(s)
          phosphatidylserine binding protein binding phospholipid transporter activity lipid binding cholesterol binding phosphatidylinositol-4-phosphate binding phosphatidylserine transfer activity
          Process(es)
          lipid transport phospholipid transport negative regulation of cell migration phosphatidylserine acyl-chain remodeling fat cell differentiation positive regulation of glucose import positive regulation of insulin receptor signaling pathway positive regulation of protein kinase B signaling triglyceride homeostasis protein localization to nuclear pore intermembrane lipid transfer
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline