Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • DRD2 Antibodies

          Alomone Labs, LTD

          D2 Dopamine Receptor (extracellular) Polyclonal Antibody

          View all (28) DRD2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite D2 Dopamine Receptor (extracellular) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          D2 Dopamine Receptor (extracellular) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          D2 Dopamine Receptor (extracellular) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          D2 Dopamine Receptor (extracellular) Antibody (ADR-002-200UL) in WB

          Western blot analysis of rat striatum (lanes 1 and 5) and hippocampus (lanes 2 and 6) membranes and of rat (lanes 3 and 7) and mouse (lanes 4 and 8) whole brain lysates: - 1-4. Anti-D2Dopamine Receptor (extracellular) Antibody (#ADR-002), (1:200).5-8. Anti-D2Dopamine Receptor (extracellular) Antibody , preincubated with D2 Dopamine Receptor (extracellular) Blocking Peptide (#BLP-DR002). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          D2 Dopamine Receptor (extracellular) Antibody in Western Blot (WB)
          D2 Dopamine Receptor (extracellular) Antibody in Flow Cytometry (Flow)
          D2 Dopamine Receptor (extracellular) Polyclonal Antibody

          Product Details

          ADR-002-200UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200
          -

          Flow Cytometry (Flow)

          5 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          (C)DDLERQNWSRPFNGSE, corresponding to amino acid residues 11-26 of rat DRD2, Extracellular, N-terminus
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.8 mg/mL

          Amount

          160 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 1% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).

          Target Information

          The DRD2 Dopamine Receptor is involved in the neuromodulation of appetitive behaviors. DRD2 may be linked to alcoholism and other substance-use disorders. Mutations in DRD2 in mice lead to the suppression of reward behavior with morphine and to the reduction of spontaneous movements, a phenotype resembling Parkinsons disease. Three alternatively spliced isoforms have been identified. DRD2 has been reported to be expressed in various regions of the brain, as well as in aorta, artery, eye, and spinal cord. ESTs have been isolated from normal pituitary and in brain, colon, lung, pancreas, and skeletal muscle cancer libraries.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: D(2) dopamine receptor; D2 receptor; Dopamine 2 receptor; Dopamine D2 receptor; dopamine receptor 2; seven transmembrane helix receptor; unnamed protein product

          View more View less

          Gene Aliases: D2DR; D2R; Drd-2; DRD2

          View more View less

          UniProt ID: (Human) P14416, (Mouse) P61168, (Rat) P61169

          View more View less

          Entrez Gene ID: (Human) 1813, (Mouse) 13489, (Rat) 24318

          View more View less

          Function(s)
          dopamine neurotransmitter receptor activity, coupled via Gi/Go G-protein alpha-subunit binding G-protein coupled receptor activity dopamine neurotransmitter receptor activity receptor binding protein binding potassium channel regulator activity heterotrimeric G-protein binding dopamine binding ionotropic glutamate receptor binding identical protein binding macromolecular complex binding signal transducer activity drug binding protein homodimerization activity protein heterodimerization activity G-protein coupled receptor
          Process(es)
          temperature homeostasis response to hypoxia neuron migration synaptic transmission, dopaminergic startle response response to amphetamine neurological system process involved in regulation of systemic arterial blood pressure regulation of heart rate regulation of sodium ion transport G-protein coupled receptor internalization positive regulation of neuroblast proliferation positive regulation of receptor internalization cellular calcium ion homeostasis autophagy signal transduction G-protein coupled receptor signaling pathway adenylate cyclase-modulating G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting dopamine receptor signaling pathway phospholipase C-activating G-protein coupled receptor signaling pathway dopamine receptor signaling pathway neuron-neuron synaptic transmission neuroblast proliferation axonogenesis synapse assembly sensory perception of smell long-term memory grooming behavior locomotory behavior adult walking behavior feeding behavior protein localization cell proliferation negative regulation of cell proliferation associative learning visual learning response to xenobiotic stimulus response to light stimulus response to toxic substance response to iron ion regulation of dopamine secretion response to inactivity Wnt signaling pathway cell migration striatum development orbitofrontal cortex development cerebral cortex GABAergic interneuron migration adenohypophysis development negative regulation of cell migration peristalsis adult behavior forebrain development auditory behavior regulation of synaptic transmission, GABAergic response to estradiol positive regulation of cytokinesis circadian regulation of gene expression negative regulation of dopamine secretion response to histamine response to nicotine negative regulation of dephosphorylation intracellular signal transduction positive regulation of urine volume positive regulation of renal sodium excretion positive regulation of multicellular organism growth response to cocaine negative regulation of circadian sleep/wake cycle, sleep dopamine metabolic process drinking behavior regulation of potassium ion transport response to morphine regulation of MAPK cascade pigmentation protein kinase B signaling response to ethanol positive regulation of G-protein coupled receptor protein signaling pathway negative regulation of blood pressure negative regulation of innate immune response positive regulation of transcription from RNA polymerase II promoter negative regulation of insulin secretion acid secretion behavioral response to cocaine behavioral response to ethanol regulation of long-term neuronal synaptic plasticity response to axon injury branching morphogenesis of a nerve arachidonic acid secretion epithelial cell proliferation negative regulation of epithelial cell proliferation negative regulation of protein secretion positive regulation of neurogenesis modulation of synaptic transmission negative regulation of synaptic transmission release of sequestered calcium ion into cytosol positive regulation of multicellular organismal process negative regulation of cytosolic calcium ion concentration regulation of neurotransmitter uptake dopamine uptake involved in synaptic transmission regulation of dopamine uptake involved in synaptic transmission positive regulation of dopamine uptake involved in synaptic transmission regulation of synapse structural plasticity negative regulation of protein kinase B signaling regulation of amine transport negative regulation of synaptic transmission, glutamatergic excitatory postsynaptic potential positive regulation of growth hormone secretion prepulse inhibition phospholipase C-activating dopamine receptor signaling pathway negative regulation of dopamine receptor signaling pathway positive regulation of ERK1 and ERK2 cascade cellular response to retinoic acid cellular response to ethanol regulation of locomotion involved in locomotory behavior postsynaptic modulation of chemical synaptic transmission presynaptic modulation of chemical synaptic transmission beta-arrestin-dependent dopamine receptor signaling pathway negative regulation of cellular response to hypoxia positive regulation of glial cell-derived neurotrophic factor secretion positive regulation of long-term synaptic potentiation regulation of secretion by cell hyaloid vascular plexus regression negative regulation of neuron migration negative regulation of adenylate cyclase activity chemical synaptic transmission regulation of cAMP metabolic process activation of protein kinase activity response to drug regulation of phosphoprotein phosphatase activity phosphatidylinositol metabolic process positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway negative regulation of voltage-gated calcium channel activity cellular potassium ion homeostasis positive regulation of mitotic nuclear division
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline