Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • GnRH1 Antibodies

          Invitrogen

          GnRH1 Polyclonal Antibody

          21 Published Figures
          25 References
          View all (12) GnRH1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite GnRH1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Published Figures (21)
          GnRH1 Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          GnRH1 Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 23

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GnRH1 Antibody (PA1-121) in IHC

          Immunolocalization of GnRH in mouse embryo brain using GnRH Polyclonal Antibody (Product # PA1-121). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GnRH1 Antibody in Immunohistochemistry (IHC)
          GnRH1 Antibody in Immunohistochemistry (IHC)
          GnRH1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          GnRH1 Antibody in Immunocytochemistry (ICC/IF)
          GnRH1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          GnRH1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          GnRH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GnRH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GnRH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GnRH1 Antibody in Immunohistochemistry (IHC)
          GnRH1 Antibody in Immunohistochemistry (IHC)
          GnRH1 Antibody in Immunohistochemistry (IHC)
          GnRH1 Antibody in Immunohistochemistry (IHC)
          GnRH1 Antibody in Immunohistochemistry (IHC)
          GnRH1 Antibody in Immunohistochemistry (IHC)
          GnRH1 Antibody in Immunohistochemistry (IHC)
          GnRH1 Antibody in Immunohistochemistry (IHC)
          GnRH1 Antibody in Immunohistochemistry (IHC)
          GnRH1 Antibody in Immunohistochemistry (IHC)
          GnRH1 Antibody in Immunohistochemistry (IHC)
          GnRH1 Antibody in Immunohistochemistry (IHC)
          GnRH1 Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          GnRH1 Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          GnRH1 Polyclonal Antibody

          Product Details

          PA1-121

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 16 publications 16 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:1,000
          View 3 publications 3 publications

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 3 publications 3 publications

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Amphibian, Mouse

          Published species

          Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues pyroE(22) H W S Y G L R P G-NH2(32) of mouse GnRH.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_325077

          Product Specific Information

          PA1-121 detects gonadotropin releasing hormone (GnRH) in mouse brain and in amphibian (Hyla cinerea) samples.

          PA1-121 has been successfully used in immunohistochemistry procedures. Immunohistochemical staining of GnRH in mouse embryo brain tissue using PA1-121 results in staining of neurons in the organum vasculosum of the lamina terminalis (OVLT).

          In immunohistochemical procedures, 4% paraformaldehyde is recommended as a fixative.

          The PA1-121 immunogen is a synthetic peptide corresponding to residues pyroE(22) H W S Y G L R P G-NH2(32) of mouse GnRH. This sequence is completely conserved in rat and tree shrew and is 78% conserved in zebrafish. The PA1-121 immunizing peptide (Cat. # PEP-168) is available for use in neutralization and control experiments.

          Target Information

          The Luteinizing Hormone Releasing Hormone (LH-RH) is produced in the hypothalamus and influences the release of Luteinizing Hormone in the pituitary gland. LH-RH is also known as Gonadotropin-releasing hormone (Gn-RH).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: gonadotropin releasing hormone 2; hypogonadal; luteinizing hormone releasing hormone; luteinizing hormone-releasing hormone I; Lutropin; Progonadoliberin I; Progonadoliberin-1

          View more View less

          Gene Aliases: Gnrh; Gnrh1; Gnrh2; hpg; LHRH; Lhrh1; Lnrh

          View more View less

          UniProt ID: (Mouse) P13562

          View more View less

          Entrez Gene ID: (Mouse) 14714

          View more View less

          Function(s)
          hormone activity gonadotropin hormone-releasing hormone activity peptide hormone intercellular signal molecule
          Process(es)
          multicellular organism development regulation of gene expression male sex determination negative regulation of immature T cell proliferation negative regulation of apoptotic process response to ethanol response to steroid hormone regulation of ovarian follicle development negative regulation of neuron migration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline