Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • alpha Actinin 1/2/3/4 Antibodies

          Invitrogen

          alpha Actinin 1/2/3/4 Polyclonal Antibody

          View all (3) alpha Actinin 1/2/3/4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite alpha Actinin 1/2/3/4 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          alpha Actinin 1/2/3/4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          alpha Actinin 1/2/3/4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          alpha Actinin 1/2/3/4 Antibody (PA5-103181) in WB

          Western blot analysis of alpha Actinin 1/2/3/4 in COLO205 cells lysate. Samples were incubated with alpha Actinin 1/2/3/4 polyclonal antibody (Product # PA5-103181). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          alpha Actinin 1/2/3/4 Antibody in Western Blot (WB)
          alpha Actinin 1/2/3/4 Polyclonal Antibody

          Product Details

          PA5-103181

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:3,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human ACTN1(Accession P12814), corresponding to amino acid residues Q167-D217.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2815957

          Product Specific Information

          Antibody detects endogenous levels of total ACTN1/2/3/4.

          Target Information

          Alpha actinins belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments. This gene encodes a nonmuscle, cytoskeletal, alpha actinin isoform and maps to the same site as the structurally similar erythroid beta spectrin gene. Three transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: actinin 1 smooth muscle; alpha actinin 1a; alpha actinin 4; Alpha-actinin cytoskeletal isoform; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 2; Alpha-actinin skeletal muscle isoform 3; Alpha-actinin-1; Alpha-actinin-2; Alpha-actinin-3; Alpha-actinin-4; epididymis secretory sperm binding protein; F-actin cross-linking protein; focal segmental glomerulosclerosis 1; non-muscle alpha-actinin 1; Non-muscle alpha-actinin 4; Non-muscle alpha-actinin-1; transcriptional coactivator; unnamed protein product

          View more View less

          Gene Aliases: 1110008F24Rik; 3110023F10Rik; ACTININ-4; ACTN1; Actn1a; ACTN2; ACTN3; ACTN3D; ACTN4; BDPLT15; C77391; CMD1AA; CMH23; CMYO8; CMYP8; FSGS; FSGS1; MPD6; MYOCOZ

          View more View less

          UniProt ID: (Human) P12814, (Human) P35609, (Human) Q08043, (Human) O43707, (Rat) Q9Z1P2, (Mouse) Q7TPR4, (Mouse) Q9JI91, (Mouse) O88990, (Mouse) P57780, (Rat) Q9QXQ0

          View more View less

          Entrez Gene ID: (Human) 87, (Human) 88, (Human) 89, (Human) 81, (Rat) 81634, (Mouse) 109711, (Mouse) 11472, (Rat) 291245, (Mouse) 11474, (Rat) 171009, (Mouse) 60595, (Rat) 63836

          View more View less

          Function(s)
          transcription coactivator activity double-stranded RNA binding actin binding integrin binding calcium ion binding protein binding cytoskeletal protein binding vinculin binding identical protein binding protein homodimerization activity ion channel binding metal ion binding actin filament binding structural constituent of postsynapse phosphatidylinositol-4,5-bisphosphate binding structural constituent of muscle protein domain specific binding LIM domain binding titin binding FATZ binding titin Z domain binding structural constituent of postsynaptic actin cytoskeleton channel activator activity RNA polymerase II regulatory region sequence-specific DNA binding nucleoside binding RNA binding ligand-dependent nuclear receptor binding chromatin DNA binding retinoic acid receptor binding ligand-dependent nuclear receptor transcription coactivator activity thyroid hormone receptor coactivator activity protein binding, bridging protein dimerization activity protein complex binding nuclear hormone receptor binding poly(A) RNA binding protein N-terminus binding
          Process(es)
          regulation of the force of skeletal muscle contraction skeletal muscle atrophy transition between fast and slow fiber response to denervation involved in regulation of muscle adaptation actin cytoskeleton organization positive regulation of fast-twitch skeletal muscle fiber contraction regulation of apoptotic process negative regulation of glycolytic process focal adhesion assembly positive regulation of skeletal muscle tissue growth positive regulation of skeletal muscle fiber development muscle cell development bone morphogenesis negative regulation of calcineurin-NFAT signaling cascade regulation of muscle system process negative regulation of oxidative phosphorylation negative regulation of cold-induced thermogenesis positive regulation of bone mineralization involved in bone maturation negative regulation of relaxation of muscle regulation of aerobic respiration positive regulation of glucose catabolic process to lactate via pyruvate actin filament bundle assembly negative regulation of cellular component movement actin crosslink formation regulation of nucleic acid-templated transcription cortical cytoskeleton organization muscle contraction microspike assembly regulation of membrane potential negative regulation of potassium ion transport positive regulation of potassium ion transport protein homotetramerization negative regulation of potassium ion transmembrane transporter activity positive regulation of potassium ion transmembrane transporter activity negative regulation of protein localization to cell surface positive regulation of receptor activity transport protein transport peroxisome proliferator activated receptor signaling pathway retinoic acid receptor signaling pathway positive regulation of pinocytosis positive regulation of cellular component movement bicellular tight junction assembly protein localization to bicellular tight junction cell adhesion sarcomere organization positive regulation of transcription, DNA-templated anatomical structure formation involved in morphogenesis actin filament uncapping cardiac muscle cell development protein localization to plasma membrane phospholipase C-activating angiotensin-activated signaling pathway postsynaptic actin cytoskeleton organization positive regulation of endocytic recycling response to hypoxia vesicle transport along actin filament positive regulation of cell migration positive regulation of sodium:proton antiporter activity tumor necrosis factor-mediated signaling pathway positive regulation of transcription from RNA polymerase II promoter negative regulation of substrate adhesion-dependent cell spreading positive regulation of NIK/NF-kappaB signaling actin filament organization platelet activation platelet formation platelet morphogenesis actin filament network formation platelet aggregation postsynapse organization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline