Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 475132_mat
          Assay Name: hsa-miR-181b-2-3p
          miRBase Accession Number: MI0000683
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 124693710 - 124693798 [+] on Build GRCh38
          Mature miRNA Sequence: CUCACUGAUCAAUGAAUGCA
          Species: Human, Chrysemys picta
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 475132_mat
          Size
          Availability Made To Order
          Catalog # 4440886
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000007, mir-181

          Mature miRNA Sequence:

          CUCACUGAUCAAUGAAUGCA

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-181b-2-3p MIMAT0031893
          Chrysemys picta cpi-miR-181b-3p MIMAT0037797

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-181b-2
          Stem-loop Accession # MI0000683
          Stem-loop Sequence
          CUGAUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUGAGUCUGAAUCAACUCACUGAUCAAUGAAUGCAAACUGCGGACCAAACA
          Chromosome Location Chr. 9 - 124693710 - 124693798 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0031893
          miRBase ID: hsa-miR-181b-2-3p
          Mature miRNA Sequence: CUCACUGAUCAAUGAAUGCA
          Chromosome Location: Chr. 9 - 124693710 - 124693798 [+] on Build GRCh38

          Chrysemys picta

          Stem-loop ID cpi-mir-181b-2
          Stem-loop Accession # MI0029504
          Stem-loop Sequence
          GCAUCUAAUGGCUGCAAUCAACAUUCAUUGCUGUCGGUGGGUUUUAGUUUGUAUCAACUCACUGAUCAAUGAAUGCAAACUGCGGACCAGGUAGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037797
          miRBase ID: cpi-miR-181b-3p
          Mature miRNA Sequence: CUCACUGAUCAAUGAAUGCA
          Chromosome Location: NA

          Stem-loop ID cpi-mir-181b-3
          Stem-loop Accession # MI0029505
          Stem-loop Sequence
          CGAAAAGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGUGUUGUUGAAGGAAAAACUCACUGAUCAAUGAAUGCAACUGUGAUUGGAAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037797
          miRBase ID: cpi-miR-181b-3p
          Mature miRNA Sequence: CUCACUGAUCAAUGAAUGCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303356_pri
          mirVana® miRNA inhibitor : MH33176
          mirVana® miRNA mimic : MC33176

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0