Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 479205_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-92a-1-5p
          Stem-loop Accession Number: MI0000093
          miRBase Version: v22.1
          Chromosome Location: Chr.13: 91351314 - 91351391 [+] on Build GRCh38
          Mature miRNA Sequence: AGGUUGGGAUCGGUUGCAAUGCU
          Species: Human, Guinea pig, Ophiophagus hannah, Pteropus alecto
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 479205_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000013, mir-25

          Mature miRNA Sequence:

          AGGUUGGGAUCGGUUGCAAUGCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-92a-1-5p MIMAT0004507
          Guinea pig cpo-miR-92a-5p MIMAT0046928
          Ophiophagus hannah oha-miR-92a-5p MIMAT0036958
          Pteropus alecto pal-miR-92a-2-5p MIMAT0039838

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-92a-1
          Stem-loop Accession # MI0000093
          Stem-loop Sequence
          CUUUCUACACAGGUUGGGAUCGGUUGCAAUGCUGUGUUUCUGUAUGGUAUUGCACUUGUCCCGGCCUGUUGAGUUUGG
          Chromosome Location Chr. 13 - 91351314 - 91351391 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004507
          miRBase ID: hsa-miR-92a-1-5p
          Mature miRNA Sequence: AGGUUGGGAUCGGUUGCAAUGCU
          Chromosome Location: Chr. 13 - 91351314 - 91351391 [+] on Build GRCh38

          Guinea pig

          Stem-loop ID cpo-mir-92a-1
          Stem-loop Accession # MI0038603
          Stem-loop Sequence
          AGGUUGGGAUCGGUUGCAAUGCUGUGUUUCUGUAUGGUAUUGCACUUGUCCCGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046928
          miRBase ID: cpo-miR-92a-5p
          Mature miRNA Sequence: AGGUUGGGAUCGGUUGCAAUGCU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-92a-1
          Stem-loop Accession # MI0031491
          Stem-loop Sequence
          ACCCCUUUCUACACAGGUUGGGAUCGGUUGCAAUGCUGUGUUAAAUCAGUGGUAUUGCACUUGUCCCGGCCUGUUGAGGUUGGUGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036958
          miRBase ID: oha-miR-92a-5p
          Mature miRNA Sequence: AGGUUGGGAUCGGUUGCAAUGCU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-92a-2
          Stem-loop Accession # MI0032355
          Stem-loop Sequence
          AGGUUGGGAUCGGUUGCAAUGCUGUGUUUCUGUAUGGUAUUGCACUUGUCCCGGCCUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039838
          miRBase ID: pal-miR-92a-2-5p
          Mature miRNA Sequence: AGGUUGGGAUCGGUUGCAAUGCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302603_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12696
          Ambion® Pre-miR™ miRNA Precursor : PM12696
          TaqMan™ MicroRNA Assay : 002137
          mirVana® miRNA inhibitor : MH12696
          mirVana® miRNA mimic : MC12696
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 2
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human B Card
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 2
          FGMIR : TaqManTM Advanced miRNA Human B Card
          MIRNA : TaqMan® Advanced miRNA Human B 96-well Plates, Standard

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline