Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM10073
          Assay Name: mmu-miR-24-1-5p
          miRBase Accession Number: MI0000231
          miRBase Version: v22.1
          Chromosome Location: Chr.13: 63301208 - 63301275 [+] on Build GRCm38
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Species: Mouse, Bovine, Chicken, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Macaca nemestrina, Ophiophagus hannah, Pan paniscus, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey, Taeniopygia guttata
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM10073
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000041, mir-24

          Mature miRNA Sequence:

          GUGCCUACUGAGCUGAUAUCAGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-24-1-5p MIMAT0000218
          Bovine bta-miR-24 MIMAT0009250
          Chicken gga-miR-24-5p MIMAT0026548
          Cricetulus griseus cgr-miR-24 MIMAT0041158
          Dasypus novemcinctus dno-miR-24a-5p MIMAT0047532
          Guinea pig cpo-miR-24-5p MIMAT0046880
          View More Rows
          Macaca nemestrina mne-miR-24-5p MIMAT0002340
          Ophiophagus hannah oha-miR-24-1-5p MIMAT0036858
          Pan paniscus ppa-miR-24-5p MIMAT0002342
          Pig ssc-miR-24-2-5p MIMAT0026468
          Pongo pygmaeus ppy-miR-24-5p MIMAT0002338
          Rabbit ocu-miR-24-5p MIMAT0048130
          Rhesus monkey mml-miR-24-5p MIMAT0002336
          Taeniopygia guttata tgu-miR-24-5p MIMAT0026994

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-24-1
          Stem-loop Accession # MI0000231
          Stem-loop Sequence
          CUCCGGUGCCUACUGAGCUGAUAUCAGUUCUCAUUUCACACACUGGCUCAGUUCAGCAGGAACAGGAG
          Chromosome Location Chr. 13 - 63301208 - 63301275 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000218
          miRBase ID: mmu-miR-24-1-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: Chr. 13 - 63301208 - 63301275 [+] on Build GRCm38

          Bovine

          Stem-loop ID bta-mir-24-1
          Stem-loop Accession # MI0009761
          Stem-loop Sequence
          CUCCGGUGCCUACUGAGCUGAUAUCAGUUCUCAUUUUACACACUGGCUCAGUUCAGCAGGAACAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009250
          miRBase ID: bta-miR-24
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-24
          Stem-loop Accession # MI0001275
          Stem-loop Sequence
          CUCCGGUGCCUACUGAGCUGAUAUCAGUUCUGAUUUUACAUACUGGCUCAGUUCAGCAGGAACAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026548
          miRBase ID: gga-miR-24-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-24-1
          Stem-loop Accession # MI0033087
          Stem-loop Sequence
          CUCCGGUGCCUACUGAGCUGAUAUCAGUUCUCAUUUCACACACUGGCUCAGUUCAGCAGGAACAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041158
          miRBase ID: cgr-miR-24
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-24a
          Stem-loop Accession # MI0038921
          Stem-loop Sequence
          GUGCCUACUGAGCUGAUAUCAGUUCUCAUUUCACACACUGGCUCAGUUCAGCAGGAACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047532
          miRBase ID: dno-miR-24a-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-24-1
          Stem-loop Accession # MI0038577
          Stem-loop Sequence
          GUGCCUACUGAGCUGAUAUCAGUUCUCUUGUACACACUGGCUCAGUUCAGCAGGAACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046880
          miRBase ID: cpo-miR-24-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-24-1
          Stem-loop Accession # MI0002639
          Stem-loop Sequence
          CUCCGGUGCCUACUGAGCUGAUAUCAGUUCUCAUUUUACACACUGGCUCAGUUCAGCAGGAACAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002340
          miRBase ID: mne-miR-24-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-24-1
          Stem-loop Accession # MI0031432
          Stem-loop Sequence
          CGUCCUCCGGUGCCUACUGAGCUGAUAUCAGUUCUGAUUUUAUAUACUGGCUCAGUUCAGCAGGAACAGGAGUCGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036858
          miRBase ID: oha-miR-24-1-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-24-1
          Stem-loop Accession # MI0002640
          Stem-loop Sequence
          CUCCGGUGCCUACUGAGCUGAUAUCAGUUCUCAUUUUACACACUGGCUCAGUUCAGCAGGAACAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002342
          miRBase ID: ppa-miR-24-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-24-2
          Stem-loop Accession # MI0013111
          Stem-loop Sequence
          GACCCGCCCUCCGGUGCCUACUGAGCUGAUAUCAGUUCUCAUUUUACACACUGGCUCAGUUCAGCAGGAACAGGAGUCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026468
          miRBase ID: ssc-miR-24-2-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-24-1
          Stem-loop Accession # MI0002638
          Stem-loop Sequence
          CUCCGGUGCCUACUGAGCUGAUAUCAGUUCUCAUUUUACACACUGGCUCAGUUCAGCAGGAACAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002338
          miRBase ID: ppy-miR-24-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-24-1
          Stem-loop Accession # MI0039236
          Stem-loop Sequence
          GUGCCUACUGAGCUGAUAUCAGUUCUCGUUUCACACACUGGCUCAGUUCAGCAGGAACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048130
          miRBase ID: ocu-miR-24-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-24-1
          Stem-loop Accession # MI0002637
          Stem-loop Sequence
          CUCCGGUGCCUACUGAGCUGAUAUCAGUUCUCAUUUUACACACUGGCUCAGUUCAGCAGGAACAGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002336
          miRBase ID: mml-miR-24-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-24
          Stem-loop Accession # MI0013753
          Stem-loop Sequence
          CCAGUCGGAAACAACUUACAGCUGCUGCUGUUGUUCCAGGCUGUUGAUGGACCCGUCCUCCGGUGCCUACUGAGCUGAUAUCAGUUCUGAUUUUACACACUGGCUCAGUUCAGCAGGAACAGGAGUCGAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026994
          miRBase ID: tgu-miR-24-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306468_pri
          Ambion® Pre-miR™ miRNA Precursor : PM10073
          TaqMan™ MicroRNA Assay : 000488
          mirVana® miRNA inhibitor : MH10073
          mirVana® miRNA mimic : MC10073
          TaqMan™ Advanced miRNA Assay : mmu482715_mir


          miRBase Alias:

          Human: hsa-miR-189 (v9.2)
          Mouse: mmu-miR-24-1* (v17)
          Mouse: mmu-miR-189 (v9)
          Mouse: mmu-miR-24* (v9.2)
          Rat: rno-miR-189 (v9.2)
          Macaca mulatta: mml-miR-24* (v18)
          Macaca mulatta: mml-miR-189 (v12)
          Macaca nemestrina: mne-miR-189 (v12)
          Macaca nemestrina: mne-miR-24* (v18)
          Pectobacterium parvum: ppa-miR-189 (v12)
          Pectobacterium parvum: ppa-miR-24* (v18)
          Pongo pygmaeus: ppy-miR-189 (v12)
          Pongo pygmaeus: ppy-miR-24* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline