Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03306468_pri
          Assay Name:
          mmu-mir-3074-1
          Stem-loop ID:
          mmu-mir-24-1 mmu-mir-27b mmu-mir-3074-1
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.13: 63301208 - 63301275 [+] on Build GRCm38
          Stem-loop Location:
          Chr.13: 63301208 - 63301275 [+] on Build GRCm38 Chr.13: 63300712 - 63300784 [+] on Build GRCm38 Chr.13: 63301199 - 63301283 [-] on Build GRCm38
          Mature miRNA ID:
          mmu-miR-24-3p mmu-miR-24-1-5p mmu-miR-27b-5p mmu-miR-27b-3p mmu-miR-3074-1-3p mmu-miR-3074-5p
          Species:
          Mouse
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Mm03306468_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          66

          Chromosome Location:

          Chr. 13 - 63301008 - 63301008 [-] on 63301008 Build GRCm38

          Species:

          Mouse

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-24-3p MIMAT0000219
          Mouse mmu-miR-24-1-5p MIMAT0000218
          Mouse mmu-miR-27b-5p MIMAT0004522
          Mouse mmu-miR-27b-3p MIMAT0000126
          Mouse mmu-miR-3074-1-3p MIMAT0014857
          Mouse mmu-miR-3074-5p MIMAT0014856

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-24-1
          Stem-loop Accession # MI0000231
          Stem-loop Sequence
          CUCCGGUGCCUACUGAGCUGAUAUCAGUUCUCAUUUCACACACUGGCUCAGUUCAGCAGGAACAGGAG
          Chromosome Location Chr. 13 - 63301208 - 63301275 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000219
          miRBase ID: mmu-miR-24-3p
          Mature miRNA Sequence: UGGCUCAGUUCAGCAGGAACAG
          Chromosome Location: Chr. 13 - 63301208 - 63301275 [+] on Build GRCm38

          miRBase Accession #: MIMAT0000218
          miRBase ID: mmu-miR-24-1-5p
          Mature miRNA Sequence: GUGCCUACUGAGCUGAUAUCAGU
          Chromosome Location: Chr. 13 - 63301208 - 63301275 [+] on Build GRCm38

          Gene Family ID MIPF0000041, mir-24
          Distance to Amplicon 173 bases
          Stem-loop ID mmu-mir-27b
          Stem-loop Accession # MI0000142
          Stem-loop Sequence
          AGGUGCAGAGCUUAGCUGAUUGGUGAACAGUGAUUGGUUUCCGCUUUGUUCACAGUGGCUAAGUUCUGCACCU
          Chromosome Location Chr. 13 - 63300712 - 63300784 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004522
          miRBase ID: mmu-miR-27b-5p
          Mature miRNA Sequence: AGAGCUUAGCUGAUUGGUGAAC
          Chromosome Location: Chr. 13 - 63300712 - 63300784 [+] on Build GRCm38

          miRBase Accession #: MIMAT0000126
          miRBase ID: mmu-miR-27b-3p
          Mature miRNA Sequence: UUCACAGUGGCUAAGUUCUGC
          Chromosome Location: Chr. 13 - 63300712 - 63300784 [+] on Build GRCm38

          Gene Family ID MIPF0000036, mir-27
          Distance to Amplicon 186 bases
          Stem-loop ID mmu-mir-3074-1
          Stem-loop Accession # MI0014037
          Stem-loop Sequence
          GGGCUCGACUCCUGUUCCUGCUGAACUGAGCCAGUGUGUGAAAUGAGAACUGAUAUCAGCUCAGUAGGCACCGGAGGGCGGGUCC
          Chromosome Location Chr. 13 - 63301199 - 63301283 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0014857
          miRBase ID: mmu-miR-3074-1-3p
          Mature miRNA Sequence: GAUAUCAGCUCAGUAGGCACCG
          Chromosome Location: Chr. 13 - 63301199 - 63301283 [-] on Build GRCm38

          miRBase Accession #: MIMAT0014856
          miRBase ID: mmu-miR-3074-5p
          Mature miRNA Sequence: GUUCCUGCUGAACUGAGCCAGU
          Chromosome Location: Chr. 13 - 63301199 - 63301283 [-] on Build GRCm38

          Gene Family ID MIPF0001103, mir-3074
          Distance to Amplicon 164 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM10737
          Ambion® Pre-miR™ miRNA Precursor : PM10737
          TaqMan™ MicroRNA Assay : 000402
          mirVana® miRNA inhibitor : MH10737
          mirVana® miRNA mimic : MC10737
          TaqMan™ Advanced miRNA Assay : 477992_mir
          TaqMan™ Advanced miRNA Assay : rno481011_mir
          TaqMan™ Advanced miRNA Assay : mmu481011_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM18636
          Ambion® Pre-miR™ miRNA Precursor : PM18636
          TaqMan™ MicroRNA Assay : 243584_mat
          mirVana® miRNA inhibitor : MH18636
          mirVana® miRNA mimic : MC18636
          TaqMan™ Advanced miRNA Assay : 478803_mir
          TaqMan™ Advanced miRNA Assay : mmu481714_mir
          TaqMan™ Advanced miRNA Assay : rno478803_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM20237
          Ambion® Pre-miR™ miRNA Precursor : PM20237
          TaqMan™ MicroRNA Assay : 462202_mat
          mirVana® miRNA inhibitor : MH20237
          mirVana® miRNA mimic : MC20237
          TaqMan™ Advanced miRNA Assay : mmu481705_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10073
          Ambion® Pre-miR™ miRNA Precursor : PM10073
          TaqMan™ MicroRNA Assay : 000488
          mirVana® miRNA inhibitor : MH10073
          mirVana® miRNA mimic : MC10073
          TaqMan™ Advanced miRNA Assay : mmu482715_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12831
          Ambion® Pre-miR™ miRNA Precursor : PM12831
          TaqMan™ MicroRNA Assay : 002174
          mirVana® miRNA inhibitor : MH12831
          mirVana® miRNA mimic : MC12831
          TaqMan™ Advanced miRNA Assay : 478789_mir
          TaqMan™ Advanced miRNA Assay : mmu481663_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10750
          Ambion® Pre-miR™ miRNA Precursor : PM10750
          TaqMan™ MicroRNA Assay : 000409
          mirVana® miRNA inhibitor : MH10750
          mirVana® miRNA mimic : MC10750
          TaqMan™ Advanced miRNA Assay : 478270_mir
          TaqMan™ Advanced miRNA Assay : mmu478270_mir
          TaqMan™ Advanced miRNA Assay : rno478270_mir


          miRBase Alias:

          Mouse: mmu-mir-27b (v21)
          Mouse: mmu-mir-24-1 (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline