Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478803_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-3074-3p
          Stem-loop Accession Number: MI0014181
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 95086014 - 95086094 [-] on Build GRCh38
          Mature miRNA Sequence: GAUAUCAGCUCAGUAGGCACCG
          Species: Human, Mouse, Rat, Artibeus jamaicensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478803_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0001103, mir-3074

          Mature miRNA Sequence:

          GAUAUCAGCUCAGUAGGCACCG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-3074-3p MIMAT0015027
          Mouse mmu-miR-3074-1-3p MIMAT0014857
          Rat rno-miR-3074 MIMAT0017815
          Artibeus jamaicensis aja-miR-3074 MIMAT0031145

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-3074
          Stem-loop Accession # MI0014181
          Stem-loop Sequence
          GCUCGACUCCUGUUCCUGCUGAACUGAGCCAGUGUGUAAAAUGAGAACUGAUAUCAGCUCAGUAGGCACCGGAGGGCGGGU
          Chromosome Location Chr. 9 - 95086014 - 95086094 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0015027
          miRBase ID: hsa-miR-3074-3p
          Mature miRNA Sequence: GAUAUCAGCUCAGUAGGCACCG
          Chromosome Location: Chr. 9 - 95086014 - 95086094 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-3074-1
          Stem-loop Accession # MI0014037
          Stem-loop Sequence
          GGGCUCGACUCCUGUUCCUGCUGAACUGAGCCAGUGUGUGAAAUGAGAACUGAUAUCAGCUCAGUAGGCACCGGAGGGCGGGUCC
          Chromosome Location Chr. 13 - 63301199 - 63301283 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0014857
          miRBase ID: mmu-miR-3074-1-3p
          Mature miRNA Sequence: GAUAUCAGCUCAGUAGGCACCG
          Chromosome Location: Chr. 13 - 63301199 - 63301283 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-3074
          Stem-loop Accession # MI0015411
          Stem-loop Sequence
          GCUUUUUGCUCAAGGGCUCGACUCCUGUUCCUGCUGAACUGAGCCAGUGUGUGAAAUGAGAACUGAUAUCAGCUCAGUAGGCACCGGAGGGCGGGUCCAAUCACAGCUGGGCAGUGC
          Chromosome Location Chr. 17 - 823940 - 824056 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017815
          miRBase ID: rno-miR-3074
          Mature miRNA Sequence: GAUAUCAGCUCAGUAGGCACCG
          Chromosome Location: Chr. 17 - 823940 - 824056 [-] on Build Rnor_6.0

          Artibeus jamaicensis

          Stem-loop ID aja-mir-3074
          Stem-loop Accession # MI0024367
          Stem-loop Sequence
          GGGCUCGACUCCUGUUCCUGCUGAACUGAGCCAGUGUGUAAAAUGAGAACUGAUAUCAGCUCAGUAGGCACCGGAGGGCGGGUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031145
          miRBase ID: aja-miR-3074
          Mature miRNA Sequence: GAUAUCAGCUCAGUAGGCACCG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465740_pri
          TaqMan™ Pri-miRNA Assay : Hs03303056_pri
          TaqMan™ Pri-miRNA Assay : Mm03306468_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM18636
          Ambion® Pre-miR™ miRNA Precursor : PM18636
          TaqMan™ MicroRNA Assay : 243584_mat
          mirVana® miRNA inhibitor : MH18636
          mirVana® miRNA mimic : MC18636
          TaqMan™ Advanced miRNA Assay : mmu481714_mir
          TaqMan™ Advanced miRNA Assay : rno478803_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline