Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12831
          Assay Name: hsa-miR-27b-5p
          miRBase Accession Number: MI0000440
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 95085445 - 95085541 [+] on Build GRCh38
          Mature miRNA Sequence: AGAGCUUAGCUGAUUGGUGAAC
          Species: Human, Mouse, Alligator mississippiensis, Anolis carolinensis, Chrysemys picta, Cricetulus griseus, Goat, Ornithorhynchus anatinus, Pteropus alecto, Salmo salar, Taeniopygia guttata
          Product Type: mirVana® miRNA mimic
          Assay ID MC12831

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000036, mir-27

          Mature miRNA Sequence:

          AGAGCUUAGCUGAUUGGUGAAC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-27b-5p MIMAT0004588
          Mouse mmu-miR-27b-5p MIMAT0004522
          Alligator mississippiensis ami-miR-27b-5p MIMAT0038069
          Anolis carolinensis aca-miR-27b-5p MIMAT0021907
          Chrysemys picta cpi-miR-27b-5p MIMAT0037666
          Cricetulus griseus cgr-miR-27b-5p MIMAT0023876
          View More Rows
          Goat chi-miR-27b-5p MIMAT0036107
          Ornithorhynchus anatinus oan-miR-27b-5p MIMAT0006956
          Pteropus alecto pal-miR-27b-5p MIMAT0039809
          Salmo salar ssa-miR-27b-5p MIMAT0032576
          Taeniopygia guttata tgu-miR-27-5p MIMAT0026991

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-27b
          Stem-loop Accession # MI0000440
          Stem-loop Sequence
          ACCUCUCUAACAAGGUGCAGAGCUUAGCUGAUUGGUGAACAGUGAUUGGUUUCCGCUUUGUUCACAGUGGCUAAGUUCUGCACCUGAAGAGAAGGUG
          Chromosome Location Chr. 9 - 95085445 - 95085541 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004588
          miRBase ID: hsa-miR-27b-5p
          Mature miRNA Sequence: AGAGCUUAGCUGAUUGGUGAAC
          Chromosome Location: Chr. 9 - 95085445 - 95085541 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-27b
          Stem-loop Accession # MI0000142
          Stem-loop Sequence
          AGGUGCAGAGCUUAGCUGAUUGGUGAACAGUGAUUGGUUUCCGCUUUGUUCACAGUGGCUAAGUUCUGCACCU
          Chromosome Location Chr. 13 - 63300712 - 63300784 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004522
          miRBase ID: mmu-miR-27b-5p
          Mature miRNA Sequence: AGAGCUUAGCUGAUUGGUGAAC
          Chromosome Location: Chr. 13 - 63300712 - 63300784 [+] on Build GRCm38

          Alligator mississippiensis

          Stem-loop ID ami-mir-27b
          Stem-loop Accession # MI0029685
          Stem-loop Sequence
          CCUCUCUGACAAGGUGCAGAGCUUAGCUGAUUGGUGAACAGUGAUUGAUUUCCCCUUUGUUCACAGUGGCUAAGUUCUGCACCUGAAGAGAAGGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038069
          miRBase ID: ami-miR-27b-5p
          Mature miRNA Sequence: AGAGCUUAGCUGAUUGGUGAAC
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-27b
          Stem-loop Accession # MI0018838
          Stem-loop Sequence
          UGAGGUGCAGAGCUUAGCUGAUUGGUGAACAGUGAUUGAUUUCCUCUUUGUUCACAGUGGCUAAGUUCUGCACCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021907
          miRBase ID: aca-miR-27b-5p
          Mature miRNA Sequence: AGAGCUUAGCUGAUUGGUGAAC
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-27b
          Stem-loop Accession # MI0029419
          Stem-loop Sequence
          ACCUCUCUAACAAGGUGCAGAGCUUAGCUGAUUGGUGAACAGUGAUUGAUUUCCUCUUUGUUCACAGUGGCUAAGUUCUGCACCUGAAGAGAAGGUGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037666
          miRBase ID: cpi-miR-27b-5p
          Mature miRNA Sequence: AGAGCUUAGCUGAUUGGUGAAC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-27b
          Stem-loop Accession # MI0020474
          Stem-loop Sequence
          AACAAGGUGCAGAGCUUAGCUGAUUGGUGAACAGUGAUUGGUUUCCGCUUUGUUCACAGUGGCUAAGUUCUGCACCUGAAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023876
          miRBase ID: cgr-miR-27b-5p
          Mature miRNA Sequence: AGAGCUUAGCUGAUUGGUGAAC
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-27b
          Stem-loop Accession # MI0030723
          Stem-loop Sequence
          GCGACGACCUCUCUGACGAGGUGCAGAGCUUAGCUGAUUGGUGAACAGUGACUGGUUUCCGCUUUGUUCACAGUGGCUAAGUUCUGCACCUGAAGAGAAGGUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036107
          miRBase ID: chi-miR-27b-5p
          Mature miRNA Sequence: AGAGCUUAGCUGAUUGGUGAAC
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-27b
          Stem-loop Accession # MI0006758
          Stem-loop Sequence
          GCCGUCUCUCUGUCUCUCUGUCGCGGCGCAGAGCUUAGCUGAUUGGUGAACAGUGAUUGCUGGACUCUUUGUUCACAGUGGCUAAGUUCUGCGCCCGCAGAGAAGGGGACGGACGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006956
          miRBase ID: oan-miR-27b-5p
          Mature miRNA Sequence: AGAGCUUAGCUGAUUGGUGAAC
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-27b
          Stem-loop Accession # MI0032340
          Stem-loop Sequence
          AGAGCUUAGCUGAUUGGUGAACAGUGACUGGUUUCCGCUUUGUUCACAGUGGCUAAGUUCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039809
          miRBase ID: pal-miR-27b-5p
          Mature miRNA Sequence: AGAGCUUAGCUGAUUGGUGAAC
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-27b-1
          Stem-loop Accession # MI0026659
          Stem-loop Sequence
          AGAGCUUAGCUGAUUGGUGAACAGUGAUUGAUCUCCUCUUUGUUCACAGUGGCUAAGUUCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032576
          miRBase ID: ssa-miR-27b-5p
          Mature miRNA Sequence: AGAGCUUAGCUGAUUGGUGAAC
          Chromosome Location: NA

          Stem-loop ID ssa-mir-27b-2
          Stem-loop Accession # MI0026660
          Stem-loop Sequence
          AGAGCUUAGCUGAUUGGUGAACAGUGAUUGAUCUCCUUUGUUCACAGUGGCUAAGUUCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032576
          miRBase ID: ssa-miR-27b-5p
          Mature miRNA Sequence: AGAGCUUAGCUGAUUGGUGAAC
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-27
          Stem-loop Accession # MI0013748
          Stem-loop Sequence
          AUCUCUCUGGCGAGGUGCAGAGCUUAGCUGAUUGGUGAACAGUGAUUGUUUUCCUCUUUGUUCACAGUGGCUAAGUUCUGCACCUGAAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026991
          miRBase ID: tgu-miR-27-5p
          Mature miRNA Sequence: AGAGCUUAGCUGAUUGGUGAAC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303051_pri
          TaqMan™ Pri-miRNA Assay : Mm03306468_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12831
          Ambion® Pre-miR™ miRNA Precursor : PM12831
          TaqMan™ MicroRNA Assay : 002174
          mirVana® miRNA inhibitor : MH12831
          TaqMan™ Advanced miRNA Assay : 478789_mir
          TaqMan™ Advanced miRNA Assay : mmu481663_mir


          miRBase Alias:

          Human: hsa-miR-27b* (v17)
          Mouse: mmu-miR-27b* (v17)
          Aeromonas caviae Ae398: aca-miR-27b* (v18)
          Ochrobactrum anthropi: oan-miR-27b* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline