Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • beta Actin Antibodies

          Invitrogen

          beta Actin Monoclonal Antibody (AC-15)

          Advanced Verification
          37 Published Figures
          191 References
          View all (99) beta Actin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite beta Actin Monoclonal Antibody (AC-15)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          • Published Figures (37)
          • Advanced Verification (2)
          beta Actin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          beta Actin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 42

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          beta Actin Antibody (AM4302) in ICC/IF

          An siRNA targeting beta-actin was labeled with Cy™3 using the Silencer™ siRNA Labeling Kit. The labeled siRNA was transfected into HeLa cells and cells were analyzed 96 hours later using a beta Actin monoclonal antibody (Product # AM4302). Green: beta-actin protein detected with anti-ß-actin (Ambion) and a FITC labeled secondary antibody. Red: Cy3 labeled siRNA. Blue: DAPI stained nuclei. (Cy™3 is a tradema... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          beta Actin Antibody in Immunocytochemistry (ICC/IF)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Immunocytochemistry (ICC/IF)
          beta Actin Antibody in Immunocytochemistry (ICC/IF)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody in Western Blot (WB)
          beta Actin Antibody
          beta Actin Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          beta Actin Monoclonal Antibody (AC-15)

          Product Details

          AM4302

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 162 publications 162 publications

          Immunohistochemistry (IHC)

          -
          View 5 publications 5 publications

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 6 publications 6 publications

          Flow Cytometry (Flow)

          -
          View 2 publications 2 publications

          Miscellaneous PubMed (Misc)

          -
          View 16 publications 16 publications
          Product Specifications

          Species Reactivity

          Bovine, Dog, Chicken, Fruit fly, Cat, Guinea pig, Human, Sheep, Pig, Rabbit

          Published species

          Bacteria, Bovine, Hamster, Human, Mouse, Non-human primate, Pig, Rabbit, Rat, Sheep, Virus, Zebrafish

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          AC-15

          Immunogen

          A slightly modified synthetic beta-cytoplasmic actin N-terminal peptide,Ac-Asp-Asp-Asp-Ile-Ala-Ala-Leu-Val-Ile-Asp-Asn-Gly-Ser-Gly-Lys, conjugated to KLH
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Amount

          100 µg

          Storage buffer

          ascites

          Contains

          15mM sodium azide

          Storage conditions

          Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C

          Shipping conditions

          Dry ice

          RRID

          AB_2536382

          Product Specific Information

          The anti-beta-actin monoclonal antibody can be used to analyze beta-actin protein expression in experimental cells by immunofluorescence and western blot and has been specifically used in combination with beta-actin siRNA control to study the specificity and effectiveness of the RNA Interference (RNAi) effect at the protein level.

          The antibody reacts with beta-actin expressing cells in human, bovine, sheep, pig, rabbit, cat, dog, mouse, rat, guinea pig, chicken, carp, leech and fruit fly tissues, but not in amoeba.

          Use Anti-beta-actin for detection of beta-actin by ELISA, immunohistology, western blotting, and immunofluorescence. The antibody specifically recognizes beta-actin in a wide variety of tissues and species using immunoblotting, immunofluorescent staining of cultured cell lines, and immunohistochemistry. The epitope recognized by the antibody is resistant to formalin-fixation and paraffin-embedding. Ethanol, B5, methacarn, or Bouin's solutions may also be used as fixatives.

          Target Information

          Beta actin is one of six actin isoforms which have been identified. Beta actin is a non-muscle cytoskeletal protein in all human cell types and is involved in cell motility, structure, and integrity. There are six different actin isoforms in human. The beta isoform of actin, along with gamma actin, coexist in most cell types as components of the cyto-skeleton. Beta actins are cytoplasmic proteins ubuquitously expressed in all eukaryotic cells. Polymerization of globular actin (G-actin) leads to a structural filament (F-actin) in the form of two stranded helix. Actins are highly conserved proteins that are involved in cell motility, structure and integrity. Because beta actin is ubiquitously expressed in all eukaryotic cells, it is frequently used as a loading control for assays involving protein detection, such as Western blotting. Antibodies to beta-actin provide a specific and useful tool in studying the intracellular distribution of beta-actin and the static and dynamic aspects of the cytoskeleton.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Actin is a protein with a high degree of evolutionary conservation that undergoes polymerization to generate filaments, which subsequently assemble into interconnected networks within the cytoplasm of cells.; actin, beta, cytoplasmic; Bact; actin; beta actin; beta cytoskeletal actin; beta-actin; cytoplasmic 1; cytoskeletal beta actin; cytoskeletal protein; gamma non-muscle actin; I(2)-actin; PS1TP5-binding protein 1; unnamed protein product

          View more View less

          Gene Aliases: ACT-5; actin; Bact; BKRNS; BNS; BRWS1; CSMH; DDS1; PS1TP5BP1; THC8

          View more View less

          UniProt ID: (Sheep) P60713, (Rabbit) P29751, (Bovine) P60712, (Pig) Q6QAQ1, (Human) P60709, (Chicken) P60706, (Dog) O18840, (Guinea pig) Q71FK5

          View more View less

          Entrez Gene ID: (Sheep) 443052, (Rabbit) 100009272, (Bovine) 280979, (Pig) 414396, (Human) 60, (Chicken) 396526, (Dog) 403580, (Guinea pig) 100135470

          View more View less

          Function(s)
          protein binding ATP binding kinesin binding Tat protein binding identical protein binding nitric-oxide synthase binding structural constituent of cytoskeleton ATPase activity protein kinase binding nitric-oxide synthase regulator activity nucleosomal DNA binding tau protein binding structural constituent of postsynaptic actin cytoskeleton transporter regulator activity actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein
          Process(es)
          morphogenesis of a polarized epithelium establishment or maintenance of cell polarity regulation of transmembrane transporter activity negative regulation of protein binding adherens junction assembly apical protein localization synaptic vesicle endocytosis regulation of norepinephrine uptake protein localization to adherens junction cellular response to cytochalasin B postsynaptic actin cytoskeleton organization regulation of protein localization to plasma membrane chromatin remodeling regulation of transcription from RNA polymerase II promoter cytoskeleton organization microtubule nucleation axonogenesis positive regulation of cell proliferation substantia nigra development regulation of mitotic metaphase/anaphase transition maintenance of blood-brain barrier regulation of apoptotic process positive regulation of T cell differentiation negative regulation of cell differentiation positive regulation of cell differentiation positive regulation of myoblast differentiation positive regulation of transcription, DNA-templated cell motility positive regulation of norepinephrine uptake regulation of cell cycle regulation of G0 to G1 transition platelet aggregation regulation of transepithelial transport regulation of synaptic vesicle endocytosis positive regulation of stem cell population maintenance positive regulation of double-strand break repair via homologous recombination regulation of G1/S transition of mitotic cell cycle regulation of double-strand break repair positive regulation of double-strand break repair regulation of nucleotide-excision repair
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5d6d8bc979-gw77j:80/100.66.76.64:80.
          git-commit: e71199a02a6c1449b2ac9ded8316d5734e8aa2de
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.62.0-2026.11.45-1.0