Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM11039
          Assay Name: hsa-miR-34c-5p
          miRBase Accession Number: MI0000743
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 111513439 - 111513515 [+] on Build GRCh38
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Species: Human, Mouse, Rat, Callithrix jacchus, Chicken, Chrysemys picta, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Dog, Goat, Guinea pig, Horse, Microcebus murinus, Monodelphis domestica, Nomascus leucogenys, Ornithorhynchus anatinus, Otolemur garnettii, Pan paniscus, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Taeniopygia guttata, Tupaia chinensis
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM11039
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000039, mir-34

          Mature miRNA Sequence:

          AGGCAGUGUAGUUAGCUGAUUGC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-34c-5p MIMAT0000686
          Mouse mmu-miR-34c-5p MIMAT0000381
          Rat rno-miR-34c-5p MIMAT0000814
          Callithrix jacchus cja-miR-34c MIMAT0039373
          Chicken gga-miR-34c-5p MIMAT0001180
          Chrysemys picta cpi-miR-34c-5p MIMAT0037696
          View More Rows
          Columba livia cli-miR-34c-5p MIMAT0038471
          Cricetulus griseus cgr-miR-34c-5p MIMAT0023929
          Dasypus novemcinctus dno-miR-34c-5p MIMAT0047579
          Dog cfa-miR-34c MIMAT0006693
          Goat chi-miR-34c-5p MIMAT0036169
          Guinea pig cpo-miR-34c-5p MIMAT0046926
          Horse eca-miR-34c MIMAT0012992
          Microcebus murinus mmr-miR-34c MIMAT0049175
          Monodelphis domestica mdo-miR-34c-5p MIMAT0028586
          Nomascus leucogenys nle-miR-34c MIMAT0049467
          Ornithorhynchus anatinus oan-miR-34a-5p MIMAT0007108
          Otolemur garnettii oga-miR-34c MIMAT0049601
          Pan paniscus ppa-miR-34c MIMAT0049104
          Pig ssc-miR-34c MIMAT0013916
          Pongo pygmaeus ppy-miR-34c-5p MIMAT0015742
          Pteropus alecto pal-miR-34c-5p MIMAT0039791
          Rabbit ocu-miR-34c-5p MIMAT0048171
          Rhesus monkey mml-miR-34c-5p MIMAT0006175
          Taeniopygia guttata tgu-miR-34b MIMAT0014537
          Tupaia chinensis tch-miR-34c-5p MIMAT0036575

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-34c
          Stem-loop Accession # MI0000743
          Stem-loop Sequence
          AGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGGUAAAAAGAUU
          Chromosome Location Chr. 11 - 111513439 - 111513515 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000686
          miRBase ID: hsa-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: Chr. 11 - 111513439 - 111513515 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-34c
          Stem-loop Accession # MI0000403
          Stem-loop Sequence
          AGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACAGCCAGGUAAAAAGACU
          Chromosome Location Chr. 9 - 51103034 - 51103110 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000381
          miRBase ID: mmu-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: Chr. 9 - 51103034 - 51103110 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-34c
          Stem-loop Accession # MI0000876
          Stem-loop Sequence
          AGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACAGCCAGGUAAAAAGACU
          Chromosome Location Chr. 8 - 55492024 - 55492100 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000814
          miRBase ID: rno-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: Chr. 8 - 55492024 - 55492100 [-] on Build Rnor_6.0

          Callithrix jacchus

          Stem-loop ID cja-mir-34c
          Stem-loop Accession # MI0031891
          Stem-loop Sequence
          GAGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGGUAAAAAGAUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039373
          miRBase ID: cja-miR-34c
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-34c
          Stem-loop Accession # MI0001261
          Stem-loop Sequence
          AGCCUGGUUACCAGGCAGUGUAGUUAGCUGAUUGCCACCAGGACCAAUCACUAACCACACAGCCAGGUAAAAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001180
          miRBase ID: gga-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-34c
          Stem-loop Accession # MI0029436
          Stem-loop Sequence
          ACAUUAGCCUAGUUACCAGGCAGUGUAGUUAGCUGAUUGCUAAAGGUACAAUCACUAACCACACAGCCAGGUAAAAAGGUAUGUAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037696
          miRBase ID: cpi-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-34c
          Stem-loop Accession # MI0029941
          Stem-loop Sequence
          AUCAGCCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCCAAAAGUACCAAUCACUAGCCACACAGCCAGGUAAAAAGGCUUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038471
          miRBase ID: cli-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-34c
          Stem-loop Accession # MI0020507
          Stem-loop Sequence
          CUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGGUAAAAAGACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023929
          miRBase ID: cgr-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-34c
          Stem-loop Accession # MI0038946
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047579
          miRBase ID: dno-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-34c
          Stem-loop Accession # MI0008106
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006693
          miRBase ID: cfa-miR-34c
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-34c
          Stem-loop Accession # MI0030759
          Stem-loop Sequence
          UGAGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAAUACCAAUCACUAACCACACGGCCAGGUAAAAAGAUUUGGGAAUUCAUCCAAAUGAGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036169
          miRBase ID: chi-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-34c
          Stem-loop Accession # MI0038602
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046926
          miRBase ID: cpo-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-34c
          Stem-loop Accession # MI0012742
          Stem-loop Sequence
          AGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAAUACCAAUCACUAACCACACGGCCAGGUAAAAAGAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012992
          miRBase ID: eca-miR-34c
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-34c
          Stem-loop Accession # MI0039893
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049175
          miRBase ID: mmr-miR-34c
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-34c
          Stem-loop Accession # MI0023815
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGCUAGCAGUAGUACCAAUCACUAACCACACAGCCAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0028586
          miRBase ID: mdo-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-34c
          Stem-loop Accession # MI0040223
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGCGAAUAGUACCAAUCACUAACCACACGGCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049467
          miRBase ID: nle-miR-34c
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-34a
          Stem-loop Accession # MI0006850
          Stem-loop Sequence
          UUGCCAUCAGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAUUAACACCAAUCACUAGCCACACAGCCAGGUAAAAAGACCUGCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007108
          miRBase ID: oan-miR-34a-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-34c
          Stem-loop Accession # MI0040382
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049601
          miRBase ID: oga-miR-34c
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-34c
          Stem-loop Accession # MI0039816
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049104
          miRBase ID: ppa-miR-34c
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-34c-1
          Stem-loop Accession # MI0013132
          Stem-loop Sequence
          AGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUGAUAAAACCAAUCACUAACCACACGGCCAGGUAAAAAGAUUUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013916
          miRBase ID: ssc-miR-34c
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Stem-loop ID ssc-mir-34c-2
          Stem-loop Accession # MI0022128
          Stem-loop Sequence
          UCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUGAUAAAACCAAUCACUAACCACACGGCCAGGUAAAAAGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013916
          miRBase ID: ssc-miR-34c
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-34c
          Stem-loop Accession # MI0014808
          Stem-loop Sequence
          AGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGGUAAAAAGAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015742
          miRBase ID: ppy-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-34c
          Stem-loop Accession # MI0032331
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039791
          miRBase ID: pal-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-34c
          Stem-loop Accession # MI0039258
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGCUGAUAGUACCAAUCACUAACCACACGGCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048171
          miRBase ID: ocu-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-34c
          Stem-loop Accession # MI0007605
          Stem-loop Sequence
          AGUCUAGUUACCAGGCAGUGUAGUUAGCUGAUUGCUGAUAGUACCAAUCACUAACCACACGGCCAGGUAAAAAGAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006175
          miRBase ID: mml-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-34b
          Stem-loop Accession # MI0013751
          Stem-loop Sequence
          AAAAUUUGGGGUGAAAGGCUCUGCCAUCAGCCUGGUUGCCAGGCAGUGUAGUUAGCUGAUUGCCCAAAGCAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014537
          miRBase ID: tgu-miR-34b
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-34c
          Stem-loop Accession # MI0031239
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACGGCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036575
          miRBase ID: tch-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465278_pri
          TaqMan™ Pri-miRNA Assay : Mm03306660_pri
          TaqMan™ Pri-miRNA Assay : Hs03295169_pri
          Ambion® Pre-miR™ miRNA Precursor : PM11039
          TaqMan™ MicroRNA Assay : 000428
          mirVana® miRNA inhibitor : MH11039
          mirVana® miRNA mimic : MC11039
          TaqMan™ Advanced miRNA Assay : 478052_mir


          miRBase Alias:

          Human: hsa-miR-34c (v9.2)
          Mouse: mmu-miR-34c (v17)
          Rat: rno-miR-34c (v18)
          Gallus gallus: gga-miR-34c (v19)
          Ochrobactrum anthropi: oan-miR-34-5p (v19)
          Ochrobactrum anthropi: oan-miR-34 (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline