Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03306660_pri
          Assay Name:
          mmu-mir-34b
          Stem-loop ID:
          mmu-mir-34b mmu-mir-34c
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.9: 51103562 - 51103645 [-] on Build GRCm38
          Stem-loop Location:
          Chr.9: 51103562 - 51103645 [-] on Build GRCm38 Chr.9: 51103034 - 51103110 [-] on Build GRCm38
          Mature miRNA ID:
          mmu-miR-34b-3p mmu-miR-34b-5p mmu-miR-34c-3p mmu-miR-34c-5p
          Species:
          Mouse
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Mm03306660_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          72

          Chromosome Location:

          Chr. 9 - 51103368 - 51103368 [+] on 51103368 Build GRCm38

          Species:

          Mouse

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-34b-3p MIMAT0004581
          Mouse mmu-miR-34b-5p MIMAT0000382
          Mouse mmu-miR-34c-3p MIMAT0004580
          Mouse mmu-miR-34c-5p MIMAT0000381

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-34b
          Stem-loop Accession # MI0000404
          Stem-loop Sequence
          GUGCUCGGUUUGUAGGCAGUGUAAUUAGCUGAUUGUAGUGCGGUGCUGACAAUCACUAACUCCACUGCCAUCAAAACAAGGCAC
          Chromosome Location Chr. 9 - 51103562 - 51103645 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004581
          miRBase ID: mmu-miR-34b-3p
          Mature miRNA Sequence: AAUCACUAACUCCACUGCCAUC
          Chromosome Location: Chr. 9 - 51103562 - 51103645 [-] on Build GRCm38

          miRBase Accession #: MIMAT0000382
          miRBase ID: mmu-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: Chr. 9 - 51103562 - 51103645 [-] on Build GRCm38

          Gene Family ID MIPF0000039, mir-34
          Distance to Amplicon 159 bases
          Stem-loop ID mmu-mir-34c
          Stem-loop Accession # MI0000403
          Stem-loop Sequence
          AGUCUAGUUACUAGGCAGUGUAGUUAGCUGAUUGCUAAUAGUACCAAUCACUAACCACACAGCCAGGUAAAAAGACU
          Chromosome Location Chr. 9 - 51103034 - 51103110 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004580
          miRBase ID: mmu-miR-34c-3p
          Mature miRNA Sequence: AAUCACUAACCACACAGCCAGG
          Chromosome Location: Chr. 9 - 51103034 - 51103110 [-] on Build GRCm38

          miRBase Accession #: MIMAT0000381
          miRBase ID: mmu-miR-34c-5p
          Mature miRNA Sequence: AGGCAGUGUAGUUAGCUGAUUGC
          Chromosome Location: Chr. 9 - 51103034 - 51103110 [-] on Build GRCm38

          Gene Family ID MIPF0000039, mir-34
          Distance to Amplicon 222 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM12245
          Ambion® Pre-miR™ miRNA Precursor : PM12245
          TaqMan™ MicroRNA Assay : 002584
          mirVana® miRNA inhibitor : MH12245
          mirVana® miRNA mimic : MC12245
          TaqMan™ Advanced miRNA Assay : mmu481802_mir
          TaqMan™ Advanced miRNA Assay : rno481085_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM11039
          Ambion® Pre-miR™ miRNA Precursor : PM11039
          TaqMan™ MicroRNA Assay : 000428
          mirVana® miRNA inhibitor : MH11039
          mirVana® miRNA mimic : MC11039
          TaqMan™ Advanced miRNA Assay : 478052_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12545
          Ambion® Pre-miR™ miRNA Precursor : PM12545
          TaqMan™ MicroRNA Assay : 002618
          mirVana® miRNA inhibitor : MH12545
          mirVana® miRNA mimic : MC12545
          TaqMan™ Advanced miRNA Assay : rno481084_mir
          TaqMan™ Advanced miRNA Assay : mmu481084_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM12558
          Ambion® Pre-miR™ miRNA Precursor : PM12558
          TaqMan™ MicroRNA Assay : 002617
          mirVana® miRNA inhibitor : MH12558
          mirVana® miRNA mimic : MC12558


          miRBase Alias:

          Mouse: mmu-mir-34c (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline