Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM12558
          Assay Name: mmu-miR-34b-5p
          miRBase Accession Number: MI0000404
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 51103562 - 51103645 [-] on Build GRCm38
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Species: Mouse, Rat, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Horse, Microcebus murinus, Otolemur garnettii, Pteropus alecto
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM12558
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000039, mir-34

          Mature miRNA Sequence:

          AGGCAGUGUAAUUAGCUGAUUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-34b-5p MIMAT0000382
          Rat rno-miR-34b-5p MIMAT0000813
          Cricetulus griseus cgr-miR-34b-5p MIMAT0023927
          Dasypus novemcinctus dno-miR-34b-5p MIMAT0047577
          Goat chi-miR-34b-5p MIMAT0036167
          Guinea pig cpo-miR-34b-5p MIMAT0046924
          View More Rows
          Horse eca-miR-34b-5p MIMAT0012990
          Microcebus murinus mmr-miR-34b MIMAT0049176
          Otolemur garnettii oga-miR-34b MIMAT0049602
          Pteropus alecto pal-miR-34b-5p MIMAT0039888

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-34b
          Stem-loop Accession # MI0000404
          Stem-loop Sequence
          GUGCUCGGUUUGUAGGCAGUGUAAUUAGCUGAUUGUAGUGCGGUGCUGACAAUCACUAACUCCACUGCCAUCAAAACAAGGCAC
          Chromosome Location Chr. 9 - 51103562 - 51103645 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000382
          miRBase ID: mmu-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: Chr. 9 - 51103562 - 51103645 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-34b
          Stem-loop Accession # MI0000875
          Stem-loop Sequence
          GUGCUCGGUUUGUAGGCAGUGUAAUUAGCUGAUUGUAGUGCGGUGCUGACAAUCACUAACUCCACUGCCAUCAAAACAAGGCAC
          Chromosome Location Chr. 8 - 55492542 - 55492625 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000813
          miRBase ID: rno-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: Chr. 8 - 55492542 - 55492625 [-] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-mir-34b
          Stem-loop Accession # MI0020506
          Stem-loop Sequence
          UCGGUUUGUAGGCAGUGUAAUUAGCUGAUUGUACCGCGGUGCUGACAAUCACUAACUCCACUGCCAUCAAAACAAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023927
          miRBase ID: cgr-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-34b
          Stem-loop Accession # MI0038945
          Stem-loop Sequence
          AGGCAGUGUAAUUAGCUGAUUGUCUGGUGCUUGCAAUCACUAACUCCACUGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047577
          miRBase ID: dno-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-34b
          Stem-loop Accession # MI0030758
          Stem-loop Sequence
          GCCCGGUGCUCGGUUUGUAGGCAGUGUAAUUAGCUGAUUGUACUCUCAUGCUUACAAUCACUAGUUCCACUGCCAUCAAAACAAGGCACAGCAUCACCCGCCGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036167
          miRBase ID: chi-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-34b
          Stem-loop Accession # MI0038601
          Stem-loop Sequence
          AGGCAGUGUAAUUAGCUGAUUGUGCUGUUGUGCUUACAAUCACUAACUCCACUGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046924
          miRBase ID: cpo-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-34b
          Stem-loop Accession # MI0012741
          Stem-loop Sequence
          GUGCUCGGUUUGUAGGCAGUGUAAUUAGCUGAUUGUACUCUGGAGCUUACAAUCACUAACUCCACUGCCAUCAAAACAAGGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012990
          miRBase ID: eca-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-34b
          Stem-loop Accession # MI0039894
          Stem-loop Sequence
          AGGCAGUGUAAUUAGCUGAUUGUACUGUGGCGCUUACAAUCACUAACUCCACUGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049176
          miRBase ID: mmr-miR-34b
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-34b
          Stem-loop Accession # MI0040383
          Stem-loop Sequence
          AGGCAGUGUAAUUAGCUGAUUGUACUGUGCUGCUUACAAUCACUAACUCCACUGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049602
          miRBase ID: oga-miR-34b
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-34b
          Stem-loop Accession # MI0032384
          Stem-loop Sequence
          AGGCAGUGUAAUUAGCUGAUUGUACUCUGGUGCUUACAAUCACUAACUCCACUGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039888
          miRBase ID: pal-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465278_pri
          TaqMan™ Pri-miRNA Assay : Mm03306660_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12558
          TaqMan™ MicroRNA Assay : 002617
          mirVana® miRNA inhibitor : MH12558
          mirVana® miRNA mimic : MC12558


          miRBase Alias:

          Rat: rno-miR-34b (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline