Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH12558
          Assay Name: mmu-miR-34b-5p
          miRBase Accession Number: MI0000404
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 51103562 - 51103645 [-] on Build GRCm38
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Species: Mouse, Rat, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Horse, Microcebus murinus, Otolemur garnettii, Pteropus alecto
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH12558

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000039, mir-34

          Mature miRNA Sequence:

          AGGCAGUGUAAUUAGCUGAUUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-34b-5p MIMAT0000382
          Rat rno-miR-34b-5p MIMAT0000813
          Cricetulus griseus cgr-miR-34b-5p MIMAT0023927
          Dasypus novemcinctus dno-miR-34b-5p MIMAT0047577
          Goat chi-miR-34b-5p MIMAT0036167
          Guinea pig cpo-miR-34b-5p MIMAT0046924
          View More Rows
          Horse eca-miR-34b-5p MIMAT0012990
          Microcebus murinus mmr-miR-34b MIMAT0049176
          Otolemur garnettii oga-miR-34b MIMAT0049602
          Pteropus alecto pal-miR-34b-5p MIMAT0039888

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-34b
          Stem-loop Accession # MI0000404
          Stem-loop Sequence
          GUGCUCGGUUUGUAGGCAGUGUAAUUAGCUGAUUGUAGUGCGGUGCUGACAAUCACUAACUCCACUGCCAUCAAAACAAGGCAC
          Chromosome Location Chr. 9 - 51103562 - 51103645 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000382
          miRBase ID: mmu-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: Chr. 9 - 51103562 - 51103645 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-34b
          Stem-loop Accession # MI0000875
          Stem-loop Sequence
          GUGCUCGGUUUGUAGGCAGUGUAAUUAGCUGAUUGUAGUGCGGUGCUGACAAUCACUAACUCCACUGCCAUCAAAACAAGGCAC
          Chromosome Location Chr. 8 - 55492542 - 55492625 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000813
          miRBase ID: rno-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: Chr. 8 - 55492542 - 55492625 [-] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-mir-34b
          Stem-loop Accession # MI0020506
          Stem-loop Sequence
          UCGGUUUGUAGGCAGUGUAAUUAGCUGAUUGUACCGCGGUGCUGACAAUCACUAACUCCACUGCCAUCAAAACAAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023927
          miRBase ID: cgr-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-34b
          Stem-loop Accession # MI0038945
          Stem-loop Sequence
          AGGCAGUGUAAUUAGCUGAUUGUCUGGUGCUUGCAAUCACUAACUCCACUGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047577
          miRBase ID: dno-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-34b
          Stem-loop Accession # MI0030758
          Stem-loop Sequence
          GCCCGGUGCUCGGUUUGUAGGCAGUGUAAUUAGCUGAUUGUACUCUCAUGCUUACAAUCACUAGUUCCACUGCCAUCAAAACAAGGCACAGCAUCACCCGCCGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036167
          miRBase ID: chi-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-34b
          Stem-loop Accession # MI0038601
          Stem-loop Sequence
          AGGCAGUGUAAUUAGCUGAUUGUGCUGUUGUGCUUACAAUCACUAACUCCACUGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046924
          miRBase ID: cpo-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-34b
          Stem-loop Accession # MI0012741
          Stem-loop Sequence
          GUGCUCGGUUUGUAGGCAGUGUAAUUAGCUGAUUGUACUCUGGAGCUUACAAUCACUAACUCCACUGCCAUCAAAACAAGGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012990
          miRBase ID: eca-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-34b
          Stem-loop Accession # MI0039894
          Stem-loop Sequence
          AGGCAGUGUAAUUAGCUGAUUGUACUGUGGCGCUUACAAUCACUAACUCCACUGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049176
          miRBase ID: mmr-miR-34b
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-34b
          Stem-loop Accession # MI0040383
          Stem-loop Sequence
          AGGCAGUGUAAUUAGCUGAUUGUACUGUGCUGCUUACAAUCACUAACUCCACUGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049602
          miRBase ID: oga-miR-34b
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-34b
          Stem-loop Accession # MI0032384
          Stem-loop Sequence
          AGGCAGUGUAAUUAGCUGAUUGUACUCUGGUGCUUACAAUCACUAACUCCACUGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039888
          miRBase ID: pal-miR-34b-5p
          Mature miRNA Sequence: AGGCAGUGUAAUUAGCUGAUUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465278_pri
          TaqMan™ Pri-miRNA Assay : Mm03306660_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12558
          Ambion® Pre-miR™ miRNA Precursor : PM12558
          TaqMan™ MicroRNA Assay : 002617
          mirVana® miRNA mimic : MC12558


          miRBase Alias:

          Rat: rno-miR-34b (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline