Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002618
          Assay Name: mmu-miR-34b-3p
          miRBase Accession Number: MI0000404
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 51103562 - 51103645 [-] on Build GRCm38
          Mature miRNA Sequence: AAUCACUAACUCCACUGCCAUC
          Species: Mouse, Rat, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Guinea pig, Horse, Monodelphis domestica, Ornithorhynchus anatinus, Rabbit
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002618
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000039, mir-34

          Mature miRNA Sequence:

          AAUCACUAACUCCACUGCCAUC

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-34b-3p MIMAT0004581
          Rat rno-miR-34b-3p MIMAT0017105
          Callithrix jacchus cja-miR-34b MIMAT0039548
          Cricetulus griseus cgr-miR-34b-3p MIMAT0023928
          Dasypus novemcinctus dno-miR-34b-3p MIMAT0047578
          Guinea pig cpo-miR-34b-3p MIMAT0046925
          View More Rows
          Horse eca-miR-34b-3p MIMAT0012991
          Monodelphis domestica mdo-miR-34b-3p MIMAT0028589
          Ornithorhynchus anatinus oan-miR-34b-3p MIMAT0028989
          Rabbit ocu-miR-34b-3p MIMAT0048170

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-34b
          Stem-loop Accession # MI0000404
          Stem-loop Sequence
          GUGCUCGGUUUGUAGGCAGUGUAAUUAGCUGAUUGUAGUGCGGUGCUGACAAUCACUAACUCCACUGCCAUCAAAACAAGGCAC
          Chromosome Location Chr. 9 - 51103562 - 51103645 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004581
          miRBase ID: mmu-miR-34b-3p
          Mature miRNA Sequence: AAUCACUAACUCCACUGCCAUC
          Chromosome Location: Chr. 9 - 51103562 - 51103645 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-34b
          Stem-loop Accession # MI0000875
          Stem-loop Sequence
          GUGCUCGGUUUGUAGGCAGUGUAAUUAGCUGAUUGUAGUGCGGUGCUGACAAUCACUAACUCCACUGCCAUCAAAACAAGGCAC
          Chromosome Location Chr. 8 - 55492542 - 55492625 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017105
          miRBase ID: rno-miR-34b-3p
          Mature miRNA Sequence: AAUCACUAACUCCACUGCCAUC
          Chromosome Location: Chr. 8 - 55492542 - 55492625 [-] on Build Rnor_6.0

          Callithrix jacchus

          Stem-loop ID cja-mir-34b
          Stem-loop Accession # MI0032079
          Stem-loop Sequence
          GUGCUCGGUUUGUAGGCAGUGUCAUUAGCUGAUUGUACUGUGCUGGUUACAAUCACUAACUCCACUGCCAUCAAAACAAGGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039548
          miRBase ID: cja-miR-34b
          Mature miRNA Sequence: AAUCACUAACUCCACUGCCAUC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-34b
          Stem-loop Accession # MI0020506
          Stem-loop Sequence
          UCGGUUUGUAGGCAGUGUAAUUAGCUGAUUGUACCGCGGUGCUGACAAUCACUAACUCCACUGCCAUCAAAACAAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023928
          miRBase ID: cgr-miR-34b-3p
          Mature miRNA Sequence: AAUCACUAACUCCACUGCCAUC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-34b
          Stem-loop Accession # MI0038945
          Stem-loop Sequence
          AGGCAGUGUAAUUAGCUGAUUGUCUGGUGCUUGCAAUCACUAACUCCACUGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047578
          miRBase ID: dno-miR-34b-3p
          Mature miRNA Sequence: AAUCACUAACUCCACUGCCAUC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-34b
          Stem-loop Accession # MI0038601
          Stem-loop Sequence
          AGGCAGUGUAAUUAGCUGAUUGUGCUGUUGUGCUUACAAUCACUAACUCCACUGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046925
          miRBase ID: cpo-miR-34b-3p
          Mature miRNA Sequence: AAUCACUAACUCCACUGCCAUC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-34b
          Stem-loop Accession # MI0012741
          Stem-loop Sequence
          GUGCUCGGUUUGUAGGCAGUGUAAUUAGCUGAUUGUACUCUGGAGCUUACAAUCACUAACUCCACUGCCAUCAAAACAAGGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012991
          miRBase ID: eca-miR-34b-3p
          Mature miRNA Sequence: AAUCACUAACUCCACUGCCAUC
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-34b
          Stem-loop Accession # MI0023816
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGUAUUCUACUGCCUACAAUCACUAACUCCACUGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0028589
          miRBase ID: mdo-miR-34b-3p
          Mature miRNA Sequence: AAUCACUAACUCCACUGCCAUC
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-34b
          Stem-loop Accession # MI0024078
          Stem-loop Sequence
          AGGCAGUGUAGUUAGCUGAUUGGACCAUCCCGCCCGCAAUCACUAACUCCACUGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0028989
          miRBase ID: oan-miR-34b-3p
          Mature miRNA Sequence: AAUCACUAACUCCACUGCCAUC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-34b
          Stem-loop Accession # MI0039257
          Stem-loop Sequence
          UAGGCAGUGUAAUUAGCUGAUUGUACUGUGGUGCUUACAAUCACUAACUCCACUGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048170
          miRBase ID: ocu-miR-34b-3p
          Mature miRNA Sequence: AAUCACUAACUCCACUGCCAUC
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pool:

          Megaplex™ RT Primers, Rodent Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465278_pri
          TaqMan™ Pri-miRNA Assay : Mm03306660_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12545
          Ambion® Pre-miR™ miRNA Precursor : PM12545
          mirVana® miRNA inhibitor : MH12545
          mirVana® miRNA mimic : MC12545
          TaqMan™ Advanced miRNA Assay : rno481084_mir
          TaqMan™ Advanced miRNA Assay : mmu481084_mir
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          FGMIR : TaqManTM Rodent MicroRNA Array A
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate


          miRBase Alias:

          Rat: rno-miR-34b* (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline