Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu481384_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-363-3p
          Stem-loop Accession Number: MI0000764
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 134169378 - 134169452 [-] on Build GRCh38
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Species: Human, Mouse, Callithrix jacchus, Cyprinus carpio, Dasypus novemcinctus, Guinea pig, Horse, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Tupaia chinensis, Xenopus laevis, Zebrafish
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu481384_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000138, mir-363

          Mature miRNA Sequence:

          AAUUGCACGGUAUCCAUCUGUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-363-3p MIMAT0000707
          Mouse mmu-miR-363-3p MIMAT0000708
          Callithrix jacchus cja-miR-363 MIMAT0039359
          Cyprinus carpio ccr-miR-363 MIMAT0026293
          Dasypus novemcinctus dno-miR-363-3p MIMAT0047834
          Guinea pig cpo-miR-363-3p MIMAT0047193
          View More Rows
          Horse eca-miR-363 MIMAT0013212
          Pongo pygmaeus ppy-miR-363 MIMAT0015852
          Pteropus alecto pal-miR-363-3p MIMAT0039877
          Rabbit ocu-miR-363-3p MIMAT0048398
          Rhesus monkey mml-miR-363-3p MIMAT0006290
          Tupaia chinensis tch-miR-363-3p MIMAT0036586
          Xenopus laevis xla-miR-363-3p MIMAT0046589
          Zebrafish dre-miR-363-3p MIMAT0001874

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-363
          Stem-loop Accession # MI0000764
          Stem-loop Sequence
          UGUUGUCGGGUGGAUCACGAUGCAAUUUUGAUGAGUAUCAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUAAACC
          Chromosome Location Chr. X - 134169378 - 134169452 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000707
          miRBase ID: hsa-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: Chr. X - 134169378 - 134169452 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-363
          Stem-loop Accession # MI0000765
          Stem-loop Sequence
          UGUUAUCAGGUGGAACACGAUGCAAUUUUGGUUGGUGUAAUAGGAGGAAAAUUGCACGGUAUCCAUCUGUAAACC
          Chromosome Location Chr. X - 52741693 - 52741767 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000708
          miRBase ID: mmu-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: Chr. X - 52741693 - 52741767 [-] on Build GRCm38

          Callithrix jacchus

          Stem-loop ID cja-mir-363
          Stem-loop Accession # MI0031873
          Stem-loop Sequence
          GUUCUGAUAUUUAGGCAUUGUAAAAUGAUCUGUUUUGCUGUUGUCGGGUGGAUCACGAUGCAAUUUUGAUUAGUAUCAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUAAACCGCAGGACCUUUGUUGGCGACAUUCCUCAUCAGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039359
          miRBase ID: cja-miR-363
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-363
          Stem-loop Accession # MI0023393
          Stem-loop Sequence
          UUUCGGGUGGAUGACCCCGCAAUUUUAAUAGUGGCAAAAAUCUUCCAACAAAAAUUGCACGGUAUCCAUCUGUAAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026293
          miRBase ID: ccr-miR-363
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-363
          Stem-loop Accession # MI0039081
          Stem-loop Sequence
          CGGGUGGAUCACGAUGCAAUUUUGAUUAGUAUAAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047834
          miRBase ID: dno-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-363
          Stem-loop Accession # MI0038744
          Stem-loop Sequence
          CGGGUGGAUCACGAUGCAAUUUUGAUUAGACUAAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047193
          miRBase ID: cpo-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-363
          Stem-loop Accession # MI0012958
          Stem-loop Sequence
          UGUUGUCGGGUGGAUCACGAUGCAAUUUUGAUUAGUAUAAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUAAACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013212
          miRBase ID: eca-miR-363
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-363
          Stem-loop Accession # MI0014910
          Stem-loop Sequence
          UGUUGUCGGGUGGAUCACGAUGCAAUUUUGAUUAGUAUCAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUAAACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015852
          miRBase ID: ppy-miR-363
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-363
          Stem-loop Accession # MI0032377
          Stem-loop Sequence
          CGGGUGGAUCACGAUGCAAUUUUGAUUAGUAUAAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039877
          miRBase ID: pal-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-363
          Stem-loop Accession # MI0039379
          Stem-loop Sequence
          CGGGUGGAUCACGAUGCAAUUUUGAUUAGUACAAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048398
          miRBase ID: ocu-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-363
          Stem-loop Accession # MI0007709
          Stem-loop Sequence
          UGUUGUCGGGUGGAUCACGAUGCAAUUUUGAUUAGUAUCAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUAAACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006290
          miRBase ID: mml-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-363
          Stem-loop Accession # MI0031252
          Stem-loop Sequence
          CGGGUGGAUCACGAUGCAAUUUUGAUUAGUAUAAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036586
          miRBase ID: tch-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-363-2
          Stem-loop Accession # MI0038327
          Stem-loop Sequence
          CGGGUGGAUCACGAUGCAAUUUUAUUGAGCUUGGUAGGAGAAAAAUUGCACGGUAUCCAUCUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046589
          miRBase ID: xla-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-363
          Stem-loop Accession # MI0002067
          Stem-loop Sequence
          UAAAUGCAAAUAUUUGUCUUGCUGUUUUCGGGUGGAUGACUCUGCAAUUUUAUUAGUGAUGGAAAAACUUCAAUAAAAAUUGCACGGUAUCCAUCUGUAAUCCGCUGGAUUCCAUUACCUGCGUUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001874
          miRBase ID: dre-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03295977_pri
          TaqMan™ Pri-miRNA Assay : Mm03307329_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10149
          Ambion® Pre-miR™ miRNA Precursor : PM10149
          TaqMan™ MicroRNA Assay : 001271
          mirVana® miRNA inhibitor : MH10149
          mirVana® miRNA mimic : MC10149
          TaqMan™ Advanced miRNA Assay : 478060_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline