Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno477804_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-20b-5p
          Stem-loop Accession Number: MI0001519
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 134169809 - 134169877 [-] on Build GRCh38
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Species: Human, Mouse, Rat, Callithrix jacchus, Chicken, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Gorilla gorilla, Horse, Monodelphis domestica, Ornithorhynchus anatinus, Pan troglodytes, Pongo pygmaeus, Pteropus alecto, Python bivittatus, Rhesus monkey, Taeniopygia guttata, Xenopus laevis, Xenopus tropicalis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno477804_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000001, mir-17

          Mature miRNA Sequence:

          CAAAGUGCUCAUAGUGCAGGUAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-20b-5p MIMAT0001413
          Mouse mmu-miR-20b-5p MIMAT0003187
          Rat rno-miR-20b-5p MIMAT0003211
          Callithrix jacchus cja-miR-20b MIMAT0039389
          Chicken gga-miR-20b-5p MIMAT0001411
          Columba livia cli-miR-20b-5p MIMAT0038434
          View More Rows
          Cricetulus griseus cgr-miR-20b MIMAT0041172
          Dasypus novemcinctus dno-miR-20b-5p MIMAT0047522
          Gorilla gorilla ggo-miR-20b MIMAT0024227
          Horse eca-miR-20b MIMAT0013202
          Monodelphis domestica mdo-miR-20b-5p MIMAT0028764
          Ornithorhynchus anatinus oan-miR-20b-5p MIMAT0006853
          Pan troglodytes ptr-miR-20b MIMAT0008069
          Pongo pygmaeus ppy-miR-20b MIMAT0015732
          Pteropus alecto pal-miR-20b-5p MIMAT0040005
          Python bivittatus pbv-miR-20b-5p MIMAT0038847
          Rhesus monkey mml-miR-20b-5p MIMAT0006164
          Taeniopygia guttata tgu-miR-20b-5p MIMAT0014591
          Xenopus laevis xla-miR-20 MIMAT0001348
          Xenopus tropicalis xtr-miR-20b MIMAT0003707

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-20b
          Stem-loop Accession # MI0001519
          Stem-loop Sequence
          AGUACCAAAGUGCUCAUAGUGCAGGUAGUUUUGGCAUGACUCUACUGUAGUAUGGGCACUUCCAGUACU
          Chromosome Location Chr. X - 134169809 - 134169877 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0001413
          miRBase ID: hsa-miR-20b-5p
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: Chr. X - 134169809 - 134169877 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-20b
          Stem-loop Accession # MI0003536
          Stem-loop Sequence
          CCUAGUAGUGCCAAAGUGCUCAUAGUGCAGGUAGUUUUUAUACCACUCUACUGCAGUGUGAGCACUUCUAGUACUCCUGG
          Chromosome Location Chr. X - 52742113 - 52742192 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003187
          miRBase ID: mmu-miR-20b-5p
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: Chr. X - 52742113 - 52742192 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-20b
          Stem-loop Accession # MI0003554
          Stem-loop Sequence
          GUAGUGCCAAAGUGCUCAUAGUGCAGGUAGGUUUUGCUGCACUCUACUGCAGUGUGAGCACUUCUGGUACUC
          Chromosome Location Chr. X - 140117518 - 140117589 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0003211
          miRBase ID: rno-miR-20b-5p
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: Chr. X - 140117518 - 140117589 [-] on Build Rnor_6.0

          Callithrix jacchus

          Stem-loop ID cja-mir-20b
          Stem-loop Accession # MI0031908
          Stem-loop Sequence
          GUGGAUAAGAUUGGGUCCUAGUAGUACCAAAGUGCUCAUAGUGCAGGUAGUUUUGGCAUGACUCUACUGUAGUGUGGGCACUUCUAGUACUCUUGGAUAACAAAUCUCUUGUUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039389
          miRBase ID: cja-miR-20b
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-20b
          Stem-loop Accession # MI0001517
          Stem-loop Sequence
          CUAGCAGUAUCAAAGUGCUCAUAGUGCAGGUAGCUUGGCAUUGGACCUACUGUAAUGUGGGCACUUACAGUACUGUUAGAUAAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001411
          miRBase ID: gga-miR-20b-5p
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-20b
          Stem-loop Accession # MI0029919
          Stem-loop Sequence
          UAGUUUUGUCCUAGCAGUAUCAAAGUGCUCAUAGUGCAGGUAGCUUUGAAUUGAACCUACUGUAAUGUGGGCACUUAUAGUACUGCUAGAUAAAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038434
          miRBase ID: cli-miR-20b-5p
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-20b
          Stem-loop Accession # MI0033102
          Stem-loop Sequence
          GUAGUACCAAAGUGCUCAUAGUGCAGGUAGUUUUGCAUCAUUCUACUGCACUGUGAGCACUUCCAGUACUCUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041172
          miRBase ID: cgr-miR-20b
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-20b
          Stem-loop Accession # MI0038916
          Stem-loop Sequence
          CAAAGUGCUCAUAGUGCAGGUAGUUUUGGCAUUACUCUACUGUAGUGUGGGCACUUUAAGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047522
          miRBase ID: dno-miR-20b-5p
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-20b
          Stem-loop Accession # MI0020774
          Stem-loop Sequence
          AGAUUGGGUCCUAGUAGUACCAAAGUGCUCAUAGUGCAGGUAGUUUUGGCAUGACUCUACUGUAAUAUGGGCACUUCCAGUACUCUUGGAUAACAAAUCUCUUGUUGAUGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024227
          miRBase ID: ggo-miR-20b
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-20b
          Stem-loop Accession # MI0012950
          Stem-loop Sequence
          AGUACCAAAGUGCUCAUAGUGCAGGUAGUUUUGGCAUUACUCUACUGUAGUGUGGGCACUUCCAGUACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013202
          miRBase ID: eca-miR-20b
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-20b
          Stem-loop Accession # MI0023938
          Stem-loop Sequence
          CAAAGUGCUCAUAGUGCAGGUAGUUUUUUGCAAAUGACUACUGUACUAUGGGCACUUCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0028764
          miRBase ID: mdo-miR-20b-5p
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-20b
          Stem-loop Accession # MI0006695
          Stem-loop Sequence
          UGGAAAUCGUAUCCUGACAGUACCAAAGUGCUCAUAGUGCAGGUAGUUUUUUUCAAUUGAUUCUACUGUAAUGUGGGCACUUACAGUACUCCAGGAUAAAGUGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006853
          miRBase ID: oan-miR-20b-5p
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-20b
          Stem-loop Accession # MI0008579
          Stem-loop Sequence
          AGUACCAAAGUGCUCAUAGUGCAGGUAGUUUUGGCAUGACUCUACUGUAGUAUGGGCACUUCCAGUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008069
          miRBase ID: ptr-miR-20b
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-20b
          Stem-loop Accession # MI0014797
          Stem-loop Sequence
          AGUACCAAAGUGCUCAUAGUGCAGGUAGUUUUGGCAUGACUCUACUGUAGUAUGGGCACUUCCAGUACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015732
          miRBase ID: ppy-miR-20b
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-20b
          Stem-loop Accession # MI0032444
          Stem-loop Sequence
          CAAAGUGCUCAUAGUGCAGGUAGUUUUGGCAUUACUCUACUGUAGUGUGGGCACUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040005
          miRBase ID: pal-miR-20b-5p
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-20b
          Stem-loop Accession # MI0030167
          Stem-loop Sequence
          GUUUGUCCUGGCAGUGCCAAAGUGCUCAUAGUGCAGGUAGUUGUGGACCCAAAUUCUACUGUAAUGUGGGCACUUCCAGUCCUGCUGGAUCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038847
          miRBase ID: pbv-miR-20b-5p
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-20b
          Stem-loop Accession # MI0007591
          Stem-loop Sequence
          AGUACCAAAGUGCUCAUAGUGCAGGUAGUUUUGGCAUGACUCUACUGUAGUGUGGGCACUUCCAGUACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006164
          miRBase ID: mml-miR-20b-5p
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-20b
          Stem-loop Accession # MI0013819
          Stem-loop Sequence
          CUAGCAGUAUCAAAGUGCUCAUAGUGCAGGUAGCUUGGAAAUGGACCUACUGUAAUGUGGGCACUUACAGUAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014591
          miRBase ID: tgu-miR-20b-5p
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-20
          Stem-loop Accession # MI0001453
          Stem-loop Sequence
          GCAGUUCCAAAGUGCUCAUAGUGCAGGUAGUUGUAUUCAUGUUCUACUGUAAUAUGGGCACUUACAGUACUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001348
          miRBase ID: xla-miR-20
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-20b
          Stem-loop Accession # MI0004961
          Stem-loop Sequence
          GCAGUUCCAAAGUGCUCAUAGUGCAGGUAGUUGUAUUGAUGUUCUACUGUAAUAUGGGCACUUACAGUACUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003707
          miRBase ID: xtr-miR-20b
          Mature miRNA Sequence: CAAAGUGCUCAUAGUGCAGGUAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306673_pri
          TaqMan™ Pri-miRNA Assay : Hs03295977_pri
          TaqMan™ Pri-miRNA Assay : Rn03465594_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10975
          Ambion® Pre-miR™ miRNA Precursor : PM10975
          TaqMan™ MicroRNA Assay : 001014
          mirVana® miRNA inhibitor : MH10975
          mirVana® miRNA mimic : MC10975
          TaqMan™ Advanced miRNA Assay : 477804_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline