Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Proteins & Peptides ›
          • PDGF-BB Proteins

          Gibco

          Human PDGF-BB Recombinant Protein, PeproTech®

          163 References
          View all (4) PDGF-BB proteins

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support

          Cite Human PDGF-BB Recombinant Protein, PeproTech®

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Testing Data (1)
          Human PDGF-BB Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.
          Human PDGF-BB Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Human PDGF-BB Protein (100-14B-1MG) in Bioactivity

          Bioassay analysis of Human PDGF-BB Recombinant Protein, PeproTech® (Product # 100-14B-1MG). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Human PDGF-BB Protein in Bioactivity (Bioactivity)

          Product Details

          100-14B-1MG

          Applications
          Tested Dilution
          Publications

          Control (Ctrl)

          Assay-dependent
          -

          In vitro Assay (In vitro)

          -
          View 137 publications 137 publications

          Miscellaneous PubMed (Misc)

          -
          View 22 publications 22 publications

          Western blot control (WB Ctrl)

          Assay-dependent
          View 1 publication 1 publication

          Bioactivity (Bioactivity)

          Assay-dependent
          -

          Cell stimulation (Cell Stm)

          -
          View 1 publication 1 publication

          Cell induction (Cell Ind)

          -
          View 2 publications 2 publications
          Product Specifications

          Species

          Human

          Published species

          Bovine, Dog, Guinea pig, Human, Mouse, Non-human primate, Pig, Rat, Sheep

          Expression System

          E. coli

          Amino acid sequence

          SLGSLTIAEP AMIAECKTRT EVFEISRRLI DRTNANFLVW PPCVEVQRCS GCCNNRNVQC RPTQVQLRPV QVRKIEIVRK KPIFKKATVT LEDHLACKCE TVAAARPVT

          Molecular weight

          24.3 kDa

          Class

          Recombinant

          Type

          Protein

          Purity

          ≥ 98% by SDS-PAGE gel and HPLC analyses.

          Endotoxin concentration

          <1 EU/µg

          Activity

          Determined by the dose-dependent stimulation of the proliferation of Balb/c 3T3 cells.The expected ED50 for this effect is 1.0-3.0 ng/ml.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Amount

          1 mg

          Purification

          purified

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          Product Specific Information

          Recombinant Human PDGF-BB is a 24.3 kDa disulfide-linked homodimer of two beta chains (218 total amino acids).

          This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

          Target Information

          The protein encoded by this gene is a member of the platelet-derived growth factor family. The four members of this family are mitogenic factors for cells of mesenchymal origin and are characterized by a motif of eight cysteines. This gene product can exist either as a homodimer (PDGF-BB) or as a heterodimer with the platelet-derived growth factor alpha polypeptide (PDGF-AB), where the dimers are connected by disulfide bonds. Mutations in this gene are associated with meningioma. Reciprocal translocations between chromosomes 22 and 7, at sites where this gene and that for COL1A1 are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans resulting from unregulated expression of growth factor. Two alternatively spliced transcript variants encoding different isoforms have been identified for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: becaplermin; epididymis secretory sperm binding protein; FLJ12858; H-PDGF-BB; orf 2; PDGF subunit B; PDGF, B chain; PDGF-2; PDGFB; platelet-derived growth factor 2; Platelet-derived growth factor B chain; Platelet-derived growth factor beta polypeptide; platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis) oncogene homolog); Platelet-derived growth factor subunit B; platelet-derived growth factor, beta polypeptide (oncogene SIS); Proto-oncogene c-Sis; unnamed protein product

          View more View less

          Gene Aliases: c-sis; IBGC5; PDGF-2; PDGF2; PDGFB; SIS; SSV

          View more View less

          UniProt ID: (Human) P01127

          View more View less

          Entrez Gene ID: (Human) 5155

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline