Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • NEK6 Antibodies

          Proteintech

          NEK6 Polyclonal Antibody

          2 References
          View all (23) NEK6 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite NEK6 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (8)
          NEK6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NEK6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NEK6 Antibody (10378-1-AP) in ICC/IF

          Immunofluorescent analysis of ( -20°C Ethanol ) fixed COS-7 cells using 10378-1-AP (NEK6 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NEK6 Antibody in Immunocytochemistry (ICC/IF)
          NEK6 Antibody in Immunocytochemistry (ICC/IF)
          NEK6 Antibody in Western Blot (WB)
          NEK6 Antibody in Western Blot (WB)
          NEK6 Antibody in Western Blot (WB)
          NEK6 Antibody in Western Blot (WB)
          NEK6 Antibody in Western Blot (WB)
          NEK6 Antibody in Western Blot (WB)
          NEK6 Polyclonal Antibody

          Product Details

          10378-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          NEK6 Fusion Protein Ag0266 (6-209 aa encoded by BC000101)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.1 mg/mL

          Amount

          15 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: GHMPHGGSS NNLCHTLGPV HPPDPQRHPN TLSFRCSLAD FQIEKKIGRG QFSEVYKATC LLDRKTVALK KVQIFEMMDA KARQDCVKEI GLLKQLNHPN IIKYLDSFIE DNELNIVLEL ADAGDLSQMI KYFKKQKRLI PERTVWKYFV QLCSAVEHMH SRRVMHRDIK PANVFITATG VVKLGDLGLG RFFSSETTAA HSLVG (6-209 aa encoded by BC000101)

          Target Information

          NEK6 kinase is a member of the NEK family. NEK6 is a serine/threonine kinase that controls the initiation of mitosis and is required for the metaphase-anaphase transition. The Aspergillus nidulans 'never in mitosis A' (NIMA) gene encodes a serine/threonine kinase that controls initiation of mitosis. NIMA-related kinases (NEKs) are a group of protein kinases that are homologous to NIMA. Evidence suggests that NEKs perform functions similar to those of NIMA. NEK6 encodes a NEK type protein kinase, which contains a C-terminal NIMA-like catalytic domain.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: NEK6; Never in mitosis A-related kinase 6; NIMA (never in mitosis gene a)-related expressed kinase 6; NIMA (never in mitosis gene a)-related kinase 6; NIMA-related expressed kinase 6; NimA-related protein kinase 6; Protein kinase SID6-1512; putative serine-threonine protein kinase; RP11-101K10.6; Serine/threonine-protein kinase Nek6; SID; unnamed protein product

          View more View less

          Gene Aliases: 1300007C09Rik; NEK6; SID6-1512

          View more View less

          UniProt ID: (Human) Q9HC98, (Mouse) Q9ES70, (Rat) P59895

          View more View less

          Entrez Gene ID: (Human) 10783, (Mouse) 59126, (Rat) 360161

          View more View less

          Function(s)
          nucleotide binding magnesium ion binding transcription corepressor binding protein kinase activity protein serine/threonine kinase activity protein binding ATP binding kinase activity transferase activity kinesin binding protein kinase binding ubiquitin protein ligase binding metal ion binding protein serine kinase activity DNA-binding transcription factor binding signal transducer activity transferase activity, transferring phosphorus-containing groups non-receptor serine/threonine protein kinase
          Process(es)
          protein phosphorylation apoptotic process mitotic spindle organization chromosome segregation mitotic nuclear envelope disassembly regulation of mitotic cell cycle peptidyl-serine phosphorylation regulation of mitotic metaphase/anaphase transition regulation of chromosome organization positive regulation of I-kappaB kinase/NF-kappaB signaling protein autophosphorylation spindle assembly cell division regulation of cellular senescence cell cycle mitotic nuclear division signal transduction regulation of signal transduction phosphorylation G2 DNA damage checkpoint
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline