Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MQTPVNIPV PVLRLPRGPD GFSRGFAPDG RRAPLRPEVP EIQECPIAQE SLESQEQRAR AALRERYLRS LLAMVGHQVS FTLHEGVRVA AHFGATDLDV ANFYVSQLQT PIGVQAEALL RCSDIISYTF KP (1-131 aa encoded by BC007793)
The protein encoded by this gene is a component of the core SMN complex, which is required for pre-mRNA splicing in the nucleus. The encoded protein is found in the nucleoplasm, in nuclear "gems" (Gemini of Cajal bodies), and in the cytoplasm. Three transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gem-associated protein 7; Gemin-7; GEMIN7; KIAA2024; SIP3; unnamed protein product; ZNF650
Gene Aliases: 2400008I04Rik; AI120175; AI461688; GEMIN7; SIP3
UniProt ID: (Human) Q9H840, (Mouse) Q9CWY4
Entrez Gene ID: (Human) 79760, (Mouse) 69731, (Rat) 499391
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support