Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • AdoHcyase 2 Antibodies

          Proteintech

          AHCYL1 Polyclonal Antibody

          22 References
          View all (10) AdoHcyase 2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite AHCYL1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (9)
          AHCYL1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          AHCYL1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          AHCYL1 Antibody (10658-3-AP) in ICC/IF

          Immunofluorescent analysis of Hela cells, using AHCYL1 antibody 10658-3-AP at 1:25 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          AHCYL1 Antibody in Immunocytochemistry (ICC/IF)
          AHCYL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AHCYL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AHCYL1 Antibody in Western Blot (WB)
          AHCYL1 Antibody in Western Blot (WB)
          AHCYL1 Antibody in Western Blot (WB)
          AHCYL1 Antibody in Western Blot (WB)
          AHCYL1 Antibody in Western Blot (WB)
          AHCYL1 Antibody in Immunoprecipitation (IP)
          AHCYL1 Polyclonal Antibody

          Product Details

          10658-3-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 19 publications 19 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:400
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Chicken, Mouse, Pig, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          AHCYL1 Fusion Protein Ag1017 (212-530 aa encoded by BC007576)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.27 mg/mL

          Amount

          40.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: IDRCVNMDG WQANMILDDG GDLTHWVYKK YPNVFKKIRG IVEESVTGVH RLYQLSKAGK LCVPAMNVND SVTKQKFDNL YCCRESILDG LKRTTDVMFG GKQVVVCGYG EVGKGCCAAL KALGAIVYIT EIDPICALQA CMDGFRVVKL NEVIRQVDVV ITCTGNKNVV TREHLDRMKN SCIVCNMGHS NTEIDVTSLR TPELTWERVR SQVDHVIWPD GKRVVLLAEG RLLNLSCSTV PTFVLSITAT TQALALIELY NAPEGRYKQD VYLLPKKMDE YVASLHLPSF DAHLTELTDD QAKYLGLNKN GPFKPNYYRY (212-530 aa encoded by BC007576)

          Target Information

          The protein encoded by this gene interacts with inositol 1,4,5-trisphosphate receptor, type 1 and may be involved in the conversion of S-adenosyl-L-homocysteine to L-homocysteine and adenosine. Several transcript variants encoding two different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: adenosylhomocysteinase 2; AdoHcyase 2; DC-expressed AHCY-like molecule; DCAL; dendritic cell expressed AHCY-like protein; dendritic cell expressed AHCY-like protein; epididymis secretory sperm binding protein; inositol 1,4,5-trisphosphate receptor-binding protein; IP(3)Rs binding protein released with IP(3); IP3R binding protein released with inositol 1,4,5-trisphosphate; IP3R-binding protein released with inositol 1,4,5-trisphosphate; IRBIT; protein phosphatase 1, regulatory subunit 78; Putative adenosylhomocysteinase 2; s adenosylhomocysteine hydrolase; S-adenosyl homocysteine hydrolase homolog; S-adenosyl-L-homocysteine hydrolase 2; S-adenosylhomocysteine hydrolase, related sequence 3; S-adenosylhomocysteine hydrolase-like 1; S-adenosylhomocysteine hydrolase-like protein 1; similar to S-adenosylhomocysteine hydrolase; unnamed protein product

          View more View less

          Gene Aliases: 1110034F20Rik; AA409031; AA414901; Ahcy-rs3; AHCYL1; DCAL; IRBIT; PPP1R78; PRO0233; XPVKONA

          View more View less

          UniProt ID: (Human) O43865, (Rat) B5DFN2, (Mouse) Q80SW1

          View more View less

          Entrez Gene ID: (Human) 10768, (Rat) 362013, (Mouse) 229709

          View more View less

          Function(s)
          RNA binding adenosylhomocysteinase activity protein binding enzyme regulator activity identical protein binding hydrolase activity hydrolase
          Process(es)
          protein export from nucleus one-carbon metabolic process apoptotic process positive regulation of sodium ion transport regulation of mRNA 3'-end processing regulation of ion transmembrane transporter activity S-adenosylmethionine cycle angiotensin-activated signaling pathway epithelial fluid transport regulation of anion transport response to calcium ion mitochondrion-ER tethering mRNA polyadenylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline