Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This is a rabbit polyclonal antibody raised against near the C-terminus of human SND1.
Immunogen sequence: TCLITFLLA GIECPRGARN LPGLVQEGEP FSEEATLFTK ELVLQREVEV EVESMDKAGN FIGWLHIDGA NLSVLLVEHA LSKVHFTAER SSYYKSLLSA EEAAKQKKEK VWAHYEEQPV EEVMPVLEEK ERSASYKPVF VTEITDDLHF YVQDVETGTQ LEKLMENMRN DIASHPPVEG SYAPRRGEFC IAKFVDGEWY RARVEKVESP AKIHVFYIDY GNREVLPSTR LGTLSPAFST RVLPAQATEY AFAFIQVPQD DDARTDAVDS VVRDIQNTQC LLNVEHLSAG CPHVTLQFAD SKGDVGLGLV KEGLVMVEVR KEKQFQKVIT EYLNAQESAK SARLNLWRYG DFRADDADEF GYSR (548-911 aa encoded by B C017180)
SND1/P100 (staphylococcal nuclease and tudor domain containing 1), also known as TudorSN, functions in the Pim-1 regulation of Myb activity and acts as a transcriptional activator of EBNA-2. It also interacts with EAV, NSP1, GTF2E1 and GTF2E2, and forms a ternary complex with Stat6 and POLR2A. The staphylococcal nuclease-like (SN)-domains directly interact with amino acids 1099-1758 of CBP. SND1/P100 plays an important role in the assembly of Stat6 transcriptome and stimulates IL-4-dependent transcription by mediating interaction between Stat6 and CBP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 100 kDa coactivator; EBNA2 coactivator p100; p100 co-activator; Staphylococcal nuclease domain-containing protein 1; testis tissue sperm-binding protein Li 82P; Tudor domain-containing protein 11; TudorSN; unnamed protein product
Gene Aliases: p100; SND1; TDRD11; TSN; Tudor-SN
UniProt ID: (Human) Q7KZF4
Entrez Gene ID: (Human) 27044
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support