Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • LDLR Antibodies

          Proteintech

          LDLR Polyclonal Antibody

          93 References
          View all (44) LDLR antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite LDLR Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (21)
          LDLR Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          LDLR Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 21

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          LDLR Antibody (10785-1-AP) in ICC/IF

          Immunofluorescent analysis of (-20°C Ethanol) fixed HeLa cells using LDLR antibody (Product # 10785-1-AP) at dilution of 1:400 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          LDLR Antibody in Immunocytochemistry (ICC/IF)
          LDLR Antibody in Immunocytochemistry (ICC/IF)
          LDLR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LDLR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LDLR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LDLR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LDLR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LDLR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LDLR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LDLR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LDLR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LDLR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          LDLR Antibody in Western Blot (WB)
          LDLR Antibody in Western Blot (WB)
          LDLR Antibody in Western Blot (WB)
          LDLR Antibody in Western Blot (WB)
          LDLR Antibody in Western Blot (WB)
          LDLR Antibody in Western Blot (WB)
          LDLR Antibody in Western Blot (WB)
          LDLR Antibody in Flow Cytometry (Flow)
          LDLR Antibody in Immunoprecipitation (IP)
          LDLR Polyclonal Antibody

          Product Details

          10785-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 75 publications 75 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 7 publications 7 publications

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cells
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications

          Not applicable (N/A)

          -
          View 4 publications 4 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Chicken, Hamster, Human, Mouse, Pig, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          LDLR Fusion Protein Ag1236 (1-350 aa encoded by BC014514)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.31 mg/mL

          Amount

          46.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MGPWGWKLR WTVALLLAAA GTAVGDRCER NEFQCQDGKC ISYKWVCDGS AECQDGSDES QETCLSVTCK SGDFSCGGRV NRCIPQFWRC DGQVDCDNGS DEQGCPPKTC SQDEFRCHDG KCISRQFVCD SDRDCLDGSD EASCPVLTCG PASFQCNSST CIPQLWACDN DPDCEDGSDE WPQRCRGLYV FQGDSSPCSA FEFHCLSGEC IHSSWRCDGG PDCKDKSDEE NCAVATCRPD EFQCSDGNCI HGSRQCDREY DCKDMSDEVG CVNVTLCEGP NKFKCHSGEC ITLDKVCNMA RDCRDWSDEP IKECGTNECL DNNGGCSHVC NDLKIGYECL CPDGFQLVAQ R (1-350 aa encoded by BC014514)

          Target Information

          The low density lipoprotein receptor (LDL-R) gene family consists of cell surface proteins involved in receptor-mediated endocytosis of specific ligands. Low density lipoprotein (LDL) is normally bound at the cell membrane and taken into the cell ending up in lysosomes where the protein is degraded and the cholesterol is made available for repression of microsomal enzyme 3-hydroxy-3-methylglutaryl coenzyme A (HMG CoA) reductase, the rate-limiting step in cholesterol synthesis. At the same time, a reciprocal stimulation of cholesterol ester synthesis takes place. Mutations in the LDL-R gene cause the autosomal dominant disorder, familial hypercholesterolemia. Along with SCARB1, CLDN1, and the tetraspanin superfamily member CD81, LDL-R has been reported to be an entry factor for the Hepatitis C virus. At least three isoforms of LDL-R are known to exist.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: LDL receptor; ldl-r; Low-density lipoprotein receptor; low-density lipoprotein receptor class A domain-containing protein 3; unnamed protein product

          View more View less

          Gene Aliases: FH; FHC; FHCL1; Hlb301; LDLCQ2; LDLR

          View more View less

          UniProt ID: (Human) P01130, (Mouse) P35951

          View more View less

          Entrez Gene ID: (Human) 3949, (Mouse) 16835

          View more View less

          Function(s)
          beta-amyloid binding virus receptor activity protease binding low-density lipoprotein receptor activity calcium ion binding protein binding low-density lipoprotein particle binding very-low-density lipoprotein particle receptor activity clathrin heavy chain binding identical protein binding binding, bridging lipoprotein particle binding glycoprotein binding apolipoprotein
          Process(es)
          retinoid metabolic process negative regulation of receptor recycling lipid metabolic process lipid transport endocytosis receptor-mediated endocytosis phagocytosis long-term memory steroid metabolic process cholesterol metabolic process negative regulation of macromolecule metabolic process positive regulation of gene expression negative regulation of gene expression positive regulation of triglyceride biosynthetic process regulation of phosphatidylcholine catabolic process negative regulation of low-density lipoprotein particle clearance phospholipid transport intestinal cholesterol absorption cholesterol transport plasma lipoprotein particle clearance low-density lipoprotein particle clearance high-density lipoprotein particle clearance lipoprotein metabolic process lipoprotein catabolic process cholesterol homeostasis viral entry into host cell artery morphogenesis positive regulation of inflammatory response negative regulation of multicellular organismal process regulation of protein metabolic process negative regulation of protein metabolic process establishment of localization in cell artery development response to caloric restriction negative regulation of astrocyte activation cholesterol import cellular response to fatty acid cellular response to low-density lipoprotein particle stimulus receptor-mediated endocytosis of low-density lipoprotein particle involved in cholesterol transport regulation of cholesterol metabolic process beta-amyloid clearance amyloid-beta clearance by cellular catabolic process negative regulation of microglial cell activation positive regulation of lysosomal protein catabolic process negative regulation of amyloid fibril formation transport regulation of cholesterol homeostasis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline