Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • ANP32B Antibodies

          Proteintech

          ANP32B Polyclonal Antibody

          12 References
          View all (12) ANP32B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ANP32B Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (14)
          ANP32B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ANP32B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 14

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ANP32B Antibody (10843-1-AP) in ICC/IF

          Immunofluorescent analysis of HeLa cells using 10843-1-AP (ANP32B antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ANP32B Antibody in Immunocytochemistry (ICC/IF)
          ANP32B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ANP32B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ANP32B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ANP32B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ANP32B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ANP32B Antibody in Western Blot (WB)
          ANP32B Antibody in Western Blot (WB)
          ANP32B Antibody in Western Blot (WB)
          ANP32B Antibody in Western Blot (WB)
          ANP32B Antibody in Western Blot (WB)
          ANP32B Antibody in Western Blot (WB)
          ANP32B Antibody in Western Blot (WB)
          ANP32B Antibody in Immunoprecipitation (IP)
          ANP32B Polyclonal Antibody

          Product Details

          10843-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2400
          View 9 publications 9 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication

          Not applicable (N/A)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ANP32B Fusion Protein Ag1274 (1-251 aa encoded by BC013003)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.13 mg/mL

          Amount

          19.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MDMKRRIHL ELRNRTPAAV RELVLDNCKS NDGKIEGLTA EFVNLEFLSL INVGLISVSN LPKLPKLKKL ELSENRIFGG LDMLAEKLPN LTHLNLSGNK LKDISTLEPL KKLECLKSLD LFNCEVTNLN DYRESVFKLL PQLTYLDGYD REDQEAPDSD AEVDGVDEEE EDEEGEDEED EDDEDGEEEE FDEEDDEDED VEGDEDDDEV SEEEEEFGLD EEDEDEDEDE EEEEGGKGEK RKRETDDEGE DD (1-251 aa encoded by BC013003)

          Target Information

          Multifunctional protein working as a cell cycle progression factor as well as a cell survival factor. Required for the progression from the G1 to the S phase. Anti-apoptotic protein which functions as a caspase-3 inhibitor. Has no phosphatase 2A (PP2A) inhibitor activity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: acidic (leucine-rich) nuclear phosphoprotein 32 family, member B; Acidic leucine-rich nuclear phosphoprotein 32 family member B; acidic nuclear phosphoprotein 32 family, member B; Acidic protein rich in leucines; OTTHUMP00000063770; PHAPI2; proliferation related acidic leucine rich protein PAL31; Proliferation-related acidic leucine-rich protein PAL31; Putative HLA-DR-associated protein I-2; Silver-stainable protein SSP29; TNF superfamily member 13; unnamed protein product

          View more View less

          Gene Aliases: 2410015B15Rik; ANP32B; APRIL; PAL31; PHAPI2; PHAPI2a; SSP29

          View more View less

          UniProt ID: (Human) Q92688, (Mouse) Q9EST5

          View more View less

          Entrez Gene ID: (Human) 10541, (Mouse) 67628

          View more View less

          Function(s)
          protein binding histone binding RNA polymerase binding chromatin/chromatin-binding, or -regulatory protein
          Process(es)
          vasculature development nucleosome assembly ventricular system development regulation of apoptotic process negative regulation of apoptotic process negative regulation of cell differentiation positive regulation of protein export from nucleus inner ear development palate development G1/S transition of mitotic cell cycle positive regulation of cell proliferation positive regulation of G1/S transition of mitotic cell cycle
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline